Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1746199-1746458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN1
Entrez Gene ID
392617
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
PPP1R28
UniGene
Hs.42896
Gene Chromosome Location
Chr.7: 1666075-1747954 on Build GRCh38
Transcripts
NM_001128636.2 XM_006715725.3 XM_006715726.2 XM_006715727.3 XM_011515397.2 XM_011515398.2 XM_011515401.2 XM_017012203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4804723 c.1824G>T p.K608N Chr.7: 1746420-1746420 on Build GRCh38
5416160 c.1607G>T p.R536L Chr.7: 1746203-1746203 on Build GRCh38
6216641 c.1787C>T p.P596L Chr.7: 1746383-1746383 on Build GRCh38
5761208 c.1725G>A p.A575A Chr.7: 1746321-1746321 on Build GRCh38
6514670 c.1628G>A p.R543Q Chr.7: 1746224-1746224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993029400 Chr.7: 1746413-1746413 on Build GRCh38
rs772138706 Chr.7: 1746321-1746321 on Build GRCh38
rs747147896 Chr.7: 1746420-1746420 on Build GRCh38
rs767496054 Chr.7: 1746235-1746235 on Build GRCh38
rs973943329 Chr.7: 1746439-1746439 on Build GRCh38
rs781682953 Chr.7: 1746220-1746220 on Build GRCh38
rs960056597 Chr.7: 1746412-1746412 on Build GRCh38
rs759414426 Chr.7: 1746248-1746248 on Build GRCh38
rs751185702 Chr.7: 1746352-1746352 on Build GRCh38
rs776023219 Chr.7: 1746227-1746227 on Build GRCh38
rs763795538 Chr.7: 1746267-1746267 on Build GRCh38
rs780538635 Chr.7: 1746315-1746315 on Build GRCh38
rs543694392 Chr.7: 1746366-1746366 on Build GRCh38
rs769154454 Chr.7: 1746318-1746318 on Build GRCh38
rs775250378 Chr.7: 1746342-1746342 on Build GRCh38
rs574178999 Chr.7: 1746359-1746359 on Build GRCh38
rs764994517 Chr.7: 1746249-1746249 on Build GRCh38
rs981115372 Chr.7: 1746428-1746428 on Build GRCh38
rs772222289 Chr.7: 1746403-1746403 on Build GRCh38
rs758039912 Chr.7: 1746256-1746256 on Build GRCh38
rs780598626 Chr.7: 1746281-1746281 on Build GRCh38
rs767035701 Chr.7: 1746360-1746360 on Build GRCh38
rs776588093 Chr.7: 1746242-1746242 on Build GRCh38
rs749945154 Chr.7: 1746363-1746363 on Build GRCh38
rs777903379 Chr.7: 1746415-1746415 on Build GRCh38
rs748376249 Chr.7: 1746397-1746397 on Build GRCh38
rs763883088 Chr.7: 1746351-1746351 on Build GRCh38
rs1010455764 Chr.7: 1746402-1746402 on Build GRCh38
rs756687874 Chr.7: 1746367-1746367 on Build GRCh38
rs774563944 Chr.7: 1746319-1746319 on Build GRCh38
rs754398185 Chr.7: 1746375-1746375 on Build GRCh38
rs780767565 Chr.7: 1746373-1746373 on Build GRCh38
rs756738684 Chr.7: 1746308-1746308 on Build GRCh38
rs1020552175 Chr.7: 1746408-1746408 on Build GRCh38
rs1007410261 Chr.7: 1746377-1746377 on Build GRCh38
rs1033180867 Chr.7: 1746390-1746390 on Build GRCh38
rs771331072 Chr.7: 1746218-1746218 on Build GRCh38
rs769042444 Chr.7: 1746230-1746230 on Build GRCh38
rs746366286 Chr.7: 1746223-1746223 on Build GRCh38
rs762680751 Chr.7: 1746350-1746350 on Build GRCh38
rs749503062 Chr.7: 1746228-1746228 on Build GRCh38
rs755351530 Chr.7: 1746376-1746376 on Build GRCh38
rs752527358 Chr.7: 1746255-1746255 on Build GRCh38
rs765765593 Chr.7: 1746316-1746316 on Build GRCh38
rs774293588 Chr.7: 1746234-1746234 on Build GRCh38
rs759437441 Chr.7: 1746330-1746330 on Build GRCh38
rs115745585 Chr.7: 1746438-1746438 on Build GRCh38
rs779164550 Chr.7: 1746386-1746386 on Build GRCh38
rs893330230 Chr.7: 1746391-1746391 on Build GRCh38
rs554272019 Chr.7: 1746270-1746270 on Build GRCh38
rs1048873887 Chr.7: 1746329-1746329 on Build GRCh38
rs376930464 Chr.7: 1746435-1746435 on Build GRCh38
rs765223697 Chr.7: 1746336-1746336 on Build GRCh38
rs770244441 Chr.7: 1746224-1746224 on Build GRCh38
rs773213925 Chr.7: 1746324-1746324 on Build GRCh38
rs748346698 Chr.7: 1746320-1746320 on Build GRCh38
rs563902266 Chr.7: 1746423-1746423 on Build GRCh38
rs746340818 Chr.7: 1746285-1746285 on Build GRCh38
rs940615071 Chr.7: 1746338-1746338 on Build GRCh38
rs751186375 Chr.7: 1746268-1746268 on Build GRCh38

View Full Product Details