Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74251543-74251802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPME1
Entrez Gene ID
51400
Gene Name
protein phosphatase methylesterase 1
Gene Aliases
PME-1
UniGene
Hs.503251
Gene Chromosome Location
Chr.11: 74170891-74254703 on Build GRCh38
Transcripts
NM_001271593.1 NM_016147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5492839 c.1075-10C>T p.? Chr.11: 74251638-74251638 on Build GRCh38
1242703 c.1106G>A p.R369Q Chr.11: 74251679-74251679 on Build GRCh38
3769447 c.1105C>T p.R369W Chr.11: 74251678-74251678 on Build GRCh38
3375948 c.1129G>A p.G377S Chr.11: 74251702-74251702 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs982870126 Chr.11: 74251733-74251733 on Build GRCh38
rs777709827 Chr.11: 74251678-74251678 on Build GRCh38
rs776432512 Chr.11: 74251754-74251754 on Build GRCh38
rs988068271 Chr.11: 74251571-74251571 on Build GRCh38
rs752814403 Chr.11: 74251637-74251637 on Build GRCh38
rs769488981 Chr.11: 74251600-74251600 on Build GRCh38
rs760541808 Chr.11: 74251740-74251740 on Build GRCh38
rs775253017 Chr.11: 74251708-74251708 on Build GRCh38
rs774962989 Chr.11: 74251601-74251601 on Build GRCh38
rs755800681 Chr.11: 74251633-74251633 on Build GRCh38
rs753029665 Chr.11: 74251755-74251755 on Build GRCh38
rs779228861 Chr.11: 74251635-74251635 on Build GRCh38
rs376787845 Chr.11: 74251723-74251723 on Build GRCh38
rs369949939 Chr.11: 74251692-74251692 on Build GRCh38
rs768282830 Chr.11: 74251718-74251718 on Build GRCh38
rs746262607 Chr.11: 74251701-74251701 on Build GRCh38
rs532685612 Chr.11: 74251619-74251619 on Build GRCh38
rs771069880 Chr.11: 74251772-74251772 on Build GRCh38
rs767449324 Chr.11: 74251724-74251724 on Build GRCh38
rs369171590 Chr.11: 74251598-74251598 on Build GRCh38
rs148795025 Chr.11: 74251605-74251605 on Build GRCh38
rs770001714 Chr.11: 74251702-74251702 on Build GRCh38
rs762486131 Chr.11: 74251711-74251711 on Build GRCh38
rs768408819 Chr.11: 74251738-74251739 on Build GRCh38
rs1036561036 Chr.11: 74251730-74251730 on Build GRCh38
rs750321437 Chr.11: 74251731-74251731 on Build GRCh38
rs373092083 Chr.11: 74251757-74251757 on Build GRCh38
rs201992998 Chr.11: 74251631-74251631 on Build GRCh38
rs530079542 Chr.11: 74251572-74251572 on Build GRCh38
rs373808614 Chr.11: 74251699-74251699 on Build GRCh38
rs751692293 Chr.11: 74251760-74251760 on Build GRCh38
rs926386340 Chr.11: 74251629-74251629 on Build GRCh38
rs566850148 Chr.11: 74251611-74251611 on Build GRCh38
rs767361449 Chr.11: 74251630-74251630 on Build GRCh38
rs764352439 Chr.11: 74251758-74251758 on Build GRCh38
rs965927023 Chr.11: 74251569-74251569 on Build GRCh38
rs773529715 Chr.11: 74251728-74251728 on Build GRCh38
rs762542606 Chr.11: 74251603-74251603 on Build GRCh38
rs758544967 Chr.11: 74251664-74251664 on Build GRCh38
rs770020360 Chr.11: 74251721-74251721 on Build GRCh38
rs371290758 Chr.11: 74251743-74251743 on Build GRCh38
rs747576476 Chr.11: 74251687-74251687 on Build GRCh38

View Full Product Details