Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 144114654-144114915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GYPA
Entrez Gene ID
2993
Gene Name
glycophorin A (MNS blood group)
Gene Aliases
CD235a, GPA, GPErik, GPSAT, HGpMiV, HGpMiXI, HGpSta(C), MN, MNS, PAS-2
UniGene
Hs.434973
Gene Chromosome Location
Chr.4: 144109303-144140854 on Build GRCh38
Transcripts
NM_001308187.1 NM_001308190.1 NM_002099.7 XM_017008134.1 XM_017008135.1 XM_017008136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4158835 c.385C>T p.P129S Chr.4: 144114740-144114740 on Build GRCh38
5532132 c.421G>A p.E141K Chr.4: 144114704-144114704 on Build GRCh38
4158834 c.386C>T p.P129L Chr.4: 144114739-144114739 on Build GRCh38
3600593 c.434C>A p.P145Q Chr.4: 144114691-144114691 on Build GRCh38
7370591 c.419T>C p.V140A Chr.4: 144114706-144114706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144978812 Chr.4: 144114742-144114742 on Build GRCh38
rs961507550 Chr.4: 144114851-144114852 on Build GRCh38
rs868445451 Chr.4: 144114731-144114731 on Build GRCh38
rs750442332 Chr.4: 144114729-144114729 on Build GRCh38
rs531436516 Chr.4: 144114865-144114865 on Build GRCh38
rs139529237 Chr.4: 144114692-144114692 on Build GRCh38
rs1053624327 Chr.4: 144114823-144114823 on Build GRCh38
rs759559849 Chr.4: 144114785-144114785 on Build GRCh38
rs191630121 Chr.4: 144114797-144114797 on Build GRCh38
rs903281704 Chr.4: 144114876-144114876 on Build GRCh38
rs559312206 Chr.4: 144114779-144114779 on Build GRCh38
rs199939598 Chr.4: 144114783-144114783 on Build GRCh38
rs949126171 Chr.4: 144114740-144114740 on Build GRCh38
rs1042581115 Chr.4: 144114868-144114868 on Build GRCh38
rs1003644198 Chr.4: 144114862-144114862 on Build GRCh38
rs1046080674 Chr.4: 144114747-144114747 on Build GRCh38
rs539321054 Chr.4: 144114738-144114738 on Build GRCh38
rs779638603 Chr.4: 144114739-144114739 on Build GRCh38
rs745744808 Chr.4: 144114866-144114866 on Build GRCh38
rs374311530 Chr.4: 144114712-144114712 on Build GRCh38
rs551233980 Chr.4: 144114887-144114887 on Build GRCh38
rs771432727 Chr.4: 144114782-144114782 on Build GRCh38
rs149908820 Chr.4: 144114698-144114698 on Build GRCh38
rs762103857 Chr.4: 144114685-144114685 on Build GRCh38
rs370205131 Chr.4: 144114807-144114807 on Build GRCh38
rs200879866 Chr.4: 144114715-144114715 on Build GRCh38
rs752904561 Chr.4: 144114800-144114800 on Build GRCh38
rs1008709177 Chr.4: 144114886-144114886 on Build GRCh38
rs761614774 Chr.4: 144114694-144114694 on Build GRCh38
rs751730509 Chr.4: 144114706-144114706 on Build GRCh38
rs369389761 Chr.4: 144114722-144114722 on Build GRCh38
rs757151303 Chr.4: 144114723-144114723 on Build GRCh38
rs761685190 Chr.4: 144114802-144114802 on Build GRCh38
rs573646560 Chr.4: 144114804-144114804 on Build GRCh38
rs766507212 Chr.4: 144114705-144114705 on Build GRCh38
rs201228614 Chr.4: 144114781-144114781 on Build GRCh38
rs149086290 Chr.4: 144114748-144114748 on Build GRCh38
rs576735595 Chr.4: 144114702-144114702 on Build GRCh38
rs142191934 Chr.4: 144114762-144114762 on Build GRCh38
rs150143712 Chr.4: 144114861-144114861 on Build GRCh38
rs562322110 Chr.4: 144114864-144114864 on Build GRCh38
rs780735675 Chr.4: 144114754-144114754 on Build GRCh38
rs767742592 Chr.4: 144114795-144114795 on Build GRCh38
rs770196015 Chr.4: 144114774-144114774 on Build GRCh38

View Full Product Details