Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129976531-129976758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRH
Entrez Gene ID
7200
Gene Name
thyrotropin releasing hormone
Gene Aliases
Pro-TRH, TRF
UniGene
Hs.182231
Gene Chromosome Location
Chr.3: 129974393-129977938 on Build GRCh38
Transcripts
NM_007117.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6807738 c.213G>A p.A71A Chr.3: 129976700-129976700 on Build GRCh38
1038149 c.252G>A p.Q84Q Chr.3: 129976739-129976739 on Build GRCh38
4735878 c.268_270delGAG p.E95delE Chr.3: 129976755-129976757 on Build GRCh38
5398135 c.219G>A p.Q73Q Chr.3: 129976706-129976706 on Build GRCh38
6807736 c.252G>T p.Q84H Chr.3: 129976739-129976739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887663209 Chr.3: 129976705-129976705 on Build GRCh38
rs757575199 Chr.3: 129976690-129976690 on Build GRCh38
rs193299790 Chr.3: 129976699-129976699 on Build GRCh38
rs763597431 Chr.3: 129976683-129976683 on Build GRCh38
rs768497875 Chr.3: 129976728-129976728 on Build GRCh38
rs766048666 Chr.3: 129976670-129976670 on Build GRCh38
rs372289257 Chr.3: 129976734-129976734 on Build GRCh38
rs573304521 Chr.3: 129976646-129976646 on Build GRCh38
rs764622593 Chr.3: 129976685-129976685 on Build GRCh38
rs538153308 Chr.3: 129976555-129976555 on Build GRCh38
rs772858139 Chr.3: 129976660-129976660 on Build GRCh38
rs13097335 Chr.3: 129976597-129976597 on Build GRCh38
rs780144608 Chr.3: 129976702-129976702 on Build GRCh38
rs147760555 Chr.3: 129976567-129976567 on Build GRCh38
rs981527636 Chr.3: 129976611-129976611 on Build GRCh38
rs61740106 Chr.3: 129976735-129976735 on Build GRCh38
rs550964427 Chr.3: 129976558-129976558 on Build GRCh38
rs756305246 Chr.3: 129976700-129976700 on Build GRCh38
rs771639796 Chr.3: 129976650-129976650 on Build GRCh38
rs950097522 Chr.3: 129976600-129976600 on Build GRCh38
rs755648187 Chr.3: 129976590-129976590 on Build GRCh38
rs114856424 Chr.3: 129976618-129976618 on Build GRCh38
rs749322206 Chr.3: 129976703-129976703 on Build GRCh38
rs35341042 Chr.3: 129976630-129976631 on Build GRCh38
rs751892697 Chr.3: 129976686-129976686 on Build GRCh38
rs989353876 Chr.3: 129976586-129976586 on Build GRCh38
rs753377630 Chr.3: 129976673-129976673 on Build GRCh38
rs374882305 Chr.3: 129976666-129976666 on Build GRCh38
rs784704 Chr.3: 129976695-129976695 on Build GRCh38
rs1028735708 Chr.3: 129976715-129976715 on Build GRCh38
rs372809068 Chr.3: 129976633-129976633 on Build GRCh38

View Full Product Details