Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24305924-24306161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIDEB
Entrez Gene ID
27141
Gene Name
cell death-inducing DFFA-like effector b
Gene Aliases
-
UniGene
Hs.642693
Gene Chromosome Location
Chr.14: 24305187-24311377 on Build GRCh38
Transcripts
NM_001318807.1 NM_014430.2 XM_011536659.2 XM_017021221.1 NM_001286367.1 NM_174913.2 XM_005267385.1 XM_011536526.1

Gene Symbol
NOP9
Entrez Gene ID
161424
Gene Name
NOP9 nucleolar protein
Gene Aliases
C14orf21
UniGene
Hs.708040
Gene Chromosome Location
Chr.14: 24299854-24309126 on Build GRCh38
Transcripts
NM_001318807.1 NM_014430.2 XM_011536659.2 XM_017021221.1 NM_001286367.1 NM_174913.2 XM_005267385.1 XM_011536526.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5015216 c.512C>T p.P171L Chr.14: 24305962-24305962 on Build GRCh38
5027832 c.424A>G p.N142D Chr.14: 24306050-24306050 on Build GRCh38
3987604 c.477C>G p.L159L Chr.14: 24305997-24305997 on Build GRCh38
1201208 c.463A>G p.T155A Chr.14: 24306011-24306011 on Build GRCh38
5542749 c.387G>A p.K129K Chr.14: 24306087-24306087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773522747 Chr.14: 24306034-24306034 on Build GRCh38
rs768036452 Chr.14: 24306017-24306017 on Build GRCh38
rs144864084 Chr.14: 24305972-24305972 on Build GRCh38
rs771494020 Chr.14: 24305989-24305990 on Build GRCh38
rs139893717 Chr.14: 24306081-24306081 on Build GRCh38
rs913330740 Chr.14: 24306008-24306008 on Build GRCh38
rs774843620 Chr.14: 24306009-24306010 on Build GRCh38
rs768385764 Chr.14: 24306058-24306058 on Build GRCh38
rs200868524 Chr.14: 24306139-24306139 on Build GRCh38
rs147564519 Chr.14: 24306091-24306091 on Build GRCh38
rs144978298 Chr.14: 24306084-24306084 on Build GRCh38
rs780437046 Chr.14: 24305997-24305997 on Build GRCh38
rs748425182 Chr.14: 24306016-24306016 on Build GRCh38
rs1042635167 Chr.14: 24306069-24306069 on Build GRCh38
rs34740317 Chr.14: 24306082-24306082 on Build GRCh38
rs199780940 Chr.14: 24306002-24306002 on Build GRCh38
rs146036691 Chr.14: 24306043-24306043 on Build GRCh38
rs750018394 Chr.14: 24306076-24306076 on Build GRCh38
rs143381849 Chr.14: 24306110-24306110 on Build GRCh38
rs760028727 Chr.14: 24306044-24306046 on Build GRCh38
rs1049339375 Chr.14: 24306045-24306045 on Build GRCh38
rs762371641 Chr.14: 24306077-24306077 on Build GRCh38
rs757875722 Chr.14: 24305994-24305994 on Build GRCh38
rs372847855 Chr.14: 24306012-24306012 on Build GRCh38
rs749965348 Chr.14: 24305986-24305986 on Build GRCh38
rs866809440 Chr.14: 24306011-24306011 on Build GRCh38
rs147369401 Chr.14: 24306080-24306080 on Build GRCh38
rs367645614 Chr.14: 24306003-24306003 on Build GRCh38
rs889659485 Chr.14: 24306109-24306109 on Build GRCh38
rs756491712 Chr.14: 24306087-24306087 on Build GRCh38
rs764766425 Chr.14: 24305964-24305964 on Build GRCh38
rs148553609 Chr.14: 24306062-24306062 on Build GRCh38
rs760648215 Chr.14: 24306057-24306057 on Build GRCh38
rs749630658 Chr.14: 24305954-24305957 on Build GRCh38
rs756881098 Chr.14: 24305951-24305951 on Build GRCh38
rs765905923 Chr.14: 24306079-24306079 on Build GRCh38
rs770979892 Chr.14: 24306044-24306044 on Build GRCh38
rs775358157 Chr.14: 24306048-24306048 on Build GRCh38
rs749522698 Chr.14: 24306095-24306095 on Build GRCh38
rs747480989 Chr.14: 24306000-24306000 on Build GRCh38
rs142940048 Chr.14: 24306063-24306063 on Build GRCh38

View Full Product Details