Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24665688-24665949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCOA1
Entrez Gene ID
8648
Gene Name
nuclear receptor coactivator 1
Gene Aliases
F-SRC-1, KAT13A, RIP160, SRC1, bHLHe42, bHLHe74
UniGene
Hs.596314
Gene Chromosome Location
Chr.2: 24492050-24770702 on Build GRCh38
Transcripts
NM_003743.4 NM_147223.2 NM_147233.2 XM_005264625.1 XM_005264626.1 XM_005264628.1 XM_017005168.1 XM_017005169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3426321 c.197A>C p.K66T Chr.2: 24665856-24665856 on Build GRCh38
5434309 c.180G>C p.L60F Chr.2: 24665839-24665839 on Build GRCh38
5643555 c.164G>A p.S55N Chr.2: 24665823-24665823 on Build GRCh38
3580210 c.224A>T p.D75V Chr.2: 24665883-24665883 on Build GRCh38
6157761 c.113A>G p.Q38R Chr.2: 24665772-24665772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904854328 Chr.2: 24665721-24665721 on Build GRCh38
rs1034875662 Chr.2: 24665745-24665745 on Build GRCh38
rs374878680 Chr.2: 24665799-24665799 on Build GRCh38
rs865928496 Chr.2: 24665873-24665873 on Build GRCh38
rs755639022 Chr.2: 24665905-24665905 on Build GRCh38
rs1003437769 Chr.2: 24665724-24665724 on Build GRCh38
rs748741953 Chr.2: 24665928-24665928 on Build GRCh38
rs767381377 Chr.2: 24665823-24665823 on Build GRCh38
rs376247972 Chr.2: 24665924-24665924 on Build GRCh38
rs748936068 Chr.2: 24665758-24665758 on Build GRCh38
rs185518589 Chr.2: 24665752-24665752 on Build GRCh38
rs748666656 Chr.2: 24665901-24665901 on Build GRCh38
rs770918930 Chr.2: 24665767-24665767 on Build GRCh38
rs781681339 Chr.2: 24665736-24665736 on Build GRCh38
rs749919133 Chr.2: 24665762-24665762 on Build GRCh38
rs758611823 Chr.2: 24665881-24665881 on Build GRCh38
rs750698456 Chr.2: 24665875-24665875 on Build GRCh38
rs149729595 Chr.2: 24665861-24665861 on Build GRCh38
rs775461907 Chr.2: 24665842-24665842 on Build GRCh38
rs769307134 Chr.2: 24665751-24665751 on Build GRCh38
rs376500275 Chr.2: 24665884-24665884 on Build GRCh38
rs751932179 Chr.2: 24665886-24665886 on Build GRCh38
rs747633445 Chr.2: 24665744-24665744 on Build GRCh38
rs200704194 Chr.2: 24665896-24665896 on Build GRCh38
rs765333120 Chr.2: 24665871-24665871 on Build GRCh38
rs759349979 Chr.2: 24665814-24665814 on Build GRCh38

View Full Product Details