Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43753540-43753783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDXK
Entrez Gene ID
8566
Gene Name
pyridoxal (pyridoxine, vitamin B6) kinase
Gene Aliases
C21orf124, C21orf97, HEL-S-1a, PKH, PNK, PRED79
UniGene
Hs.284491
Gene Chromosome Location
Chr.21: 43719097-43762307 on Build GRCh38
Transcripts
NM_003681.4 XM_005261195.2 XM_005261196.3 XM_005261198.2 XM_005261199.2 XM_011529758.2 XM_011529760.2 XM_011529761.1 XM_017028483.1 XM_017028484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3551282 c.630C>G p.P210P Chr.21: 43753590-43753590 on Build GRCh38
6559167 c.759G>T p.K253N Chr.21: 43753719-43753719 on Build GRCh38
1031183 c.685G>A p.V229I Chr.21: 43753645-43753645 on Build GRCh38
3357297 c.659G>A p.R220Q Chr.21: 43753619-43753619 on Build GRCh38
6046983 c.650A>T p.E217V Chr.21: 43753610-43753610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111247841 Chr.21: 43753582-43753582 on Build GRCh38
rs750792358 Chr.21: 43753656-43753656 on Build GRCh38
rs369432283 Chr.21: 43753737-43753737 on Build GRCh38
rs780854378 Chr.21: 43753713-43753713 on Build GRCh38
rs769761064 Chr.21: 43753725-43753725 on Build GRCh38
rs200762097 Chr.21: 43753641-43753641 on Build GRCh38
rs760868476 Chr.21: 43753753-43753753 on Build GRCh38
rs752826014 Chr.21: 43753567-43753567 on Build GRCh38
rs780565048 Chr.21: 43753680-43753680 on Build GRCh38
rs984066690 Chr.21: 43753666-43753666 on Build GRCh38
rs770571042 Chr.21: 43753727-43753728 on Build GRCh38
rs775393291 Chr.21: 43753730-43753730 on Build GRCh38
rs368214047 Chr.21: 43753578-43753578 on Build GRCh38
rs894625127 Chr.21: 43753748-43753748 on Build GRCh38
rs758511237 Chr.21: 43753573-43753573 on Build GRCh38
rs963330330 Chr.21: 43753590-43753590 on Build GRCh38
rs376453520 Chr.21: 43753735-43753735 on Build GRCh38
rs765292715 Chr.21: 43753620-43753620 on Build GRCh38
rs942564020 Chr.21: 43753675-43753675 on Build GRCh38
rs746827852 Chr.21: 43753598-43753617 on Build GRCh38
rs201546479 Chr.21: 43753591-43753591 on Build GRCh38
rs772759626 Chr.21: 43753601-43753601 on Build GRCh38
rs754200347 Chr.21: 43753760-43753760 on Build GRCh38
rs370448964 Chr.21: 43753736-43753736 on Build GRCh38
rs757480516 Chr.21: 43753642-43753642 on Build GRCh38
rs756627812 Chr.21: 43753598-43753598 on Build GRCh38
rs902871109 Chr.21: 43753719-43753719 on Build GRCh38
rs755450554 Chr.21: 43753689-43753689 on Build GRCh38
rs749846062 Chr.21: 43753683-43753683 on Build GRCh38
rs912365069 Chr.21: 43753674-43753674 on Build GRCh38
rs764118392 Chr.21: 43753638-43753638 on Build GRCh38
rs767722136 Chr.21: 43753644-43753644 on Build GRCh38
rs371501001 Chr.21: 43753612-43753612 on Build GRCh38
rs775520000 Chr.21: 43753630-43753630 on Build GRCh38
rs911028893 Chr.21: 43753682-43753682 on Build GRCh38
rs372098146 Chr.21: 43753698-43753698 on Build GRCh38
rs770490767 Chr.21: 43753709-43753709 on Build GRCh38
rs111425602 Chr.21: 43753581-43753581 on Build GRCh38
rs145100052 Chr.21: 43753596-43753596 on Build GRCh38
rs200647993 Chr.21: 43753613-43753613 on Build GRCh38
rs759333796 Chr.21: 43753619-43753619 on Build GRCh38
rs750598827 Chr.21: 43753747-43753747 on Build GRCh38
rs1041367550 Chr.21: 43753714-43753714 on Build GRCh38
rs745893841 Chr.21: 43753587-43753587 on Build GRCh38
rs768626421 Chr.21: 43753732-43753732 on Build GRCh38
rs963890245 Chr.21: 43753684-43753684 on Build GRCh38
rs758463498 Chr.21: 43753731-43753731 on Build GRCh38
rs374293056 Chr.21: 43753575-43753575 on Build GRCh38
rs367927142 Chr.21: 43753603-43753603 on Build GRCh38
rs61737063 Chr.21: 43753631-43753631 on Build GRCh38
rs746467291 Chr.21: 43753702-43753702 on Build GRCh38
rs370764393 Chr.21: 43753665-43753665 on Build GRCh38
rs8127335 Chr.21: 43753599-43753599 on Build GRCh38
rs202036508 Chr.21: 43753600-43753600 on Build GRCh38
rs138572525 Chr.21: 43753716-43753716 on Build GRCh38
rs776301044 Chr.21: 43753618-43753618 on Build GRCh38
rs772689161 Chr.21: 43753681-43753681 on Build GRCh38
rs766772841 Chr.21: 43753759-43753759 on Build GRCh38

View Full Product Details