Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149713101-149713358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEA11
Entrez Gene ID
4110
Gene Name
MAGE family member A11
Gene Aliases
CT1.11, MAGE-11, MAGE11, MAGEA-11
UniGene
Hs.670252
Gene Chromosome Location
Chr.X: 149688202-149717268 on Build GRCh38
Transcripts
NM_001011544.1 NM_005366.4 XM_011531164.2 XM_017029521.1 XM_017029522.1

SNP Detail

SNP ID Chromosome Location
rs782123949 Chr.X: 149713165-149713165 on Build GRCh38
rs782214984 Chr.X: 149713206-149713206 on Build GRCh38
rs781986273 Chr.X: 149713157-149713157 on Build GRCh38
rs142651260 Chr.X: 149713170-149713170 on Build GRCh38
rs782494490 Chr.X: 149713231-149713231 on Build GRCh38
rs782026785 Chr.X: 149713262-149713262 on Build GRCh38
rs782095623 Chr.X: 149713292-149713292 on Build GRCh38
rs368566721 Chr.X: 149713269-149713269 on Build GRCh38
rs1970766 Chr.X: 149713293-149713293 on Build GRCh38
rs782402135 Chr.X: 149713260-149713260 on Build GRCh38
rs782519445 Chr.X: 149713201-149713201 on Build GRCh38
rs781878857 Chr.X: 149713306-149713306 on Build GRCh38
rs782393534 Chr.X: 149713273-149713273 on Build GRCh38
rs781903897 Chr.X: 149713183-149713183 on Build GRCh38
rs782195407 Chr.X: 149713250-149713250 on Build GRCh38
rs782557635 Chr.X: 149713309-149713309 on Build GRCh38
rs782796516 Chr.X: 149713176-149713176 on Build GRCh38
rs782568667 Chr.X: 149713154-149713154 on Build GRCh38
rs782456489 Chr.X: 149713175-149713175 on Build GRCh38
rs375047042 Chr.X: 149713294-149713294 on Build GRCh38
rs782801638 Chr.X: 149713168-149713168 on Build GRCh38
rs782563478 Chr.X: 149713249-149713249 on Build GRCh38
rs186341107 Chr.X: 149713127-149713127 on Build GRCh38
rs782765668 Chr.X: 149713305-149713305 on Build GRCh38
rs376083752 Chr.X: 149713209-149713209 on Build GRCh38
rs782497523 Chr.X: 149713317-149713317 on Build GRCh38
rs782649959 Chr.X: 149713203-149713203 on Build GRCh38
rs782770625 Chr.X: 149713275-149713275 on Build GRCh38
rs782491762 Chr.X: 149713298-149713298 on Build GRCh38

View Full Product Details