Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45823790-45824041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNNC2
Entrez Gene ID
7125
Gene Name
troponin C2, fast skeletal type
Gene Aliases
-
UniGene
Hs.182421
Gene Chromosome Location
Chr.20: 45823214-45833329 on Build GRCh38
Transcripts
NM_003279.2 XM_011529031.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4400718 c.409A>C p.K137Q Chr.20: 45824033-45824033 on Build GRCh38
6805404 c.415G>A p.G139S Chr.20: 45824027-45824027 on Build GRCh38
5790019 c.415G>C p.G139R Chr.20: 45824027-45824027 on Build GRCh38
97846 c.408G>T p.M136I Chr.20: 45824034-45824034 on Build GRCh38
1027287 c.417C>T p.G139G Chr.20: 45824025-45824025 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759345093 Chr.20: 45823951-45823951 on Build GRCh38
rs964792496 Chr.20: 45823908-45823908 on Build GRCh38
rs747329067 Chr.20: 45824011-45824011 on Build GRCh38
rs948382357 Chr.20: 45824000-45824000 on Build GRCh38
rs781622000 Chr.20: 45824009-45824009 on Build GRCh38
rs900335382 Chr.20: 45824001-45824001 on Build GRCh38
rs551547278 Chr.20: 45823907-45823907 on Build GRCh38
rs117822901 Chr.20: 45823920-45823920 on Build GRCh38
rs764660804 Chr.20: 45823929-45823929 on Build GRCh38
rs989529171 Chr.20: 45823911-45823911 on Build GRCh38
rs977637654 Chr.20: 45823924-45823924 on Build GRCh38
rs764214103 Chr.20: 45823970-45823970 on Build GRCh38
rs535210380 Chr.20: 45823828-45823828 on Build GRCh38
rs924565990 Chr.20: 45823931-45823931 on Build GRCh38
rs754004139 Chr.20: 45823980-45823980 on Build GRCh38
rs757336473 Chr.20: 45823992-45823992 on Build GRCh38
rs944529682 Chr.20: 45823827-45823827 on Build GRCh38
rs947240829 Chr.20: 45823921-45823921 on Build GRCh38
rs530394831 Chr.20: 45823817-45823817 on Build GRCh38
rs778920626 Chr.20: 45823995-45823995 on Build GRCh38
rs767395238 Chr.20: 45823961-45823961 on Build GRCh38
rs1017998505 Chr.20: 45823854-45823854 on Build GRCh38
rs758805359 Chr.20: 45824005-45824005 on Build GRCh38
rs761028276 Chr.20: 45823967-45823967 on Build GRCh38
rs955116956 Chr.20: 45823818-45823818 on Build GRCh38
rs113240353 Chr.20: 45824002-45824002 on Build GRCh38
rs537198656 Chr.20: 45823963-45823963 on Build GRCh38
rs780503233 Chr.20: 45824010-45824010 on Build GRCh38

View Full Product Details