Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137214357-137214608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDOR1
Entrez Gene ID
27158
Gene Name
NADPH dependent diflavin oxidoreductase 1
Gene Aliases
NR1, bA350O14.9
UniGene
Hs.512564
Gene Chromosome Location
Chr.9: 137205667-137219361 on Build GRCh38
Transcripts
NM_001144026.2 NM_001144027.2 NM_001144028.2 NM_014434.3 XM_011518545.1 XM_011518547.1 XM_011518548.2 XM_017014626.1 XM_017014627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6449031 c.747G>A p.L249L Chr.9: 137214594-137214594 on Build GRCh38
7222756 c.722+3G>A p.? Chr.9: 137214416-137214416 on Build GRCh38
5430229 c.668C>T p.S223F Chr.9: 137214359-137214359 on Build GRCh38
5021659 c.714T>C p.S238S Chr.9: 137214405-137214405 on Build GRCh38
3395692 c.739G>A p.V247M Chr.9: 137214586-137214586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949237149 Chr.9: 137214511-137214511 on Build GRCh38
rs776535516 Chr.9: 137214455-137214455 on Build GRCh38
rs771613393 Chr.9: 137214438-137214438 on Build GRCh38
rs920378373 Chr.9: 137214489-137214489 on Build GRCh38
rs745593114 Chr.9: 137214554-137214554 on Build GRCh38
rs768704351 Chr.9: 137214457-137214457 on Build GRCh38
rs778359036 Chr.9: 137214534-137214534 on Build GRCh38
rs139883255 Chr.9: 137214465-137214465 on Build GRCh38
rs749805431 Chr.9: 137214464-137214466 on Build GRCh38
rs760476937 Chr.9: 137214387-137214387 on Build GRCh38
rs761142430 Chr.9: 137214526-137214526 on Build GRCh38
rs538940688 Chr.9: 137214512-137214512 on Build GRCh38
rs766907612 Chr.9: 137214529-137214529 on Build GRCh38
rs780378837 Chr.9: 137214423-137214423 on Build GRCh38
rs538445745 Chr.9: 137214564-137214564 on Build GRCh38
rs76600786 Chr.9: 137214464-137214464 on Build GRCh38
rs747551558 Chr.9: 137214433-137214433 on Build GRCh38
rs773212598 Chr.9: 137214384-137214384 on Build GRCh38
rs759427646 Chr.9: 137214456-137214456 on Build GRCh38
rs77926343 Chr.9: 137214517-137214517 on Build GRCh38
rs754149051 Chr.9: 137214532-137214532 on Build GRCh38
rs974220049 Chr.9: 137214480-137214480 on Build GRCh38
rs73563696 Chr.9: 137214405-137214405 on Build GRCh38
rs769264248 Chr.9: 137214462-137214463 on Build GRCh38
rs770843750 Chr.9: 137214388-137214388 on Build GRCh38
rs764213624 Chr.9: 137214520-137214520 on Build GRCh38
rs777213725 Chr.9: 137214439-137214439 on Build GRCh38
rs947820513 Chr.9: 137214498-137214498 on Build GRCh38
rs774639597 Chr.9: 137214461-137214461 on Build GRCh38
rs776623658 Chr.9: 137214394-137214394 on Build GRCh38
rs757220701 Chr.9: 137214540-137214540 on Build GRCh38
rs377723853 Chr.9: 137214441-137214441 on Build GRCh38
rs373365492 Chr.9: 137214420-137214420 on Build GRCh38
rs937909292 Chr.9: 137214479-137214479 on Build GRCh38
rs113736428 Chr.9: 137214397-137214397 on Build GRCh38
rs373933917 Chr.9: 137214576-137214576 on Build GRCh38
rs750938696 Chr.9: 137214415-137214415 on Build GRCh38
rs199964308 Chr.9: 137214537-137214537 on Build GRCh38
rs754856659 Chr.9: 137214537-137214538 on Build GRCh38
rs750054866 Chr.9: 137214578-137214578 on Build GRCh38
rs1043566115 Chr.9: 137214530-137214530 on Build GRCh38
rs891101100 Chr.9: 137214559-137214559 on Build GRCh38
rs371399787 Chr.9: 137214442-137214442 on Build GRCh38
rs752387696 Chr.9: 137214535-137214535 on Build GRCh38
rs778687528 Chr.9: 137214586-137214588 on Build GRCh38
rs764416218 Chr.9: 137214400-137214400 on Build GRCh38
rs368880107 Chr.9: 137214536-137214536 on Build GRCh38
rs746556804 Chr.9: 137214539-137214539 on Build GRCh38
rs566904031 Chr.9: 137214508-137214508 on Build GRCh38
rs1004201299 Chr.9: 137214572-137214572 on Build GRCh38
rs62587578 Chr.9: 137214585-137214585 on Build GRCh38
rs367981974 Chr.9: 137214533-137214533 on Build GRCh38
rs550756832 Chr.9: 137214428-137214428 on Build GRCh38
rs762197152 Chr.9: 137214465-137214466 on Build GRCh38
rs767846125 Chr.9: 137214412-137214412 on Build GRCh38
rs117858988 Chr.9: 137214416-137214416 on Build GRCh38
rs368322055 Chr.9: 137214474-137214474 on Build GRCh38
rs149701237 Chr.9: 137214464-137214465 on Build GRCh38
rs979034674 Chr.9: 137214471-137214471 on Build GRCh38
rs769629715 Chr.9: 137214557-137214557 on Build GRCh38
rs547006074 Chr.9: 137214487-137214487 on Build GRCh38
rs773564096 Chr.9: 137214463-137214464 on Build GRCh38
rs762009913 Chr.9: 137214519-137214519 on Build GRCh38
rs375495144 Chr.9: 137214521-137214521 on Build GRCh38
rs897202277 Chr.9: 137214515-137214515 on Build GRCh38
rs569376463 Chr.9: 137214432-137214432 on Build GRCh38
rs995597945 Chr.9: 137214516-137214516 on Build GRCh38
rs761968397 Chr.9: 137214411-137214411 on Build GRCh38

View Full Product Details