Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138459066-138459339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESYT3
Entrez Gene ID
83850
Gene Name
extended synaptotagmin 3
Gene Aliases
CHR3SYT, E-Syt3, FAM62C
UniGene
Hs.477711
Gene Chromosome Location
Chr.3: 138434573-138479942 on Build GRCh38
Transcripts
NM_001322831.1 NM_001322834.1 NM_031913.4 NR_136409.1 XM_017007307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6656588 c.589G>A p.G197R Chr.3: 138459194-138459194 on Build GRCh38
4398631 c.642G>A p.G214G Chr.3: 138459247-138459247 on Build GRCh38
3846356 c.648+2T>G p.? Chr.3: 138459255-138459255 on Build GRCh38
5465728 c.621G>T p.K207N Chr.3: 138459226-138459226 on Build GRCh38
728174 c.645C>T p.I215I Chr.3: 138459250-138459250 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766504589 Chr.3: 138459302-138459302 on Build GRCh38
rs527514639 Chr.3: 138459245-138459245 on Build GRCh38
rs771757852 Chr.3: 138459148-138459148 on Build GRCh38
rs751794447 Chr.3: 138459195-138459195 on Build GRCh38
rs536590426 Chr.3: 138459282-138459282 on Build GRCh38
rs1007702261 Chr.3: 138459179-138459179 on Build GRCh38
rs954139311 Chr.3: 138459315-138459315 on Build GRCh38
rs759951737 Chr.3: 138459294-138459295 on Build GRCh38
rs781357781 Chr.3: 138459238-138459238 on Build GRCh38
rs113171647 Chr.3: 138459172-138459172 on Build GRCh38
rs189455154 Chr.3: 138459261-138459261 on Build GRCh38
rs759014154 Chr.3: 138459145-138459145 on Build GRCh38
rs552818597 Chr.3: 138459096-138459096 on Build GRCh38
rs1028798908 Chr.3: 138459308-138459308 on Build GRCh38
rs768342169 Chr.3: 138459267-138459267 on Build GRCh38
rs930336227 Chr.3: 138459101-138459101 on Build GRCh38
rs139594127 Chr.3: 138459193-138459193 on Build GRCh38
rs199936858 Chr.3: 138459266-138459266 on Build GRCh38
rs750573202 Chr.3: 138459176-138459176 on Build GRCh38
rs752082758 Chr.3: 138459113-138459113 on Build GRCh38
rs551467666 Chr.3: 138459130-138459130 on Build GRCh38
rs371677867 Chr.3: 138459276-138459276 on Build GRCh38
rs556194356 Chr.3: 138459305-138459305 on Build GRCh38
rs372035292 Chr.3: 138459259-138459259 on Build GRCh38
rs774820681 Chr.3: 138459151-138459152 on Build GRCh38
rs897909979 Chr.3: 138459162-138459162 on Build GRCh38
rs987728046 Chr.3: 138459247-138459247 on Build GRCh38
rs760492296 Chr.3: 138459153-138459153 on Build GRCh38
rs907444913 Chr.3: 138459221-138459221 on Build GRCh38
rs778318367 Chr.3: 138459257-138459257 on Build GRCh38
rs893091197 Chr.3: 138459287-138459287 on Build GRCh38
rs776357118 Chr.3: 138459273-138459273 on Build GRCh38
rs763729082 Chr.3: 138459160-138459160 on Build GRCh38
rs144490386 Chr.3: 138459191-138459191 on Build GRCh38
rs775127784 Chr.3: 138459264-138459264 on Build GRCh38
rs896421126 Chr.3: 138459090-138459090 on Build GRCh38
rs368357672 Chr.3: 138459178-138459178 on Build GRCh38
rs531646765 Chr.3: 138459126-138459126 on Build GRCh38
rs755402736 Chr.3: 138459210-138459210 on Build GRCh38
rs761715535 Chr.3: 138459170-138459170 on Build GRCh38
rs377068443 Chr.3: 138459155-138459155 on Build GRCh38
rs145007016 Chr.3: 138459244-138459244 on Build GRCh38
rs775074414 Chr.3: 138459149-138459149 on Build GRCh38

View Full Product Details