Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154478260-154478515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAGE3
Entrez Gene ID
8270
Gene Name
L antigen family member 3
Gene Aliases
CVG5, DXS9879E, DXS9951E, ESO3, ITBA2, Pcc1
UniGene
Hs.444619
Gene Chromosome Location
Chr.X: 154477769-154479257 on Build GRCh38
Transcripts
NM_006014.4

SNP Detail

SNP ID Chromosome Location
rs1037257365 Chr.X: 154478298-154478298 on Build GRCh38
rs782731371 Chr.X: 154478283-154478283 on Build GRCh38
rs782792256 Chr.X: 154478294-154478294 on Build GRCh38
rs782430798 Chr.X: 154478326-154478326 on Build GRCh38
rs782011954 Chr.X: 154478440-154478440 on Build GRCh38
rs782103437 Chr.X: 154478432-154478432 on Build GRCh38
rs377753533 Chr.X: 154478387-154478387 on Build GRCh38
rs782207715 Chr.X: 154478333-154478333 on Build GRCh38
rs782344105 Chr.X: 154478413-154478413 on Build GRCh38
rs199499929 Chr.X: 154478407-154478407 on Build GRCh38
rs782593315 Chr.X: 154478330-154478330 on Build GRCh38
rs376379947 Chr.X: 154478369-154478369 on Build GRCh38
rs201041758 Chr.X: 154478313-154478313 on Build GRCh38
rs1007283352 Chr.X: 154478476-154478476 on Build GRCh38
rs782785819 Chr.X: 154478433-154478433 on Build GRCh38
rs11538410 Chr.X: 154478340-154478340 on Build GRCh38
rs781852260 Chr.X: 154478288-154478288 on Build GRCh38
rs782695876 Chr.X: 154478452-154478452 on Build GRCh38
rs781872781 Chr.X: 154478462-154478462 on Build GRCh38
rs782164177 Chr.X: 154478350-154478350 on Build GRCh38
rs371344200 Chr.X: 154478370-154478370 on Build GRCh38
rs782283522 Chr.X: 154478363-154478363 on Build GRCh38
rs781878793 Chr.X: 154478496-154478496 on Build GRCh38
rs374116161 Chr.X: 154478386-154478386 on Build GRCh38
rs12392443 Chr.X: 154478445-154478445 on Build GRCh38
rs782786534 Chr.X: 154478417-154478417 on Build GRCh38
rs782479618 Chr.X: 154478460-154478460 on Build GRCh38
rs782480742 Chr.X: 154478455-154478455 on Build GRCh38
rs897387368 Chr.X: 154478331-154478331 on Build GRCh38
rs782336210 Chr.X: 154478453-154478454 on Build GRCh38
rs782229180 Chr.X: 154478435-154478435 on Build GRCh38

View Full Product Details