Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14843412-14843639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs759020755 Chr.X: 14843594-14843594 on Build GRCh38
rs2375726 Chr.X: 14843553-14843553 on Build GRCh38
rs920824221 Chr.X: 14843468-14843468 on Build GRCh38
rs745355429 Chr.X: 14843559-14843564 on Build GRCh38
rs769023794 Chr.X: 14843528-14843528 on Build GRCh38
rs143434225 Chr.X: 14843501-14843501 on Build GRCh38
rs763533295 Chr.X: 14843566-14843566 on Build GRCh38
rs764467478 Chr.X: 14843438-14843438 on Build GRCh38
rs773885877 Chr.X: 14843584-14843584 on Build GRCh38
rs187611308 Chr.X: 14843534-14843534 on Build GRCh38
rs757677526 Chr.X: 14843488-14843488 on Build GRCh38
rs935647794 Chr.X: 14843525-14843525 on Build GRCh38
rs755704953 Chr.X: 14843496-14843496 on Build GRCh38
rs773654012 Chr.X: 14843564-14843564 on Build GRCh38
rs771278010 Chr.X: 14843572-14843572 on Build GRCh38
rs927015579 Chr.X: 14843509-14843509 on Build GRCh38
rs781315536 Chr.X: 14843443-14843443 on Build GRCh38
rs992299568 Chr.X: 14843459-14843459 on Build GRCh38
rs948292480 Chr.X: 14843493-14843493 on Build GRCh38
rs951545082 Chr.X: 14843455-14843455 on Build GRCh38
rs147973478 Chr.X: 14843469-14843469 on Build GRCh38
rs770685871 Chr.X: 14843593-14843593 on Build GRCh38

View Full Product Details