Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120461924-120462197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATC
Entrez Gene ID
283459
Gene Name
glutamyl-tRNA amidotransferase subunit C
Gene Aliases
15E1.2
UniGene
Hs.706889
Gene Chromosome Location
Chr.12: 120446438-120463753 on Build GRCh38
Transcripts
NM_176818.2 NR_033684.1 NM_003769.2

Gene Symbol
SRSF9
Entrez Gene ID
8683
Gene Name
serine and arginine rich splicing factor 9
Gene Aliases
SFRS9, SRp30c
UniGene
Hs.706889
Gene Chromosome Location
Chr.12: 120461668-120469755 on Build GRCh38
Transcripts
NM_176818.2 NR_033684.1 NM_003769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4636883 c.658C>T p.P220S Chr.12: 120462027-120462027 on Build GRCh38
3359579 c.533C>T p.S178F Chr.12: 120462152-120462152 on Build GRCh38
6790644 c.580C>T p.R194W Chr.12: 120462105-120462105 on Build GRCh38
4039402 c.531T>G p.T177T Chr.12: 120462154-120462154 on Build GRCh38
1127534 c.576C>A p.Y192* Chr.12: 120462109-120462109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755150306 Chr.12: 120462004-120462004 on Build GRCh38
rs999382570 Chr.12: 120461956-120461956 on Build GRCh38
rs748166545 Chr.12: 120462019-120462019 on Build GRCh38
rs780370849 Chr.12: 120461996-120461996 on Build GRCh38
rs146381618 Chr.12: 120461972-120461972 on Build GRCh38
rs752857364 Chr.12: 120462111-120462111 on Build GRCh38
rs750984394 Chr.12: 120462062-120462062 on Build GRCh38
rs779936408 Chr.12: 120462165-120462166 on Build GRCh38
rs750852833 Chr.12: 120461970-120461972 on Build GRCh38
rs745603265 Chr.12: 120462170-120462170 on Build GRCh38
rs781270406 Chr.12: 120462008-120462008 on Build GRCh38
rs751870495 Chr.12: 120462097-120462097 on Build GRCh38
rs760167949 Chr.12: 120462057-120462057 on Build GRCh38
rs751817320 Chr.12: 120461999-120461999 on Build GRCh38
rs940710190 Chr.12: 120462047-120462047 on Build GRCh38
rs766758485 Chr.12: 120462081-120462081 on Build GRCh38
rs749288623 Chr.12: 120462144-120462144 on Build GRCh38
rs770810155 Chr.12: 120462043-120462043 on Build GRCh38
rs757222703 Chr.12: 120462148-120462148 on Build GRCh38
rs370436477 Chr.12: 120462041-120462041 on Build GRCh38
rs758963911 Chr.12: 120461986-120461987 on Build GRCh38
rs758851796 Chr.12: 120461994-120461994 on Build GRCh38
rs201844994 Chr.12: 120462109-120462109 on Build GRCh38
rs148192217 Chr.12: 120462116-120462116 on Build GRCh38
rs756275363 Chr.12: 120462121-120462121 on Build GRCh38
rs139360328 Chr.12: 120462080-120462080 on Build GRCh38
rs763511362 Chr.12: 120462058-120462058 on Build GRCh38
rs759295143 Chr.12: 120462051-120462051 on Build GRCh38
rs775156072 Chr.12: 120462056-120462056 on Build GRCh38
rs755203606 Chr.12: 120462102-120462102 on Build GRCh38
rs920881426 Chr.12: 120462085-120462085 on Build GRCh38
rs776930825 Chr.12: 120462038-120462038 on Build GRCh38
rs376520463 Chr.12: 120461983-120461983 on Build GRCh38
rs777693373 Chr.12: 120462143-120462143 on Build GRCh38
rs201309578 Chr.12: 120461954-120461954 on Build GRCh38
rs769622746 Chr.12: 120462026-120462026 on Build GRCh38
rs778875064 Chr.12: 120462154-120462154 on Build GRCh38
rs774258544 Chr.12: 120462046-120462046 on Build GRCh38
rs771804226 Chr.12: 120462052-120462052 on Build GRCh38
rs777833144 Chr.12: 120462031-120462031 on Build GRCh38
rs372204543 Chr.12: 120461969-120461969 on Build GRCh38

View Full Product Details