Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 88801559-88801784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNGTT
Entrez Gene ID
8732
Gene Name
RNA guanylyltransferase and 5'-phosphatase
Gene Aliases
CAP1A, HCE, HCE1, hCAP
UniGene
Hs.567378
Gene Chromosome Location
Chr.6: 88609897-88963716 on Build GRCh38
Transcripts
NM_001286426.1 NM_001286428.1 NM_003800.4 XM_017011401.1 XM_017011402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4863388 c.1294G>T p.E432* Chr.6: 88801608-88801608 on Build GRCh38
1082413 c.1294G>T p.E432* Chr.6: 88801608-88801608 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs545177478 Chr.6: 88801742-88801742 on Build GRCh38
rs941611828 Chr.6: 88801749-88801749 on Build GRCh38
rs188536650 Chr.6: 88801711-88801711 on Build GRCh38
rs979160063 Chr.6: 88801709-88801709 on Build GRCh38
rs910166925 Chr.6: 88801743-88801743 on Build GRCh38
rs1018104722 Chr.6: 88801688-88801688 on Build GRCh38
rs12189561 Chr.6: 88801714-88801714 on Build GRCh38
rs753971554 Chr.6: 88801595-88801595 on Build GRCh38
rs764206640 Chr.6: 88801588-88801588 on Build GRCh38
rs747846239 Chr.6: 88801658-88801658 on Build GRCh38
rs148889826 Chr.6: 88801629-88801629 on Build GRCh38
rs780992342 Chr.6: 88801647-88801647 on Build GRCh38
rs929971667 Chr.6: 88801728-88801728 on Build GRCh38
rs753265745 Chr.6: 88801671-88801671 on Build GRCh38
rs769410063 Chr.6: 88801660-88801660 on Build GRCh38
rs772550399 Chr.6: 88801668-88801668 on Build GRCh38
rs138793662 Chr.6: 88801701-88801701 on Build GRCh38
rs748832392 Chr.6: 88801670-88801670 on Build GRCh38
rs554018152 Chr.6: 88801750-88801750 on Build GRCh38
rs372615584 Chr.6: 88801645-88801645 on Build GRCh38
rs59654511 Chr.6: 88801651-88801651 on Build GRCh38
rs758230940 Chr.6: 88801596-88801596 on Build GRCh38
rs373661527 Chr.6: 88801612-88801612 on Build GRCh38
rs922608424 Chr.6: 88801726-88801726 on Build GRCh38
rs769607503 Chr.6: 88801635-88801635 on Build GRCh38
rs983000719 Chr.6: 88801732-88801732 on Build GRCh38
rs780046328 Chr.6: 88801623-88801623 on Build GRCh38
rs771281901 Chr.6: 88801674-88801674 on Build GRCh38
rs754878389 Chr.6: 88801637-88801637 on Build GRCh38
rs7449471 Chr.6: 88801735-88801735 on Build GRCh38

View Full Product Details