Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178149662-178149895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHP2
Entrez Gene ID
55651
Gene Name
NHP2 ribonucleoprotein
Gene Aliases
DKCB2, NHP2P, NOLA2
UniGene
Hs.744074
Gene Chromosome Location
Chr.5: 178149463-178153960 on Build GRCh38
Transcripts
NM_001034833.1 NM_017838.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1066457 c.435G>A p.E145E Chr.5: 178149740-178149740 on Build GRCh38
1066458 c.352G>A p.A118T Chr.5: 178149823-178149823 on Build GRCh38
1066459 c.348A>G p.A116A Chr.5: 178149827-178149827 on Build GRCh38
449413 c.403T>C p.Y135H Chr.5: 178149772-178149772 on Build GRCh38
3661744 c.395A>T p.H132L Chr.5: 178149780-178149780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745927459 Chr.5: 178149697-178149697 on Build GRCh38
rs747921841 Chr.5: 178149778-178149778 on Build GRCh38
rs750874926 Chr.5: 178149820-178149820 on Build GRCh38
rs747261406 Chr.5: 178149699-178149699 on Build GRCh38
rs770411732 Chr.5: 178149785-178149785 on Build GRCh38
rs200122862 Chr.5: 178149690-178149690 on Build GRCh38
rs201618462 Chr.5: 178149691-178149691 on Build GRCh38
rs121908091 Chr.5: 178149715-178149715 on Build GRCh38
rs759145032 Chr.5: 178149792-178149792 on Build GRCh38
rs375285426 Chr.5: 178149806-178149806 on Build GRCh38
rs1044969988 Chr.5: 178149722-178149722 on Build GRCh38
rs944016113 Chr.5: 178149682-178149682 on Build GRCh38
rs780412873 Chr.5: 178149683-178149683 on Build GRCh38
rs748734070 Chr.5: 178149860-178149860 on Build GRCh38
rs143501297 Chr.5: 178149741-178149741 on Build GRCh38
rs140275983 Chr.5: 178149819-178149819 on Build GRCh38
rs755971433 Chr.5: 178149853-178149853 on Build GRCh38
rs554358239 Chr.5: 178149704-178149704 on Build GRCh38
rs770801671 Chr.5: 178149868-178149868 on Build GRCh38
rs749194693 Chr.5: 178149864-178149864 on Build GRCh38
rs752392516 Chr.5: 178149811-178149811 on Build GRCh38
rs781096695 Chr.5: 178149767-178149767 on Build GRCh38
rs778922299 Chr.5: 178149695-178149695 on Build GRCh38
rs769022015 Chr.5: 178149749-178149749 on Build GRCh38
rs121908090 Chr.5: 178149799-178149799 on Build GRCh38
rs759194933 Chr.5: 178149716-178149716 on Build GRCh38
rs773752330 Chr.5: 178149786-178149786 on Build GRCh38
rs121908089 Chr.5: 178149760-178149760 on Build GRCh38
rs754228756 Chr.5: 178149728-178149728 on Build GRCh38
rs762150542 Chr.5: 178149812-178149812 on Build GRCh38
rs757780182 Chr.5: 178149692-178149692 on Build GRCh38
rs146766601 Chr.5: 178149781-178149781 on Build GRCh38
rs757655789 Chr.5: 178149734-178149734 on Build GRCh38
rs1004048449 Chr.5: 178149794-178149794 on Build GRCh38
rs561948156 Chr.5: 178149836-178149836 on Build GRCh38
rs563885344 Chr.5: 178149743-178149743 on Build GRCh38
rs769885801 Chr.5: 178149703-178149703 on Build GRCh38
rs369301843 Chr.5: 178149707-178149707 on Build GRCh38
rs766666396 Chr.5: 178149683-178149683 on Build GRCh38
rs755460344 Chr.5: 178149779-178149780 on Build GRCh38
rs769763413 Chr.5: 178149779-178149779 on Build GRCh38
rs201984406 Chr.5: 178149681-178149681 on Build GRCh38
rs762870297 Chr.5: 178149784-178149784 on Build GRCh38
rs200781371 Chr.5: 178149702-178149702 on Build GRCh38
rs537891538 Chr.5: 178149684-178149684 on Build GRCh38
rs567317314 Chr.5: 178149758-178149758 on Build GRCh38
rs768498847 Chr.5: 178149701-178149701 on Build GRCh38
rs772091448 Chr.5: 178149698-178149698 on Build GRCh38
rs372641598 Chr.5: 178149689-178149689 on Build GRCh38
rs779366459 Chr.5: 178149791-178149792 on Build GRCh38
rs139588879 Chr.5: 178149823-178149823 on Build GRCh38
rs781544363 Chr.5: 178149844-178149844 on Build GRCh38
rs150844606 Chr.5: 178149705-178149705 on Build GRCh38
rs747197452 Chr.5: 178149744-178149744 on Build GRCh38
rs754309481 Chr.5: 178149723-178149723 on Build GRCh38
rs1023073784 Chr.5: 178149843-178149843 on Build GRCh38
rs547248309 Chr.5: 178149863-178149863 on Build GRCh38
rs1028921372 Chr.5: 178149849-178149849 on Build GRCh38
rs185752917 Chr.5: 178149709-178149709 on Build GRCh38
rs764390414 Chr.5: 178149718-178149718 on Build GRCh38
rs115334254 Chr.5: 178149824-178149824 on Build GRCh38
rs779452625 Chr.5: 178149736-178149736 on Build GRCh38
rs201653595 Chr.5: 178149852-178149852 on Build GRCh38
rs147575606 Chr.5: 178149867-178149867 on Build GRCh38
rs149168003 Chr.5: 178149840-178149840 on Build GRCh38

View Full Product Details