Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46494958-46495195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNMAL2
Entrez Gene ID
57469
Gene Name
paraneoplastic Ma antigen family-like 2
Gene Aliases
-
UniGene
Hs.7193
Gene Chromosome Location
Chr.19: 46491191-46495912 on Build GRCh38
Transcripts
NM_020709.2 NM_001205281.1

Gene Symbol
PPP5D1
Entrez Gene ID
100506012
Gene Name
PPP5 tetratricopeptide repeat domain containing 1
Gene Aliases
-
UniGene
Hs.671661
Gene Chromosome Location
Chr.19: 46480788-46601200 on Build GRCh38
Transcripts
NM_020709.2 NM_001205281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6751354 c.395C>T p.A132V Chr.19: 46495071-46495071 on Build GRCh38
3707236 c.332T>C p.L111P Chr.19: 46495134-46495134 on Build GRCh38
3707237 c.332T>C p.L111P Chr.19: 46495134-46495134 on Build GRCh38
5714167 c.353C>T p.A118V Chr.19: 46495113-46495113 on Build GRCh38
5505980 c.353G>A p.R118Q Chr.19: 46494985-46494985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761589351 Chr.19: 46495098-46495098 on Build GRCh38
rs780603949 Chr.19: 46495063-46495063 on Build GRCh38
rs764616898 Chr.19: 46495052-46495052 on Build GRCh38
rs4803967 Chr.19: 46494979-46494979 on Build GRCh38
rs975093022 Chr.19: 46495056-46495056 on Build GRCh38
rs78382198 Chr.19: 46495115-46495115 on Build GRCh38
rs201169835 Chr.19: 46495079-46495079 on Build GRCh38
rs755398401 Chr.19: 46495126-46495126 on Build GRCh38
rs769704644 Chr.19: 46494982-46494982 on Build GRCh38
rs372628659 Chr.19: 46494987-46494987 on Build GRCh38
rs761396001 Chr.19: 46495046-46495046 on Build GRCh38
rs148479589 Chr.19: 46495033-46495033 on Build GRCh38
rs756259750 Chr.19: 46495136-46495136 on Build GRCh38
rs758805358 Chr.19: 46495061-46495061 on Build GRCh38
rs779973734 Chr.19: 46495166-46495166 on Build GRCh38
rs763657526 Chr.19: 46494988-46495008 on Build GRCh38
rs781577305 Chr.19: 46495072-46495072 on Build GRCh38
rs759344802 Chr.19: 46494995-46494995 on Build GRCh38
rs749317461 Chr.19: 46495153-46495153 on Build GRCh38
rs775236274 Chr.19: 46495020-46495020 on Build GRCh38
rs767953355 Chr.19: 46495129-46495129 on Build GRCh38
rs763619828 Chr.19: 46495112-46495112 on Build GRCh38
rs745860177 Chr.19: 46495080-46495080 on Build GRCh38
rs926756614 Chr.19: 46495004-46495004 on Build GRCh38
rs768465930 Chr.19: 46495175-46495175 on Build GRCh38
rs757271622 Chr.19: 46495165-46495165 on Build GRCh38
rs755306751 Chr.19: 46495070-46495070 on Build GRCh38
rs751965570 Chr.19: 46495068-46495068 on Build GRCh38
rs924167856 Chr.19: 46495090-46495090 on Build GRCh38
rs538950722 Chr.19: 46495144-46495144 on Build GRCh38
rs141479764 Chr.19: 46495088-46495088 on Build GRCh38
rs139994198 Chr.19: 46495077-46495077 on Build GRCh38
rs1016921349 Chr.19: 46495151-46495151 on Build GRCh38
rs146030178 Chr.19: 46495044-46495044 on Build GRCh38
rs373192275 Chr.19: 46495133-46495133 on Build GRCh38
rs750958683 Chr.19: 46495057-46495057 on Build GRCh38
rs774349867 Chr.19: 46494985-46494985 on Build GRCh38
rs760388098 Chr.19: 46495026-46495026 on Build GRCh38
rs539300965 Chr.19: 46495084-46495084 on Build GRCh38
rs775324606 Chr.19: 46495087-46495087 on Build GRCh38
rs752053547 Chr.19: 46495123-46495123 on Build GRCh38
rs572994007 Chr.19: 46495109-46495109 on Build GRCh38
rs746913214 Chr.19: 46495174-46495174 on Build GRCh38
rs146188530 Chr.19: 46495097-46495097 on Build GRCh38
rs1056186421 Chr.19: 46495032-46495032 on Build GRCh38
rs776264051 Chr.19: 46495176-46495176 on Build GRCh38
rs771904712 Chr.19: 46495015-46495015 on Build GRCh38
rs776381212 Chr.19: 46495096-46495096 on Build GRCh38

View Full Product Details