Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1775909-1776140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EME2
Entrez Gene ID
197342
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
SLX2B, gs125
UniGene
Hs.7247
Gene Chromosome Location
Chr.16: 1773222-1776714 on Build GRCh38
Transcripts
NM_001257370.1 XM_017023025.1 XM_017023026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3988269 c.1166C>T p.A389V Chr.16: 1776069-1776069 on Build GRCh38
1716098 c.1124C>T p.A375V Chr.16: 1775946-1775946 on Build GRCh38
6653664 c.1150C>T p.R384C Chr.16: 1775972-1775972 on Build GRCh38
3944397 c.1167G>A p.A389A Chr.16: 1776070-1776070 on Build GRCh38
5850290 c.1142C>T p.P381L Chr.16: 1775964-1775964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771589337 Chr.16: 1775966-1775967 on Build GRCh38
rs747923894 Chr.16: 1776000-1776000 on Build GRCh38
rs199593669 Chr.16: 1776074-1776074 on Build GRCh38
rs766792983 Chr.16: 1776068-1776068 on Build GRCh38
rs1025604407 Chr.16: 1775930-1775930 on Build GRCh38
rs534005461 Chr.16: 1776103-1776103 on Build GRCh38
rs141577357 Chr.16: 1776109-1776109 on Build GRCh38
rs761145404 Chr.16: 1776067-1776067 on Build GRCh38
rs759022450 Chr.16: 1775967-1775967 on Build GRCh38
rs571431327 Chr.16: 1776102-1776102 on Build GRCh38
rs769388005 Chr.16: 1776089-1776089 on Build GRCh38
rs779513633 Chr.16: 1775946-1775946 on Build GRCh38
rs777091490 Chr.16: 1776099-1776099 on Build GRCh38
rs80194604 Chr.16: 1776010-1776010 on Build GRCh38
rs745411622 Chr.16: 1775961-1775965 on Build GRCh38
rs765733601 Chr.16: 1776101-1776101 on Build GRCh38
rs768907545 Chr.16: 1775939-1775939 on Build GRCh38
rs750409117 Chr.16: 1776115-1776115 on Build GRCh38
rs190542947 Chr.16: 1776040-1776040 on Build GRCh38
rs770066405 Chr.16: 1775935-1775935 on Build GRCh38
rs558222106 Chr.16: 1776112-1776112 on Build GRCh38
rs746956947 Chr.16: 1776042-1776042 on Build GRCh38
rs201406919 Chr.16: 1776030-1776030 on Build GRCh38
rs142106391 Chr.16: 1776039-1776039 on Build GRCh38
rs61753375 Chr.16: 1775981-1775981 on Build GRCh38
rs745463843 Chr.16: 1776049-1776049 on Build GRCh38
rs571694867 Chr.16: 1776006-1776006 on Build GRCh38
rs138348894 Chr.16: 1775965-1775965 on Build GRCh38
rs563874572 Chr.16: 1775964-1775964 on Build GRCh38
rs1008242861 Chr.16: 1775948-1775948 on Build GRCh38
rs529141086 Chr.16: 1775971-1775971 on Build GRCh38
rs146182784 Chr.16: 1776111-1776111 on Build GRCh38
rs757852342 Chr.16: 1775931-1775954 on Build GRCh38
rs781130749 Chr.16: 1776113-1776113 on Build GRCh38
rs377219719 Chr.16: 1776034-1776034 on Build GRCh38
rs1041231585 Chr.16: 1776026-1776026 on Build GRCh38
rs774399655 Chr.16: 1775940-1775940 on Build GRCh38
rs9929680 Chr.16: 1776022-1776022 on Build GRCh38
rs763553959 Chr.16: 1776061-1776061 on Build GRCh38
rs373958816 Chr.16: 1776088-1776088 on Build GRCh38
rs773077546 Chr.16: 1775951-1775951 on Build GRCh38
rs776464907 Chr.16: 1776047-1776047 on Build GRCh38
rs376865399 Chr.16: 1776092-1776092 on Build GRCh38
rs139809312 Chr.16: 1776070-1776070 on Build GRCh38
rs531529878 Chr.16: 1776097-1776097 on Build GRCh38
rs771754192 Chr.16: 1775946-1775946 on Build GRCh38
rs201221323 Chr.16: 1776098-1776098 on Build GRCh38
rs746949623 Chr.16: 1776079-1776079 on Build GRCh38
rs376076255 Chr.16: 1776116-1776116 on Build GRCh38
rs138047300 Chr.16: 1775972-1775972 on Build GRCh38
rs764710854 Chr.16: 1776008-1776008 on Build GRCh38
rs143964421 Chr.16: 1775968-1775968 on Build GRCh38
rs538069713 Chr.16: 1775992-1775992 on Build GRCh38
rs756471725 Chr.16: 1776025-1776025 on Build GRCh38
rs150341255 Chr.16: 1775970-1775970 on Build GRCh38
rs761910064 Chr.16: 1775941-1775941 on Build GRCh38
rs371864567 Chr.16: 1776029-1776029 on Build GRCh38
rs746671635 Chr.16: 1776086-1776087 on Build GRCh38
rs776232995 Chr.16: 1776001-1776001 on Build GRCh38
rs754262901 Chr.16: 1776069-1776069 on Build GRCh38
rs770751090 Chr.16: 1776045-1776045 on Build GRCh38
rs543882375 Chr.16: 1775938-1775938 on Build GRCh38
rs777777467 Chr.16: 1776078-1776078 on Build GRCh38
rs528910143 Chr.16: 1776087-1776087 on Build GRCh38
rs779930109 Chr.16: 1775936-1775936 on Build GRCh38
rs978424213 Chr.16: 1775993-1775993 on Build GRCh38
rs766090627 Chr.16: 1775961-1775961 on Build GRCh38
rs201967594 Chr.16: 1775997-1775997 on Build GRCh38
rs952397000 Chr.16: 1775966-1775966 on Build GRCh38
rs775063615 Chr.16: 1776010-1776011 on Build GRCh38
rs200657371 Chr.16: 1776073-1776073 on Build GRCh38
rs760181032 Chr.16: 1775957-1775957 on Build GRCh38
rs563835971 Chr.16: 1776059-1776059 on Build GRCh38
rs779770797 Chr.16: 1776123-1776123 on Build GRCh38
rs773765005 Chr.16: 1776064-1776064 on Build GRCh38
rs748044681 Chr.16: 1775977-1775977 on Build GRCh38
rs891128727 Chr.16: 1775947-1775947 on Build GRCh38
rs371199330 Chr.16: 1776093-1776093 on Build GRCh38
rs185444041 Chr.16: 1776014-1776014 on Build GRCh38
rs746063686 Chr.16: 1775929-1775929 on Build GRCh38
rs772844873 Chr.16: 1775985-1775985 on Build GRCh38
rs769439253 Chr.16: 1776058-1776058 on Build GRCh38
rs762160902 Chr.16: 1776013-1776013 on Build GRCh38
rs149531255 Chr.16: 1775973-1775973 on Build GRCh38

View Full Product Details