Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10284480-10284741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM1
Entrez Gene ID
3383
Gene Name
intercellular adhesion molecule 1
Gene Aliases
BB2, CD54, P3.58
UniGene
Hs.643447
Gene Chromosome Location
Chr.19: 10270841-10286615 on Build GRCh38
Transcripts
NM_000201.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3891428 c.1063C>T p.Q355* Chr.19: 10284540-10284540 on Build GRCh38
5149908 c.1099C>T p.R367C Chr.19: 10284576-10284576 on Build GRCh38
990480 c.1147C>A p.H383N Chr.19: 10284624-10284624 on Build GRCh38
5183070 c.1096G>A p.G366R Chr.19: 10284573-10284573 on Build GRCh38
390794 c.1171C>T p.R391C Chr.19: 10284648-10284648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758965302 Chr.19: 10284592-10284592 on Build GRCh38
rs139178890 Chr.19: 10284576-10284576 on Build GRCh38
rs200879569 Chr.19: 10284667-10284667 on Build GRCh38
rs771711919 Chr.19: 10284649-10284649 on Build GRCh38
rs779307629 Chr.19: 10284510-10284510 on Build GRCh38
rs779022009 Chr.19: 10284706-10284706 on Build GRCh38
rs755044702 Chr.19: 10284676-10284676 on Build GRCh38
rs747223390 Chr.19: 10284514-10284514 on Build GRCh38
rs775039982 Chr.19: 10284516-10284536 on Build GRCh38
rs763423696 Chr.19: 10284666-10284666 on Build GRCh38
rs144226309 Chr.19: 10284648-10284648 on Build GRCh38
rs770551007 Chr.19: 10284561-10284561 on Build GRCh38
rs754190930 Chr.19: 10284506-10284506 on Build GRCh38
rs757208462 Chr.19: 10284702-10284702 on Build GRCh38
rs141326678 Chr.19: 10284499-10284499 on Build GRCh38
rs868375367 Chr.19: 10284542-10284542 on Build GRCh38
rs762182509 Chr.19: 10284650-10284650 on Build GRCh38
rs766445571 Chr.19: 10284606-10284606 on Build GRCh38
rs376622574 Chr.19: 10284612-10284612 on Build GRCh38
rs765509208 Chr.19: 10284656-10284656 on Build GRCh38
rs150014298 Chr.19: 10284608-10284608 on Build GRCh38
rs756424583 Chr.19: 10284682-10284682 on Build GRCh38
rs753734473 Chr.19: 10284700-10284700 on Build GRCh38
rs755868188 Chr.19: 10284615-10284615 on Build GRCh38
rs758480023 Chr.19: 10284512-10284512 on Build GRCh38
rs757757223 Chr.19: 10284509-10284509 on Build GRCh38
rs752847261 Chr.19: 10284679-10284679 on Build GRCh38
rs139290643 Chr.19: 10284584-10284584 on Build GRCh38
rs777679809 Chr.19: 10284625-10284625 on Build GRCh38
rs201383296 Chr.19: 10284533-10284533 on Build GRCh38
rs983667097 Chr.19: 10284526-10284526 on Build GRCh38
rs759319472 Chr.19: 10284500-10284500 on Build GRCh38
rs867627594 Chr.19: 10284717-10284717 on Build GRCh38
rs780195275 Chr.19: 10284513-10284513 on Build GRCh38
rs146000551 Chr.19: 10284577-10284577 on Build GRCh38
rs1038470076 Chr.19: 10284711-10284720 on Build GRCh38
rs200152703 Chr.19: 10284689-10284689 on Build GRCh38
rs751590636 Chr.19: 10284674-10284674 on Build GRCh38
rs370481619 Chr.19: 10284681-10284681 on Build GRCh38
rs2071441 Chr.19: 10284632-10284632 on Build GRCh38
rs199672020 Chr.19: 10284564-10284564 on Build GRCh38
rs773636684 Chr.19: 10284658-10284658 on Build GRCh38
rs181563357 Chr.19: 10284639-10284639 on Build GRCh38
rs755206701 Chr.19: 10284610-10284610 on Build GRCh38
rs767714183 Chr.19: 10284678-10284678 on Build GRCh38
rs781009982 Chr.19: 10284518-10284518 on Build GRCh38
rs765707240 Chr.19: 10284578-10284578 on Build GRCh38
rs1036180125 Chr.19: 10284622-10284622 on Build GRCh38
rs771319016 Chr.19: 10284515-10284535 on Build GRCh38
rs768938327 Chr.19: 10284516-10284516 on Build GRCh38
rs770432918 Chr.19: 10284640-10284640 on Build GRCh38
rs1027972658 Chr.19: 10284517-10284517 on Build GRCh38
rs900625084 Chr.19: 10284693-10284693 on Build GRCh38
rs370415118 Chr.19: 10284573-10284573 on Build GRCh38
rs775659075 Chr.19: 10284720-10284720 on Build GRCh38
rs13306429 Chr.19: 10284572-10284572 on Build GRCh38
rs1801714 Chr.19: 10284532-10284532 on Build GRCh38
rs377420632 Chr.19: 10284557-10284557 on Build GRCh38

View Full Product Details