Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26227037-26227304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF4B
Entrez Gene ID
6875
Gene Name
TATA-box binding protein associated factor 4b
Gene Aliases
SPGF13, TAF2C2, TAFII105
UniGene
Hs.369519
Gene Chromosome Location
Chr.18: 26226491-26391685 on Build GRCh38
Transcripts
NM_001293725.1 NM_005640.2 NR_121653.1 XM_005258339.3 XM_011526153.2 XM_017025932.1 XM_017025933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
289793 c.141G>T p.K47N Chr.18: 26227074-26227074 on Build GRCh38
4960214 c.235C>T p.P79S Chr.18: 26227168-26227168 on Build GRCh38
5761990 c.334_335insTT p.P113fs*7 Chr.18: 26227267-26227268 on Build GRCh38
3524676 c.238C>T p.P80S Chr.18: 26227171-26227171 on Build GRCh38
4543775 c.343G>A p.G115R Chr.18: 26227276-26227276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759189467 Chr.18: 26227169-26227169 on Build GRCh38
rs770783086 Chr.18: 26227232-26227232 on Build GRCh38
rs201019228 Chr.18: 26227265-26227265 on Build GRCh38
rs780195768 Chr.18: 26227209-26227209 on Build GRCh38
rs772871310 Chr.18: 26227079-26227079 on Build GRCh38
rs776977422 Chr.18: 26227068-26227068 on Build GRCh38
rs932705973 Chr.18: 26227145-26227145 on Build GRCh38
rs756317230 Chr.18: 26227212-26227212 on Build GRCh38
rs776603147 Chr.18: 26227168-26227168 on Build GRCh38
rs372215570 Chr.18: 26227198-26227198 on Build GRCh38
rs777929270 Chr.18: 26227057-26227057 on Build GRCh38
rs759288826 Chr.18: 26227093-26227093 on Build GRCh38
rs761399100 Chr.18: 26227188-26227189 on Build GRCh38
rs892697795 Chr.18: 26227157-26227157 on Build GRCh38
rs375315702 Chr.18: 26227161-26227161 on Build GRCh38
rs745916304 Chr.18: 26227071-26227071 on Build GRCh38
rs368327854 Chr.18: 26227162-26227162 on Build GRCh38
rs945275966 Chr.18: 26227065-26227065 on Build GRCh38
rs749429923 Chr.18: 26227144-26227144 on Build GRCh38
rs773873936 Chr.18: 26227177-26227179 on Build GRCh38
rs764475904 Chr.18: 26227195-26227195 on Build GRCh38
rs368485870 Chr.18: 26227215-26227215 on Build GRCh38
rs771042881 Chr.18: 26227066-26227066 on Build GRCh38
rs764992017 Chr.18: 26227172-26227172 on Build GRCh38
rs201210120 Chr.18: 26227275-26227275 on Build GRCh38
rs753580212 Chr.18: 26227087-26227087 on Build GRCh38
rs778884350 Chr.18: 26227226-26227226 on Build GRCh38
rs752239406 Chr.18: 26227186-26227186 on Build GRCh38
rs770714283 Chr.18: 26227163-26227163 on Build GRCh38
rs780051411 Chr.18: 26227143-26227143 on Build GRCh38
rs758915043 Chr.18: 26227109-26227109 on Build GRCh38
rs936529564 Chr.18: 26227091-26227091 on Build GRCh38
rs771901325 Chr.18: 26227153-26227153 on Build GRCh38
rs370628700 Chr.18: 26227097-26227097 on Build GRCh38
rs751863912 Chr.18: 26227124-26227124 on Build GRCh38
rs778400017 Chr.18: 26227120-26227120 on Build GRCh38
rs746921063 Chr.18: 26227227-26227227 on Build GRCh38
rs752255940 Chr.18: 26227101-26227101 on Build GRCh38
rs942404662 Chr.18: 26227069-26227069 on Build GRCh38
rs775578058 Chr.18: 26227076-26227076 on Build GRCh38
rs769909933 Chr.18: 26227075-26227075 on Build GRCh38
rs781247197 Chr.18: 26227136-26227136 on Build GRCh38
rs752156674 Chr.18: 26227204-26227204 on Build GRCh38
rs374723586 Chr.18: 26227239-26227239 on Build GRCh38
rs35709083 Chr.18: 26227103-26227104 on Build GRCh38
rs749338378 Chr.18: 26227218-26227218 on Build GRCh38
rs986737217 Chr.18: 26227252-26227252 on Build GRCh38
rs746040332 Chr.18: 26227137-26227137 on Build GRCh38
rs60311681 Chr.18: 26227080-26227080 on Build GRCh38
rs372334200 Chr.18: 26227190-26227190 on Build GRCh38
rs757726259 Chr.18: 26227206-26227206 on Build GRCh38
rs537350359 Chr.18: 26227125-26227125 on Build GRCh38
rs973861788 Chr.18: 26227219-26227219 on Build GRCh38
rs769568198 Chr.18: 26227259-26227259 on Build GRCh38
rs200194030 Chr.18: 26227220-26227220 on Build GRCh38
rs78262333 Chr.18: 26227063-26227063 on Build GRCh38
rs761673468 Chr.18: 26227078-26227078 on Build GRCh38
rs762349186 Chr.18: 26227273-26227273 on Build GRCh38

View Full Product Details