Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30532446-30532707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC14L6
Entrez Gene ID
730005
Gene Name
SEC14 like lipid binding 6
Gene Aliases
-
UniGene
Hs.505605
Gene Chromosome Location
Chr.22: 30522797-30546717 on Build GRCh38
Transcripts
NM_001193336.2 XM_011530357.2 XM_011530358.2 XM_011530360.2 XM_017028931.1 XM_017028932.1 XM_017028933.1 XM_017028934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5948990 c.423+1G>T p.? Chr.22: 30532524-30532524 on Build GRCh38
4989591 c.268G>A p.G90S Chr.22: 30532680-30532680 on Build GRCh38
7274387 c.384C>T p.C128C Chr.22: 30532564-30532564 on Build GRCh38
3842473 c.329G>T p.G110V Chr.22: 30532619-30532619 on Build GRCh38
5607614 c.276C>A p.D92E Chr.22: 30532672-30532672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763730535 Chr.22: 30532672-30532672 on Build GRCh38
rs978205334 Chr.22: 30532560-30532560 on Build GRCh38
rs751127034 Chr.22: 30532674-30532674 on Build GRCh38
rs747613072 Chr.22: 30532476-30532476 on Build GRCh38
rs529698329 Chr.22: 30532493-30532493 on Build GRCh38
rs1020046988 Chr.22: 30532551-30532551 on Build GRCh38
rs937316122 Chr.22: 30532678-30532678 on Build GRCh38
rs149479913 Chr.22: 30532643-30532643 on Build GRCh38
rs985350784 Chr.22: 30532580-30532580 on Build GRCh38
rs943833702 Chr.22: 30532619-30532619 on Build GRCh38
rs556339011 Chr.22: 30532484-30532484 on Build GRCh38
rs1041161965 Chr.22: 30532481-30532481 on Build GRCh38
rs755389496 Chr.22: 30532688-30532688 on Build GRCh38
rs772150274 Chr.22: 30532545-30532545 on Build GRCh38
rs767119945 Chr.22: 30532680-30532680 on Build GRCh38
rs776464574 Chr.22: 30532590-30532590 on Build GRCh38
rs1019303909 Chr.22: 30532596-30532596 on Build GRCh38
rs576175820 Chr.22: 30532487-30532487 on Build GRCh38
rs748211105 Chr.22: 30532535-30532535 on Build GRCh38
rs183903211 Chr.22: 30532632-30532632 on Build GRCh38
rs137955219 Chr.22: 30532633-30532633 on Build GRCh38
rs759316456 Chr.22: 30532607-30532607 on Build GRCh38
rs907972107 Chr.22: 30532670-30532670 on Build GRCh38
rs984887427 Chr.22: 30532657-30532657 on Build GRCh38
rs568733949 Chr.22: 30532572-30532572 on Build GRCh38
rs893008357 Chr.22: 30532516-30532516 on Build GRCh38
rs1055728423 Chr.22: 30532683-30532683 on Build GRCh38
rs1008318910 Chr.22: 30532564-30532564 on Build GRCh38
rs773354534 Chr.22: 30532550-30532550 on Build GRCh38
rs940692140 Chr.22: 30532468-30532468 on Build GRCh38
rs1048157735 Chr.22: 30532663-30532663 on Build GRCh38
rs532725817 Chr.22: 30532671-30532671 on Build GRCh38
rs560957647 Chr.22: 30532563-30532563 on Build GRCh38
rs191426209 Chr.22: 30532490-30532490 on Build GRCh38
rs565164447 Chr.22: 30532492-30532492 on Build GRCh38
rs1029195295 Chr.22: 30532635-30532635 on Build GRCh38
rs966816868 Chr.22: 30532558-30532558 on Build GRCh38
rs779459989 Chr.22: 30532523-30532523 on Build GRCh38
rs1055702009 Chr.22: 30532501-30532501 on Build GRCh38
rs540993479 Chr.22: 30532555-30532555 on Build GRCh38
rs761508538 Chr.22: 30532675-30532675 on Build GRCh38
rs754221381 Chr.22: 30532681-30532681 on Build GRCh38

View Full Product Details