Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28987674-28987933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
UniGene
Hs.175437
Gene Chromosome Location
Chr.1: 28887091-29120046 on Build GRCh38
Transcripts
NM_001166005.1 NM_001166006.1 NM_001166007.1 NM_004437.3 NM_203342.2 NM_203343.2 XM_005245753.1 XM_005245757.1 XM_005245760.1 XM_005245761.1 XM_005245763.1 XM_005245764.1 XM_005245765.1 XM_005245768.1 XM_005245769.1 XM_005245770.1 XM_005245772.4 XM_005245773.4 XM_005245774.1 XM_006710434.1 XM_006710439.1 XM_011540956.1 XM_011540957.1 XM_011540958.1 XM_011540959.1 XM_011540960.1 XM_011540961.1 XM_011540962.1 XM_011540963.2 XM_011540964.1 XM_011540965.2 XM_017000581.1 XM_017000582.1 XM_017000583.1 XM_017000584.1 XM_017000585.1 XM_017000586.1 XM_017000587.1 XM_017000588.1 XM_017000589.1 XM_017000590.1 XM_017000591.1 XM_017000592.1 XM_017000593.1 XM_017000594.1 XM_017000595.1 XM_017000596.1 XM_017000597.1 XM_017000598.1 XM_017000599.1 XM_017000600.1 XM_017000601.1 XM_017000602.1 XM_017000603.1 XM_017000604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1251239 c.261C>T p.L87L Chr.1: 28987698-28987698 on Build GRCh38
6654561 c.285C>A p.S95S Chr.1: 28987722-28987722 on Build GRCh38
6391477 c.418A>G p.K140E Chr.1: 28987855-28987855 on Build GRCh38
6024690 c.378C>A p.I126I Chr.1: 28987815-28987815 on Build GRCh38
4409667 c.241C>T p.R81* Chr.1: 28987678-28987678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774634104 Chr.1: 28987883-28987883 on Build GRCh38
rs1046166710 Chr.1: 28987905-28987905 on Build GRCh38
rs371403877 Chr.1: 28987814-28987814 on Build GRCh38
rs985496985 Chr.1: 28987812-28987812 on Build GRCh38
rs201345838 Chr.1: 28987775-28987775 on Build GRCh38
rs200186018 Chr.1: 28987847-28987847 on Build GRCh38
rs755507695 Chr.1: 28987848-28987848 on Build GRCh38
rs929710377 Chr.1: 28987908-28987908 on Build GRCh38
rs759883733 Chr.1: 28987890-28987890 on Build GRCh38
rs766702018 Chr.1: 28987715-28987715 on Build GRCh38
rs897659215 Chr.1: 28987889-28987889 on Build GRCh38
rs752920448 Chr.1: 28987912-28987912 on Build GRCh38
rs745437081 Chr.1: 28987858-28987858 on Build GRCh38
rs752730549 Chr.1: 28987732-28987732 on Build GRCh38
rs781775828 Chr.1: 28987906-28987907 on Build GRCh38
rs866162009 Chr.1: 28987905-28987906 on Build GRCh38
rs968524834 Chr.1: 28987743-28987743 on Build GRCh38
rs781461033 Chr.1: 28987723-28987723 on Build GRCh38
rs755218198 Chr.1: 28987722-28987722 on Build GRCh38
rs769244959 Chr.1: 28987849-28987849 on Build GRCh38
rs768207973 Chr.1: 28987821-28987821 on Build GRCh38
rs766900734 Chr.1: 28987874-28987874 on Build GRCh38
rs776279425 Chr.1: 28987831-28987831 on Build GRCh38
rs966891385 Chr.1: 28987698-28987698 on Build GRCh38
rs979954102 Chr.1: 28987749-28987749 on Build GRCh38
rs560164744 Chr.1: 28987761-28987761 on Build GRCh38
rs763330269 Chr.1: 28987712-28987712 on Build GRCh38
rs140065972 Chr.1: 28987790-28987790 on Build GRCh38
rs772686614 Chr.1: 28987852-28987852 on Build GRCh38
rs751918017 Chr.1: 28987717-28987717 on Build GRCh38
rs992883620 Chr.1: 28987796-28987796 on Build GRCh38
rs369294048 Chr.1: 28987772-28987772 on Build GRCh38
rs369976028 Chr.1: 28987859-28987859 on Build GRCh38
rs767667572 Chr.1: 28987902-28987902 on Build GRCh38
rs779019847 Chr.1: 28987771-28987771 on Build GRCh38
rs141680585 Chr.1: 28987700-28987700 on Build GRCh38
rs929146973 Chr.1: 28987897-28987897 on Build GRCh38
rs368670866 Chr.1: 28987848-28987848 on Build GRCh38

View Full Product Details