Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31535354-31535581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G2-DDX39B
Entrez Gene ID
100532737
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31546848 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Gene Symbol
DDX39B
Entrez Gene ID
7919
Gene Name
DEAD-box helicase 39B
Gene Aliases
BAT1, D6S81E, UAP56
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31542475 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624551 c.540C>T p.F180F Chr.6: 31535421-31535421 on Build GRCh38
3624552 c.654C>T p.T218T Chr.6: 31535448-31535448 on Build GRCh38
3702977 c.684A>G p.E228E Chr.6: 31535463-31535463 on Build GRCh38
3624549 c.681C>T p.F227F Chr.6: 31535421-31535421 on Build GRCh38
3624554 c.513C>T p.T171T Chr.6: 31535448-31535448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544979742 Chr.6: 31535541-31535541 on Build GRCh38
rs775080061 Chr.6: 31535522-31535522 on Build GRCh38
rs183952677 Chr.6: 31535552-31535552 on Build GRCh38
rs746746089 Chr.6: 31535481-31535481 on Build GRCh38
rs267600954 Chr.6: 31535456-31535456 on Build GRCh38
rs777506169 Chr.6: 31535492-31535492 on Build GRCh38
rs556229987 Chr.6: 31535490-31535490 on Build GRCh38
rs773279301 Chr.6: 31535490-31535492 on Build GRCh38
rs55799418 Chr.6: 31535442-31535442 on Build GRCh38
rs968346579 Chr.6: 31535538-31535538 on Build GRCh38
rs113533670 Chr.6: 31535487-31535487 on Build GRCh38
rs943788740 Chr.6: 31535535-31535535 on Build GRCh38
rs989722483 Chr.6: 31535388-31535388 on Build GRCh38
rs745507081 Chr.6: 31535498-31535498 on Build GRCh38
rs777773721 Chr.6: 31535469-31535469 on Build GRCh38
rs760736319 Chr.6: 31535528-31535554 on Build GRCh38
rs560131473 Chr.6: 31535550-31535550 on Build GRCh38
rs758214637 Chr.6: 31535446-31535446 on Build GRCh38
rs1021311483 Chr.6: 31535527-31535527 on Build GRCh38
rs191731842 Chr.6: 31535507-31535507 on Build GRCh38
rs769002730 Chr.6: 31535533-31535533 on Build GRCh38
rs780105836 Chr.6: 31535412-31535412 on Build GRCh38
rs34885180 Chr.6: 31535556-31535557 on Build GRCh38
rs986272591 Chr.6: 31535510-31535510 on Build GRCh38

View Full Product Details