Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17561251-17561508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXADR
Entrez Gene ID
1525
Gene Name
coxsackie virus and adenovirus receptor
Gene Aliases
CAR, CAR4/6, HCAR
UniGene
Hs.627078
Gene Chromosome Location
Chr.21: 17512906-17636213 on Build GRCh38
Transcripts
NM_001207063.1 NM_001207064.1 NM_001207065.1 NM_001207066.1 NM_001338.4 XM_011529476.2 XM_011529477.2 XM_011529478.2 XM_011529479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1713857 c.822T>A p.H274Q Chr.21: 17561465-17561465 on Build GRCh38
5392721 c.814G>A p.E272K Chr.21: 17561457-17561457 on Build GRCh38
6587429 c.770T>A p.I257N Chr.21: 17561413-17561413 on Build GRCh38
1713856 c.814G>A p.E272K Chr.21: 17561457-17561457 on Build GRCh38
3764905 c.753G>A p.A251A Chr.21: 17561396-17561396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912371525 Chr.21: 17561365-17561365 on Build GRCh38
rs770704671 Chr.21: 17561373-17561373 on Build GRCh38
rs908332571 Chr.21: 17561288-17561288 on Build GRCh38
rs766509347 Chr.21: 17561299-17561299 on Build GRCh38
rs148881358 Chr.21: 17561395-17561395 on Build GRCh38
rs1047358778 Chr.21: 17561338-17561338 on Build GRCh38
rs759569781 Chr.21: 17561397-17561397 on Build GRCh38
rs756589633 Chr.21: 17561343-17561343 on Build GRCh38
rs533643575 Chr.21: 17561410-17561410 on Build GRCh38
rs141988381 Chr.21: 17561308-17561308 on Build GRCh38
rs778349868 Chr.21: 17561428-17561428 on Build GRCh38
rs760582039 Chr.21: 17561377-17561377 on Build GRCh38
rs1014454826 Chr.21: 17561400-17561400 on Build GRCh38
rs537375317 Chr.21: 17561376-17561376 on Build GRCh38
rs756679498 Chr.21: 17561424-17561424 on Build GRCh38
rs781568544 Chr.21: 17561309-17561309 on Build GRCh38
rs771570606 Chr.21: 17561375-17561375 on Build GRCh38
rs757637973 Chr.21: 17561433-17561433 on Build GRCh38
rs200360457 Chr.21: 17561393-17561393 on Build GRCh38
rs763147801 Chr.21: 17561298-17561298 on Build GRCh38
rs754261158 Chr.21: 17561432-17561432 on Build GRCh38
rs767607873 Chr.21: 17561406-17561406 on Build GRCh38
rs778263762 Chr.21: 17561344-17561344 on Build GRCh38
rs778453029 Chr.21: 17561434-17561434 on Build GRCh38
rs751593551 Chr.21: 17561304-17561304 on Build GRCh38
rs899509385 Chr.21: 17561425-17561425 on Build GRCh38
rs778561658 Chr.21: 17561374-17561374 on Build GRCh38
rs186473514 Chr.21: 17561306-17561306 on Build GRCh38
rs137939398 Chr.21: 17561477-17561477 on Build GRCh38
rs779083564 Chr.21: 17561308-17561309 on Build GRCh38
rs776403577 Chr.21: 17561379-17561379 on Build GRCh38
rs201624272 Chr.21: 17561391-17561391 on Build GRCh38
rs372788152 Chr.21: 17561469-17561469 on Build GRCh38
rs772388424 Chr.21: 17561323-17561325 on Build GRCh38
rs748426780 Chr.21: 17561318-17561321 on Build GRCh38
rs751752573 Chr.21: 17561396-17561396 on Build GRCh38
rs749007607 Chr.21: 17561362-17561362 on Build GRCh38
rs779600277 Chr.21: 17561468-17561468 on Build GRCh38
rs143478011 Chr.21: 17561436-17561436 on Build GRCh38
rs768582795 Chr.21: 17561472-17561472 on Build GRCh38
rs757895680 Chr.21: 17561448-17561448 on Build GRCh38
rs750692034 Chr.21: 17561292-17561292 on Build GRCh38
rs150493172 Chr.21: 17561318-17561318 on Build GRCh38

View Full Product Details