Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42117499-42117764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT2A
Entrez Gene ID
2648
Gene Name
lysine acetyltransferase 2A
Gene Aliases
GCN5, GCN5L2, PCAF-b, hGCN5
UniGene
Hs.463045
Gene Chromosome Location
Chr.17: 42113111-42121393 on Build GRCh38
Transcripts
NM_021078.2 XM_006721817.3 XM_006721818.3 XM_017024448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
979419 c.1442C>T p.S481L Chr.17: 42117583-42117583 on Build GRCh38
5055482 c.1453G>A p.A485T Chr.17: 42117572-42117572 on Build GRCh38
979417 c.1486C>T p.R496C Chr.17: 42117539-42117539 on Build GRCh38
3989031 c.1363C>G p.P455A Chr.17: 42117743-42117743 on Build GRCh38
3517615 c.1471C>T p.R491C Chr.17: 42117554-42117554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369510636 Chr.17: 42117698-42117698 on Build GRCh38
rs948140872 Chr.17: 42117690-42117690 on Build GRCh38
rs782142091 Chr.17: 42117608-42117608 on Build GRCh38
rs782350177 Chr.17: 42117704-42117704 on Build GRCh38
rs782373861 Chr.17: 42117577-42117577 on Build GRCh38
rs782651040 Chr.17: 42117554-42117554 on Build GRCh38
rs782092850 Chr.17: 42117612-42117612 on Build GRCh38
rs782570466 Chr.17: 42117689-42117689 on Build GRCh38
rs369605791 Chr.17: 42117619-42117619 on Build GRCh38
rs782253780 Chr.17: 42117731-42117731 on Build GRCh38
rs782360789 Chr.17: 42117573-42117573 on Build GRCh38
rs782433159 Chr.17: 42117631-42117631 on Build GRCh38
rs782603888 Chr.17: 42117574-42117574 on Build GRCh38
rs199558402 Chr.17: 42117621-42117621 on Build GRCh38
rs371666045 Chr.17: 42117614-42117614 on Build GRCh38
rs782412917 Chr.17: 42117594-42117594 on Build GRCh38
rs781914542 Chr.17: 42117611-42117611 on Build GRCh38
rs561719000 Chr.17: 42117622-42117622 on Build GRCh38
rs782790918 Chr.17: 42117624-42117624 on Build GRCh38
rs781829541 Chr.17: 42117646-42117646 on Build GRCh38
rs573714349 Chr.17: 42117663-42117663 on Build GRCh38
rs782527099 Chr.17: 42117546-42117546 on Build GRCh38
rs782691157 Chr.17: 42117533-42117533 on Build GRCh38
rs553979667 Chr.17: 42117609-42117609 on Build GRCh38
rs781906152 Chr.17: 42117555-42117555 on Build GRCh38
rs782522405 Chr.17: 42117661-42117661 on Build GRCh38
rs782625819 Chr.17: 42117644-42117644 on Build GRCh38
rs377719705 Chr.17: 42117726-42117726 on Build GRCh38
rs782725616 Chr.17: 42117639-42117640 on Build GRCh38
rs781881327 Chr.17: 42117522-42117522 on Build GRCh38
rs373945603 Chr.17: 42117717-42117717 on Build GRCh38
rs538458369 Chr.17: 42117568-42117568 on Build GRCh38
rs781810893 Chr.17: 42117628-42117628 on Build GRCh38
rs782800781 Chr.17: 42117521-42117521 on Build GRCh38
rs782262277 Chr.17: 42117575-42117575 on Build GRCh38
rs373511745 Chr.17: 42117623-42117623 on Build GRCh38
rs896313763 Chr.17: 42117675-42117675 on Build GRCh38
rs941043389 Chr.17: 42117616-42117616 on Build GRCh38
rs782063917 Chr.17: 42117527-42117527 on Build GRCh38
rs577725633 Chr.17: 42117537-42117537 on Build GRCh38
rs782292863 Chr.17: 42117666-42117666 on Build GRCh38
rs903845358 Chr.17: 42117727-42117727 on Build GRCh38
rs985616741 Chr.17: 42117567-42117567 on Build GRCh38
rs782186147 Chr.17: 42117570-42117570 on Build GRCh38
rs142273465 Chr.17: 42117538-42117538 on Build GRCh38

View Full Product Details