Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57884235-57884496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB1
Entrez Gene ID
1258
Gene Name
cyclic nucleotide gated channel beta 1
Gene Aliases
CNCG2, CNCG3L, CNCG4, CNG4, CNGB1B, GAR1, GARP, GARP2, RCNC2, RCNCb, RCNCbeta, RP45
UniGene
Hs.147062
Gene Chromosome Location
Chr.16: 57882340-57971116 on Build GRCh38
Transcripts
NM_001286130.1 NM_001297.4 XM_011522870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3794942 c.3649G>A p.E1217K Chr.16: 57884271-57884271 on Build GRCh38
3969611 c.3543C>T p.T1181T Chr.16: 57884377-57884377 on Build GRCh38
5606438 c.3557C>T p.P1186L Chr.16: 57884363-57884363 on Build GRCh38
1378606 c.3538G>A p.A1180T Chr.16: 57884382-57884382 on Build GRCh38
5470542 c.3671G>T p.R1224M Chr.16: 57884249-57884249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777912112 Chr.16: 57884349-57884349 on Build GRCh38
rs761341606 Chr.16: 57884309-57884309 on Build GRCh38
rs529956339 Chr.16: 57884403-57884403 on Build GRCh38
rs78154829 Chr.16: 57884275-57884275 on Build GRCh38
rs866203302 Chr.16: 57884291-57884291 on Build GRCh38
rs773308978 Chr.16: 57884265-57884265 on Build GRCh38
rs561289103 Chr.16: 57884331-57884331 on Build GRCh38
rs190049030 Chr.16: 57884474-57884474 on Build GRCh38
rs765506955 Chr.16: 57884307-57884307 on Build GRCh38
rs778536843 Chr.16: 57884341-57884341 on Build GRCh38
rs564918974 Chr.16: 57884361-57884361 on Build GRCh38
rs770663676 Chr.16: 57884435-57884435 on Build GRCh38
rs760012294 Chr.16: 57884302-57884302 on Build GRCh38
rs771439069 Chr.16: 57884266-57884266 on Build GRCh38
rs776024444 Chr.16: 57884437-57884437 on Build GRCh38
rs752493968 Chr.16: 57884344-57884344 on Build GRCh38
rs547549398 Chr.16: 57884368-57884368 on Build GRCh38
rs754864561 Chr.16: 57884338-57884338 on Build GRCh38
rs886052191 Chr.16: 57884281-57884281 on Build GRCh38
rs200169932 Chr.16: 57884427-57884427 on Build GRCh38
rs367858527 Chr.16: 57884332-57884332 on Build GRCh38
rs767684787 Chr.16: 57884477-57884477 on Build GRCh38
rs770227227 Chr.16: 57884414-57884414 on Build GRCh38
rs781217584 Chr.16: 57884407-57884407 on Build GRCh38
rs868830996 Chr.16: 57884289-57884289 on Build GRCh38
rs763846389 Chr.16: 57884465-57884465 on Build GRCh38
rs752446297 Chr.16: 57884395-57884395 on Build GRCh38
rs753863333 Chr.16: 57884382-57884382 on Build GRCh38
rs201186180 Chr.16: 57884336-57884336 on Build GRCh38
rs751766451 Chr.16: 57884405-57884405 on Build GRCh38
rs749621708 Chr.16: 57884422-57884422 on Build GRCh38
rs749978261 Chr.16: 57884330-57884330 on Build GRCh38
rs375729452 Chr.16: 57884446-57884446 on Build GRCh38
rs1009948267 Chr.16: 57884350-57884350 on Build GRCh38
rs759022981 Chr.16: 57884439-57884439 on Build GRCh38
rs772230733 Chr.16: 57884256-57884256 on Build GRCh38
rs745921689 Chr.16: 57884411-57884411 on Build GRCh38
rs773233624 Chr.16: 57884474-57884475 on Build GRCh38
rs866734925 Chr.16: 57884288-57884288 on Build GRCh38
rs758569244 Chr.16: 57884346-57884346 on Build GRCh38
rs375919369 Chr.16: 57884377-57884377 on Build GRCh38
rs7190372 Chr.16: 57884364-57884364 on Build GRCh38
rs543712958 Chr.16: 57884360-57884360 on Build GRCh38
rs186853874 Chr.16: 57884432-57884432 on Build GRCh38
rs752667944 Chr.16: 57884447-57884447 on Build GRCh38
rs1132032 Chr.16: 57884297-57884297 on Build GRCh38
rs769866157 Chr.16: 57884349-57884350 on Build GRCh38
rs747094811 Chr.16: 57884356-57884356 on Build GRCh38
rs763330590 Chr.16: 57884367-57884367 on Build GRCh38
rs775750549 Chr.16: 57884365-57884365 on Build GRCh38
rs758143647 Chr.16: 57884399-57884399 on Build GRCh38
rs376024334 Chr.16: 57884440-57884440 on Build GRCh38
rs767160156 Chr.16: 57884324-57884324 on Build GRCh38
rs765169492 Chr.16: 57884394-57884394 on Build GRCh38
rs933868691 Chr.16: 57884296-57884296 on Build GRCh38
rs780383898 Chr.16: 57884343-57884343 on Build GRCh38
rs777055578 Chr.16: 57884286-57884286 on Build GRCh38
rs1056514077 Chr.16: 57884271-57884271 on Build GRCh38
rs73562944 Chr.16: 57884393-57884393 on Build GRCh38
rs760331816 Chr.16: 57884374-57884374 on Build GRCh38
rs774103056 Chr.16: 57884308-57884308 on Build GRCh38
rs780554214 Chr.16: 57884415-57884415 on Build GRCh38
rs868226949 Chr.16: 57884376-57884376 on Build GRCh38
rs769142173 Chr.16: 57884424-57884424 on Build GRCh38

View Full Product Details