Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79970315-79970568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSBP1L1
Entrez Gene ID
440498
Gene Name
heat shock factor binding protein 1 like 1
Gene Aliases
-
UniGene
Hs.191582
Gene Chromosome Location
Chr.18: 79964582-79970822 on Build GRCh38
Transcripts
NM_001136180.1

SNP Detail

SNP ID Chromosome Location
rs560324030 Chr.18: 79970457-79970457 on Build GRCh38
rs367795289 Chr.18: 79970353-79970353 on Build GRCh38
rs541201956 Chr.18: 79970361-79970361 on Build GRCh38
rs371874697 Chr.18: 79970430-79970430 on Build GRCh38
rs756187255 Chr.18: 79970477-79970477 on Build GRCh38
rs142821156 Chr.18: 79970500-79970500 on Build GRCh38
rs907888018 Chr.18: 79970506-79970506 on Build GRCh38
rs765733029 Chr.18: 79970397-79970397 on Build GRCh38
rs376590127 Chr.18: 79970447-79970447 on Build GRCh38
rs368829105 Chr.18: 79970543-79970543 on Build GRCh38
rs371695008 Chr.18: 79970384-79970384 on Build GRCh38
rs951554708 Chr.18: 79970362-79970362 on Build GRCh38
rs928376129 Chr.18: 79970519-79970519 on Build GRCh38
rs559389966 Chr.18: 79970373-79970373 on Build GRCh38
rs116269936 Chr.18: 79970460-79970460 on Build GRCh38
rs900336632 Chr.18: 79970525-79970525 on Build GRCh38
rs780798081 Chr.18: 79970423-79970424 on Build GRCh38
rs954401822 Chr.18: 79970394-79970394 on Build GRCh38
rs779982143 Chr.18: 79970491-79970491 on Build GRCh38
rs925405698 Chr.18: 79970435-79970435 on Build GRCh38
rs959559167 Chr.18: 79970437-79970437 on Build GRCh38
rs942080051 Chr.18: 79970508-79970508 on Build GRCh38
rs564051300 Chr.18: 79970411-79970411 on Build GRCh38
rs567996271 Chr.18: 79970543-79970543 on Build GRCh38
rs937189554 Chr.18: 79970526-79970526 on Build GRCh38
rs371714498 Chr.18: 79970539-79970539 on Build GRCh38
rs945702224 Chr.18: 79970496-79970496 on Build GRCh38
rs1037757966 Chr.18: 79970523-79970523 on Build GRCh38
rs118028499 Chr.18: 79970410-79970410 on Build GRCh38
rs1045481464 Chr.18: 79970509-79970509 on Build GRCh38
rs753140521 Chr.18: 79970404-79970404 on Build GRCh38
rs996005596 Chr.18: 79970531-79970531 on Build GRCh38
rs117741599 Chr.18: 79970544-79970544 on Build GRCh38
rs749143458 Chr.18: 79970498-79970498 on Build GRCh38
rs758492045 Chr.18: 79970407-79970407 on Build GRCh38

View Full Product Details