Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34097143-34097394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS6
Entrez Gene ID
64968
Gene Name
mitochondrial ribosomal protein S6
Gene Aliases
C21orf101, MRP-S6, RPMS6, S6mt
UniGene
Hs.302742
Gene Chromosome Location
Chr.21: 34073523-34143034 on Build GRCh38
Transcripts
NM_032476.3 NM_006933.6

Gene Symbol
SLC5A3
Entrez Gene ID
6526
Gene Name
solute carrier family 5 member 3
Gene Aliases
BCW2, SMIT, SMIT1, SMIT2
UniGene
Hs.302742
Gene Chromosome Location
Chr.21: 34073523-34106262 on Build GRCh38
Transcripts
NM_032476.3 NM_006933.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4853217 c.1971C>G p.F657L Chr.21: 34097169-34097169 on Build GRCh38
252826 c.2075G>A p.R692Q Chr.21: 34097273-34097273 on Build GRCh38
5572784 c.1961A>C p.Y654S Chr.21: 34097159-34097159 on Build GRCh38
5366246 c.2155T>A p.*719R Chr.21: 34097353-34097353 on Build GRCh38
3379174 c.1947T>C p.D649D Chr.21: 34097145-34097145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769963063 Chr.21: 34097256-34097256 on Build GRCh38
rs753374778 Chr.21: 34097294-34097294 on Build GRCh38
rs752139372 Chr.21: 34097302-34097302 on Build GRCh38
rs551314834 Chr.21: 34097265-34097265 on Build GRCh38
rs759259884 Chr.21: 34097274-34097274 on Build GRCh38
rs775198034 Chr.21: 34097276-34097276 on Build GRCh38
rs555822620 Chr.21: 34097278-34097278 on Build GRCh38
rs760343616 Chr.21: 34097283-34097283 on Build GRCh38
rs752399933 Chr.21: 34097165-34097165 on Build GRCh38
rs758968680 Chr.21: 34097205-34097205 on Build GRCh38
rs369745240 Chr.21: 34097275-34097275 on Build GRCh38
rs755529407 Chr.21: 34097171-34097171 on Build GRCh38
rs368773190 Chr.21: 34097167-34097167 on Build GRCh38
rs376446722 Chr.21: 34097238-34097238 on Build GRCh38
rs763637483 Chr.21: 34097198-34097198 on Build GRCh38
rs755394502 Chr.21: 34097216-34097216 on Build GRCh38
rs763718792 Chr.21: 34097287-34097287 on Build GRCh38
rs780761515 Chr.21: 34097208-34097208 on Build GRCh38
rs138321081 Chr.21: 34097253-34097253 on Build GRCh38
rs781226805 Chr.21: 34097231-34097231 on Build GRCh38
rs773313197 Chr.21: 34097262-34097262 on Build GRCh38
rs958804216 Chr.21: 34097281-34097281 on Build GRCh38
rs770843858 Chr.21: 34097272-34097272 on Build GRCh38
rs957735388 Chr.21: 34097213-34097213 on Build GRCh38
rs142721463 Chr.21: 34097328-34097328 on Build GRCh38
rs1009845624 Chr.21: 34097248-34097248 on Build GRCh38
rs756837280 Chr.21: 34097296-34097296 on Build GRCh38
rs746485136 Chr.21: 34097348-34097349 on Build GRCh38
rs766924510 Chr.21: 34097297-34097297 on Build GRCh38
rs781602182 Chr.21: 34097363-34097363 on Build GRCh38
rs753327329 Chr.21: 34097203-34097203 on Build GRCh38
rs777616705 Chr.21: 34097244-34097247 on Build GRCh38
rs755341560 Chr.21: 34097306-34097306 on Build GRCh38
rs774287970 Chr.21: 34097273-34097273 on Build GRCh38
rs747317104 Chr.21: 34097214-34097214 on Build GRCh38

View Full Product Details