Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 48007098-48007371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUN3
Entrez Gene ID
256979
Gene Name
Sad1 and UNC84 domain containing 3
Gene Aliases
SUNC1
UniGene
Hs.406741
Gene Chromosome Location
Chr.7: 47987148-48035899 on Build GRCh38
Transcripts
NM_001030019.1 NM_001284350.1 NM_152782.3 XM_017011930.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5961346 c.392G>T p.G131V Chr.7: 48007229-48007229 on Build GRCh38
5447765 c.398T>C p.L133P Chr.7: 48007259-48007259 on Build GRCh38
1090206 c.362G>A p.R121H Chr.7: 48007295-48007295 on Build GRCh38
348118 c.382G>T p.E128* Chr.7: 48007275-48007275 on Build GRCh38
5928987 c.379G>A p.D127N Chr.7: 48007242-48007242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868188521 Chr.7: 48007182-48007182 on Build GRCh38
rs763409732 Chr.7: 48007286-48007286 on Build GRCh38
rs747059775 Chr.7: 48007246-48007246 on Build GRCh38
rs186734784 Chr.7: 48007231-48007231 on Build GRCh38
rs1034112472 Chr.7: 48007233-48007233 on Build GRCh38
rs916742074 Chr.7: 48007127-48007127 on Build GRCh38
rs759642395 Chr.7: 48007285-48007286 on Build GRCh38
rs574552304 Chr.7: 48007336-48007336 on Build GRCh38
rs948410066 Chr.7: 48007293-48007293 on Build GRCh38
rs758677844 Chr.7: 48007337-48007338 on Build GRCh38
rs776910129 Chr.7: 48007268-48007268 on Build GRCh38
rs754500828 Chr.7: 48007201-48007201 on Build GRCh38
rs752378125 Chr.7: 48007211-48007211 on Build GRCh38
rs150352113 Chr.7: 48007177-48007177 on Build GRCh38
rs770141892 Chr.7: 48007276-48007276 on Build GRCh38
rs761259164 Chr.7: 48007202-48007203 on Build GRCh38
rs542421917 Chr.7: 48007238-48007238 on Build GRCh38
rs766732610 Chr.7: 48007249-48007250 on Build GRCh38
rs75522639 Chr.7: 48007134-48007134 on Build GRCh38
rs115827032 Chr.7: 48007178-48007178 on Build GRCh38
rs753032576 Chr.7: 48007290-48007291 on Build GRCh38
rs755805894 Chr.7: 48007226-48007226 on Build GRCh38
rs777506868 Chr.7: 48007144-48007144 on Build GRCh38
rs1017687830 Chr.7: 48007143-48007143 on Build GRCh38
rs778725412 Chr.7: 48007316-48007316 on Build GRCh38
rs763655544 Chr.7: 48007296-48007296 on Build GRCh38
rs537944642 Chr.7: 48007320-48007320 on Build GRCh38
rs754516906 Chr.7: 48007123-48007123 on Build GRCh38
rs145833872 Chr.7: 48007275-48007275 on Build GRCh38
rs556391462 Chr.7: 48007234-48007234 on Build GRCh38
rs372507142 Chr.7: 48007180-48007180 on Build GRCh38
rs753642528 Chr.7: 48007228-48007228 on Build GRCh38
rs762227525 Chr.7: 48007192-48007192 on Build GRCh38
rs368996415 Chr.7: 48007145-48007145 on Build GRCh38
rs75207929 Chr.7: 48007199-48007199 on Build GRCh38
rs780318172 Chr.7: 48007240-48007240 on Build GRCh38
rs940317870 Chr.7: 48007210-48007210 on Build GRCh38
rs562292083 Chr.7: 48007295-48007295 on Build GRCh38
rs765732753 Chr.7: 48007194-48007194 on Build GRCh38
rs766965264 Chr.7: 48007203-48007203 on Build GRCh38
rs906902510 Chr.7: 48007214-48007214 on Build GRCh38
rs780830682 Chr.7: 48007128-48007128 on Build GRCh38
rs1574242 Chr.7: 48007117-48007117 on Build GRCh38
rs754005799 Chr.7: 48007284-48007285 on Build GRCh38
rs553383777 Chr.7: 48007227-48007227 on Build GRCh38
rs754397235 Chr.7: 48007115-48007115 on Build GRCh38
rs769060684 Chr.7: 48007253-48007253 on Build GRCh38
rs369223693 Chr.7: 48007161-48007161 on Build GRCh38
rs753472883 Chr.7: 48007301-48007301 on Build GRCh38
rs776906820 Chr.7: 48007189-48007189 on Build GRCh38
rs376354129 Chr.7: 48007330-48007330 on Build GRCh38
rs757742306 Chr.7: 48007116-48007116 on Build GRCh38
rs765388747 Chr.7: 48007287-48007288 on Build GRCh38
rs757023687 Chr.7: 48007306-48007306 on Build GRCh38
rs982171583 Chr.7: 48007142-48007142 on Build GRCh38
rs750245825 Chr.7: 48007184-48007184 on Build GRCh38
rs17852360 Chr.7: 48007278-48007278 on Build GRCh38
rs145043926 Chr.7: 48007174-48007174 on Build GRCh38
rs751090481 Chr.7: 48007118-48007118 on Build GRCh38
rs745939558 Chr.7: 48007165-48007165 on Build GRCh38
rs948241396 Chr.7: 48007132-48007132 on Build GRCh38
rs747155704 Chr.7: 48007341-48007341 on Build GRCh38
rs752255194 Chr.7: 48007287-48007287 on Build GRCh38
rs747742642 Chr.7: 48007133-48007133 on Build GRCh38
rs866078716 Chr.7: 48007196-48007196 on Build GRCh38

View Full Product Details