Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9930270-9930531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LZIC
Entrez Gene ID
84328
Gene Name
leucine zipper and CTNNBIP1 domain containing
Gene Aliases
-
UniGene
Hs.327252
Gene Chromosome Location
Chr.1: 9922113-9943427 on Build GRCh38
Transcripts
NM_001316973.1 NM_001316974.1 NM_001316975.1 NM_001316976.1 NM_032368.4 XM_005263506.2 XM_005263509.2 XM_017002546.1 XM_017002547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889170 c.515G>T p.G172V Chr.1: 9930457-9930457 on Build GRCh38
6490645 c.515-3delT p.? Chr.1: 9930460-9930460 on Build GRCh38
5953951 c.619G>C p.E207Q Chr.1: 9930416-9930416 on Build GRCh38
6705429 c.633delA p.K211fs? Chr.1: 9930402-9930402 on Build GRCh38
5953950 c.556G>C p.E186Q Chr.1: 9930416-9930416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766417125 Chr.1: 9930468-9930468 on Build GRCh38
rs776067258 Chr.1: 9930505-9930505 on Build GRCh38
rs755460957 Chr.1: 9930368-9930369 on Build GRCh38
rs866524214 Chr.1: 9930431-9930431 on Build GRCh38
rs753939939 Chr.1: 9930471-9930471 on Build GRCh38
rs750567681 Chr.1: 9930464-9930464 on Build GRCh38
rs776178658 Chr.1: 9930413-9930413 on Build GRCh38
rs745384264 Chr.1: 9930507-9930507 on Build GRCh38
rs758668718 Chr.1: 9930407-9930409 on Build GRCh38
rs777879453 Chr.1: 9930429-9930435 on Build GRCh38
rs748518908 Chr.1: 9930360-9930360 on Build GRCh38
rs756326455 Chr.1: 9930467-9930467 on Build GRCh38
rs767745716 Chr.1: 9930456-9930456 on Build GRCh38
rs888270230 Chr.1: 9930421-9930421 on Build GRCh38
rs779313258 Chr.1: 9930402-9930402 on Build GRCh38
rs201530461 Chr.1: 9930501-9930501 on Build GRCh38
rs375762992 Chr.1: 9930493-9930493 on Build GRCh38
rs760255793 Chr.1: 9930385-9930385 on Build GRCh38
rs755535414 Chr.1: 9930299-9930299 on Build GRCh38
rs753028254 Chr.1: 9930401-9930402 on Build GRCh38
rs770308947 Chr.1: 9930389-9930389 on Build GRCh38
rs758788147 Chr.1: 9930484-9930484 on Build GRCh38
rs779477038 Chr.1: 9930355-9930355 on Build GRCh38
rs980820735 Chr.1: 9930323-9930323 on Build GRCh38
rs914181082 Chr.1: 9930458-9930458 on Build GRCh38
rs765149898 Chr.1: 9930430-9930430 on Build GRCh38
rs905113940 Chr.1: 9930487-9930487 on Build GRCh38
rs542810040 Chr.1: 9930477-9930477 on Build GRCh38
rs779495324 Chr.1: 9930478-9930478 on Build GRCh38
rs759443241 Chr.1: 9930419-9930419 on Build GRCh38
rs371896710 Chr.1: 9930376-9930376 on Build GRCh38
rs572325999 Chr.1: 9930325-9930325 on Build GRCh38
rs754311665 Chr.1: 9930360-9930362 on Build GRCh38
rs777514290 Chr.1: 9930488-9930488 on Build GRCh38
rs762674523 Chr.1: 9930434-9930434 on Build GRCh38
rs771855432 Chr.1: 9930375-9930375 on Build GRCh38
rs926800909 Chr.1: 9930359-9930359 on Build GRCh38
rs752769691 Chr.1: 9930482-9930482 on Build GRCh38
rs367641955 Chr.1: 9930354-9930354 on Build GRCh38
rs542213528 Chr.1: 9930296-9930296 on Build GRCh38

View Full Product Details