Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42634799-42635036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STARD9
Entrez Gene ID
57519
Gene Name
StAR related lipid transfer domain containing 9
Gene Aliases
KIF16A
UniGene
Hs.122061
Gene Chromosome Location
Chr.15: 42575646-42720998 on Build GRCh38
Transcripts
NM_020759.2 XM_011521831.2 XM_011521832.2 XM_011521833.2 XM_011521834.2 XM_011521835.2 XM_011521836.2 XM_011521837.2 XM_017022439.1 XM_017022440.1 XM_017022441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6015732 c.272T>C p.V91A Chr.15: 42634893-42634893 on Build GRCh38
3956627 c.263T>A p.L88Q Chr.15: 42634884-42634884 on Build GRCh38
5350257 c.350C>T p.P117L Chr.15: 42634971-42634971 on Build GRCh38
6383180 c.250G>T p.G84W Chr.15: 42634871-42634871 on Build GRCh38
1629560 c.266C>T p.S89F Chr.15: 42634887-42634887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010817113 Chr.15: 42634893-42634893 on Build GRCh38
rs773552277 Chr.15: 42635009-42635009 on Build GRCh38
rs937584761 Chr.15: 42634846-42634846 on Build GRCh38
rs769181914 Chr.15: 42634909-42634909 on Build GRCh38
rs189043638 Chr.15: 42634933-42634933 on Build GRCh38
rs780123175 Chr.15: 42634961-42634961 on Build GRCh38
rs893422455 Chr.15: 42634885-42634885 on Build GRCh38
rs775693640 Chr.15: 42634903-42634903 on Build GRCh38
rs904545235 Chr.15: 42634932-42634932 on Build GRCh38
rs777055958 Chr.15: 42634916-42634916 on Build GRCh38
rs936083314 Chr.15: 42634938-42634938 on Build GRCh38
rs534934951 Chr.15: 42634900-42634900 on Build GRCh38
rs377240528 Chr.15: 42634990-42634990 on Build GRCh38
rs913824870 Chr.15: 42634832-42634832 on Build GRCh38
rs755982653 Chr.15: 42634989-42634989 on Build GRCh38
rs17767439 Chr.15: 42634984-42634984 on Build GRCh38
rs745900641 Chr.15: 42634838-42634838 on Build GRCh38
rs745500128 Chr.15: 42634875-42634875 on Build GRCh38
rs894616110 Chr.15: 42634972-42634975 on Build GRCh38
rs778682289 Chr.15: 42634861-42634861 on Build GRCh38
rs868612063 Chr.15: 42634971-42634971 on Build GRCh38
rs1028925325 Chr.15: 42634954-42634954 on Build GRCh38
rs769249865 Chr.15: 42634885-42634885 on Build GRCh38
rs756765568 Chr.15: 42634836-42634836 on Build GRCh38

View Full Product Details