Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205098950-205099201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP5
Entrez Gene ID
5929
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
Gene Aliases
RBQ3, SWD1
UniGene
Hs.519230
Gene Chromosome Location
Chr.1: 205086142-205122022 on Build GRCh38
Transcripts
NM_001193272.1 NM_001193273.1 NM_005057.3 XM_011509833.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4608252 c.1083G>T p.E361D Chr.1: 205099002-205099002 on Build GRCh38
5417404 c.986G>T p.W329L Chr.1: 205099099-205099099 on Build GRCh38
902723 c.1012G>T p.E338* Chr.1: 205099073-205099073 on Build GRCh38
6459959 c.1059T>C p.D353D Chr.1: 205099026-205099026 on Build GRCh38
4720574 c.1096+2T>C p.? Chr.1: 205098987-205098987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047879423 Chr.1: 205099015-205099015 on Build GRCh38
rs577555468 Chr.1: 205098978-205098978 on Build GRCh38
rs560350657 Chr.1: 205099164-205099164 on Build GRCh38
rs767402413 Chr.1: 205099156-205099156 on Build GRCh38
rs755223800 Chr.1: 205098972-205098972 on Build GRCh38
rs143332925 Chr.1: 205099041-205099041 on Build GRCh38
rs778203449 Chr.1: 205099010-205099010 on Build GRCh38
rs575450921 Chr.1: 205099137-205099137 on Build GRCh38
rs779147043 Chr.1: 205098973-205098973 on Build GRCh38
rs918716585 Chr.1: 205099127-205099127 on Build GRCh38
rs762720670 Chr.1: 205099120-205099120 on Build GRCh38
rs745325699 Chr.1: 205099058-205099058 on Build GRCh38
rs758788378 Chr.1: 205098985-205098985 on Build GRCh38
rs761704078 Chr.1: 205099149-205099149 on Build GRCh38
rs776315962 Chr.1: 205099050-205099050 on Build GRCh38
rs763932836 Chr.1: 205099124-205099124 on Build GRCh38
rs756723042 Chr.1: 205099112-205099112 on Build GRCh38
rs189854779 Chr.1: 205099023-205099023 on Build GRCh38
rs769348224 Chr.1: 205099111-205099111 on Build GRCh38
rs774951245 Chr.1: 205098982-205098983 on Build GRCh38
rs1039193516 Chr.1: 205099152-205099152 on Build GRCh38

View Full Product Details