Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28621867-28622134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF12
Entrez Gene ID
6883
Gene Name
TATA-box binding protein associated factor 12
Gene Aliases
TAF2J, TAFII20
UniGene
Hs.530251
Gene Chromosome Location
Chr.1: 28603096-28648707 on Build GRCh38
Transcripts
NM_001135218.1 NM_005644.3 XM_017002186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6396363 c.122C>T p.P41L Chr.1: 28621960-28621960 on Build GRCh38
121996 c.36C>G p.L12L Chr.1: 28622046-28622046 on Build GRCh38
4031206 c.21A>G p.S7S Chr.1: 28622061-28622061 on Build GRCh38
6527563 c.38C>G p.S13C Chr.1: 28622044-28622044 on Build GRCh38
4481327 c.24C>T p.A8A Chr.1: 28622058-28622058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755173784 Chr.1: 28622050-28622050 on Build GRCh38
rs1804642 Chr.1: 28621989-28621989 on Build GRCh38
rs754383510 Chr.1: 28621899-28621899 on Build GRCh38
rs932515133 Chr.1: 28622073-28622073 on Build GRCh38
rs748810993 Chr.1: 28621949-28621949 on Build GRCh38
rs758834398 Chr.1: 28622089-28622089 on Build GRCh38
rs867724132 Chr.1: 28622050-28622050 on Build GRCh38
rs138962568 Chr.1: 28622041-28622041 on Build GRCh38
rs760955522 Chr.1: 28621984-28621984 on Build GRCh38
rs200737548 Chr.1: 28621919-28621919 on Build GRCh38
rs761840003 Chr.1: 28622013-28622013 on Build GRCh38
rs796797289 Chr.1: 28622065-28622065 on Build GRCh38
rs777359513 Chr.1: 28621948-28621948 on Build GRCh38
rs778342112 Chr.1: 28622076-28622076 on Build GRCh38
rs779433369 Chr.1: 28621961-28621961 on Build GRCh38
rs752379232 Chr.1: 28622038-28622038 on Build GRCh38
rs534981616 Chr.1: 28622064-28622064 on Build GRCh38
rs376623498 Chr.1: 28621963-28621963 on Build GRCh38
rs776767504 Chr.1: 28622093-28622093 on Build GRCh38
rs776898097 Chr.1: 28621988-28621988 on Build GRCh38
rs190752579 Chr.1: 28621973-28621973 on Build GRCh38
rs781086540 Chr.1: 28621937-28621937 on Build GRCh38
rs752431769 Chr.1: 28621944-28621944 on Build GRCh38
rs770466818 Chr.1: 28621953-28621953 on Build GRCh38
rs536479244 Chr.1: 28621985-28621985 on Build GRCh38
rs375818698 Chr.1: 28622078-28622078 on Build GRCh38
rs756805497 Chr.1: 28622074-28622074 on Build GRCh38
rs766482084 Chr.1: 28621897-28621897 on Build GRCh38
rs767427390 Chr.1: 28622029-28622029 on Build GRCh38
rs748353865 Chr.1: 28622099-28622099 on Build GRCh38
rs905139105 Chr.1: 28622067-28622067 on Build GRCh38
rs200582502 Chr.1: 28622090-28622090 on Build GRCh38
rs772446054 Chr.1: 28621970-28621970 on Build GRCh38
rs922441114 Chr.1: 28622049-28622049 on Build GRCh38
rs372852548 Chr.1: 28621969-28621969 on Build GRCh38
rs759507763 Chr.1: 28622023-28622023 on Build GRCh38
rs755657743 Chr.1: 28621946-28621946 on Build GRCh38
rs11540607 Chr.1: 28622071-28622071 on Build GRCh38
rs149193832 Chr.1: 28622022-28622022 on Build GRCh38
rs202043421 Chr.1: 28621905-28621905 on Build GRCh38
rs573195656 Chr.1: 28622052-28622052 on Build GRCh38
rs199614348 Chr.1: 28621894-28621894 on Build GRCh38
rs746402511 Chr.1: 28622081-28622081 on Build GRCh38
rs371905728 Chr.1: 28621936-28621936 on Build GRCh38
rs765386621 Chr.1: 28622014-28622014 on Build GRCh38

View Full Product Details