Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13214298-13214543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1A
Entrez Gene ID
773
Gene Name
calcium voltage-gated channel subunit alpha1 A
Gene Aliases
APCA, BI, CACNL1A4, CAV2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6
UniGene
Hs.501632
Gene Chromosome Location
Chr.19: 13206442-13506460 on Build GRCh38
Transcripts
NM_000068.3 NM_001127221.1 NM_001127222.1 NM_001174080.1 NM_023035.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1304067 c.5802C>A p.S1934S Chr.19: 13214538-13214538 on Build GRCh38
3529009 c.5802C>T p.S1934S Chr.19: 13214538-13214538 on Build GRCh38
1304063 c.5815G>C p.D1939H Chr.19: 13214525-13214525 on Build GRCh38
3529005 c.5858G>A p.G1953E Chr.19: 13214315-13214315 on Build GRCh38
4947723 c.5854G>A p.V1952M Chr.19: 13214319-13214319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556502230 Chr.19: 13214350-13214350 on Build GRCh38
rs372261381 Chr.19: 13214340-13214340 on Build GRCh38
rs767035894 Chr.19: 13214451-13214451 on Build GRCh38
rs368203745 Chr.19: 13214329-13214329 on Build GRCh38
rs772598066 Chr.19: 13214340-13214341 on Build GRCh38
rs750093351 Chr.19: 13214458-13214458 on Build GRCh38
rs951196653 Chr.19: 13214500-13214500 on Build GRCh38
rs184658006 Chr.19: 13214347-13214347 on Build GRCh38
rs765769753 Chr.19: 13214352-13214352 on Build GRCh38
rs560179255 Chr.19: 13214477-13214477 on Build GRCh38
rs761332254 Chr.19: 13214374-13214374 on Build GRCh38
rs367895273 Chr.19: 13214372-13214372 on Build GRCh38
rs772163130 Chr.19: 13214339-13214339 on Build GRCh38
rs17846925 Chr.19: 13214415-13214415 on Build GRCh38
rs371733571 Chr.19: 13214491-13214491 on Build GRCh38
rs755806752 Chr.19: 13214460-13214460 on Build GRCh38
rs750284095 Chr.19: 13214435-13214435 on Build GRCh38
rs896854918 Chr.19: 13214450-13214450 on Build GRCh38
rs776065476 Chr.19: 13214494-13214495 on Build GRCh38
rs1005508013 Chr.19: 13214424-13214424 on Build GRCh38
rs748279762 Chr.19: 13214330-13214330 on Build GRCh38
rs971098914 Chr.19: 13214511-13214511 on Build GRCh38
rs758507856 Chr.19: 13214509-13214509 on Build GRCh38
rs188329345 Chr.19: 13214421-13214421 on Build GRCh38
rs534218994 Chr.19: 13214320-13214320 on Build GRCh38
rs748649290 Chr.19: 13214329-13214330 on Build GRCh38
rs771333825 Chr.19: 13214349-13214349 on Build GRCh38
rs779787185 Chr.19: 13214317-13214317 on Build GRCh38
rs993807681 Chr.19: 13214468-13214468 on Build GRCh38
rs749251058 Chr.19: 13214319-13214319 on Build GRCh38
rs777815999 Chr.19: 13214520-13214520 on Build GRCh38
rs756095565 Chr.19: 13214486-13214486 on Build GRCh38
rs766183151 Chr.19: 13214463-13214463 on Build GRCh38

View Full Product Details