The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080432.2 | NP_001073901.1 | 8 - 9 | 1584 | 62 |
XM_011523313.2 | XP_011521615.1 | 8 - 9 | 1578 | 62 | |
GenBank mRNA | AB051539.1 | - | 8 - 9 | 1368 | 62 |
AK096554.1 | - | 8 - 9 | 1391 | 62 | |
AK295196.1 | - | 7 - 8 | 737 | 62 | |
BC030798.1 | - | 2 - 3 | 352 | 62 | |
BC132892.1 | - | 3 - 4 | 287 | 62 | |
BC137091.1 | - | 3 - 4 | 267 | 62 |