Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 71787562-71788102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM86C1
Entrez Gene ID
55199
Gene Name
family with sequence similarity 86 member C1
Gene Aliases
FAM86C
UniGene
Hs.567527
Gene Chromosome Location
Chr.11: 71787511-71801234 on Build GRCh38
Transcripts
NM_001099653.1 NM_018172.2 NM_152563.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3749186 c.89G>C p.R30P Chr.11: 71787625-71787625 on Build GRCh38
3721744 c.66A>G p.A22A Chr.11: 71787602-71787602 on Build GRCh38
3749187 c.89G>C p.R30P Chr.11: 71787625-71787625 on Build GRCh38
5773749 c.73G>C p.A25P Chr.11: 71787609-71787609 on Build GRCh38
3721745 c.66A>G p.A22A Chr.11: 71787602-71787602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747599179 Chr.11: 71787594-71787594 on Build GRCh38
rs185917759 Chr.11: 71787945-71787945 on Build GRCh38
rs747825632 Chr.11: 71787617-71787617 on Build GRCh38
rs773236014 Chr.11: 71787597-71787597 on Build GRCh38
rs12283346 Chr.11: 71787625-71787625 on Build GRCh38
rs928557934 Chr.11: 71787771-71787771 on Build GRCh38
rs543596115 Chr.11: 71787736-71787736 on Build GRCh38
rs186708274 Chr.11: 71787821-71787821 on Build GRCh38
rs778643510 Chr.11: 71787592-71787592 on Build GRCh38
rs770943212 Chr.11: 71787601-71787601 on Build GRCh38
rs989945340 Chr.11: 71787708-71787708 on Build GRCh38
rs1031376860 Chr.11: 71787689-71787689 on Build GRCh38
rs1035520602 Chr.11: 71787883-71787883 on Build GRCh38
rs749628675 Chr.11: 71787649-71787669 on Build GRCh38
rs774651868 Chr.11: 71787766-71787766 on Build GRCh38
rs540940080 Chr.11: 71787822-71787822 on Build GRCh38
rs563362203 Chr.11: 71787855-71787855 on Build GRCh38
rs576178514 Chr.11: 71787725-71787725 on Build GRCh38
rs979960896 Chr.11: 71787769-71787769 on Build GRCh38
rs11234943 Chr.11: 71787834-71787834 on Build GRCh38
rs61889008 Chr.11: 71787938-71787938 on Build GRCh38
rs923495441 Chr.11: 71787961-71787961 on Build GRCh38
rs770715596 Chr.11: 71787635-71787635 on Build GRCh38
rs760825003 Chr.11: 71787660-71787660 on Build GRCh38
rs1021395775 Chr.11: 71787711-71787711 on Build GRCh38
rs35109270 Chr.11: 71787611-71787612 on Build GRCh38
rs902095673 Chr.11: 71787823-71787823 on Build GRCh38
rs866910498 Chr.11: 71788027-71788027 on Build GRCh38
rs756605716 Chr.11: 71787609-71787609 on Build GRCh38
rs9734986 Chr.11: 71787923-71787923 on Build GRCh38
rs879935239 Chr.11: 71788020-71788020 on Build GRCh38
rs891428240 Chr.11: 71788014-71788014 on Build GRCh38
rs796385804 Chr.11: 71787932-71787934 on Build GRCh38
rs948627494 Chr.11: 71787603-71787603 on Build GRCh38
rs758122009 Chr.11: 71787678-71787678 on Build GRCh38
rs190108203 Chr.11: 71787846-71787846 on Build GRCh38
rs754405004 Chr.11: 71787667-71787667 on Build GRCh38
rs1053391161 Chr.11: 71787981-71787981 on Build GRCh38
rs375753901 Chr.11: 71787717-71787717 on Build GRCh38
rs755378642 Chr.11: 71787613-71787613 on Build GRCh38
rs955977006 Chr.11: 71787695-71787695 on Build GRCh38
rs572072020 Chr.11: 71787639-71787639 on Build GRCh38
rs775310062 Chr.11: 71787652-71787652 on Build GRCh38
rs760667166 Chr.11: 71787598-71787598 on Build GRCh38
rs749725857 Chr.11: 71787588-71787588 on Build GRCh38
rs768357623 Chr.11: 71787585-71787600 on Build GRCh38
rs773264145 Chr.11: 71787659-71787659 on Build GRCh38
rs767558028 Chr.11: 71787606-71787606 on Build GRCh38
rs181678084 Chr.11: 71787877-71787877 on Build GRCh38
rs34076970 Chr.11: 71787780-71787780 on Build GRCh38
rs561694708 Chr.11: 71787819-71787819 on Build GRCh38
rs939369381 Chr.11: 71787641-71787641 on Build GRCh38
rs535728045 Chr.11: 71787615-71787615 on Build GRCh38
rs938650951 Chr.11: 71787778-71787778 on Build GRCh38
rs751838321 Chr.11: 71787605-71787605 on Build GRCh38
rs767791088 Chr.11: 71787657-71787657 on Build GRCh38
rs929361803 Chr.11: 71787813-71787813 on Build GRCh38
rs1049190096 Chr.11: 71787814-71787814 on Build GRCh38
rs769652472 Chr.11: 71787651-71787651 on Build GRCh38
rs191079008 Chr.11: 71788038-71788038 on Build GRCh38
rs771742917 Chr.11: 71787596-71787596 on Build GRCh38
rs969831800 Chr.11: 71787879-71787879 on Build GRCh38
rs1039214187 Chr.11: 71787957-71787957 on Build GRCh38
rs867789019 Chr.11: 71787774-71787774 on Build GRCh38
rs531991627 Chr.11: 71787865-71787865 on Build GRCh38
rs909297679 Chr.11: 71787941-71787941 on Build GRCh38
rs750539545 Chr.11: 71787608-71787608 on Build GRCh38
rs979902006 Chr.11: 71787880-71787880 on Build GRCh38
rs997596731 Chr.11: 71787670-71787670 on Build GRCh38
rs559614644 Chr.11: 71787832-71787832 on Build GRCh38
rs750551442 Chr.11: 71788017-71788017 on Build GRCh38
rs753153102 Chr.11: 71787677-71787677 on Build GRCh38
rs1011718495 Chr.11: 71787866-71787866 on Build GRCh38
rs765613565 Chr.11: 71787676-71787676 on Build GRCh38
rs746585417 Chr.11: 71787628-71787628 on Build GRCh38
rs755252941 Chr.11: 71787669-71787669 on Build GRCh38
rs969884295 Chr.11: 71787741-71787741 on Build GRCh38
rs557806487 Chr.11: 71787656-71787656 on Build GRCh38
rs11234947 Chr.11: 71787984-71787984 on Build GRCh38
rs12277291 Chr.11: 71787602-71787602 on Build GRCh38
rs551167138 Chr.11: 71788072-71788072 on Build GRCh38
rs764155176 Chr.11: 71787604-71787604 on Build GRCh38
rs933550635 Chr.11: 71787974-71787977 on Build GRCh38
rs959916709 Chr.11: 71788053-71788053 on Build GRCh38
rs919225620 Chr.11: 71787913-71787913 on Build GRCh38
rs865878507 Chr.11: 71787772-71787772 on Build GRCh38
rs768183363 Chr.11: 71787590-71787590 on Build GRCh38
rs745610316 Chr.11: 71787645-71787645 on Build GRCh38
rs1017881364 Chr.11: 71787666-71787666 on Build GRCh38
rs919293636 Chr.11: 71787787-71787787 on Build GRCh38
rs11234944 Chr.11: 71787856-71787856 on Build GRCh38
rs960215531 Chr.11: 71787901-71787901 on Build GRCh38
rs905611456 Chr.11: 71788024-71788024 on Build GRCh38
rs780672506 Chr.11: 71787610-71787610 on Build GRCh38
rs754095233 Chr.11: 71787611-71787611 on Build GRCh38
rs11234942 Chr.11: 71787648-71787648 on Build GRCh38
rs34702231 Chr.11: 71787590-71787591 on Build GRCh38
rs367665207 Chr.11: 71787909-71787909 on Build GRCh38
rs771900192 Chr.11: 71787622-71787622 on Build GRCh38
rs766732417 Chr.11: 71787663-71787663 on Build GRCh38

View Full Product Details