Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72229377-72229872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6850225 c.772G>A p.E258K Chr.11: 72229681-72229681 on Build GRCh38
6318626 c.719G>T p.S240I Chr.11: 72229524-72229524 on Build GRCh38
3810362 c.827C>G p.S276* Chr.11: 72229736-72229736 on Build GRCh38
5557883 c.799A>G p.K267E Chr.11: 72229708-72229708 on Build GRCh38
6315502 c.825G>T p.L275L Chr.11: 72229734-72229734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548467739 Chr.11: 72229796-72229796 on Build GRCh38
rs776498366 Chr.11: 72229594-72229594 on Build GRCh38
rs760757952 Chr.11: 72229752-72229752 on Build GRCh38
rs774613122 Chr.11: 72229728-72229728 on Build GRCh38
rs777376401 Chr.11: 72229677-72229677 on Build GRCh38
rs758056866 Chr.11: 72229781-72229781 on Build GRCh38
rs746518063 Chr.11: 72229683-72229683 on Build GRCh38
rs761235393 Chr.11: 72229496-72229496 on Build GRCh38
rs375761722 Chr.11: 72229635-72229635 on Build GRCh38
rs779654573 Chr.11: 72229454-72229454 on Build GRCh38
rs568210147 Chr.11: 72229759-72229759 on Build GRCh38
rs377498321 Chr.11: 72229585-72229585 on Build GRCh38
rs746501297 Chr.11: 72229798-72229798 on Build GRCh38
rs367992744 Chr.11: 72229588-72229588 on Build GRCh38
rs1007477970 Chr.11: 72229828-72229828 on Build GRCh38
rs186423971 Chr.11: 72229407-72229407 on Build GRCh38
rs767762060 Chr.11: 72229734-72229734 on Build GRCh38
rs778596194 Chr.11: 72229664-72229664 on Build GRCh38
rs766325854 Chr.11: 72229528-72229528 on Build GRCh38
rs776376700 Chr.11: 72229782-72229783 on Build GRCh38
rs201713633 Chr.11: 72229659-72229659 on Build GRCh38
rs1017166222 Chr.11: 72229406-72229406 on Build GRCh38
rs749022309 Chr.11: 72229462-72229462 on Build GRCh38
rs797044468 Chr.11: 72229677-72229677 on Build GRCh38
rs373301495 Chr.11: 72229438-72229438 on Build GRCh38
rs771989660 Chr.11: 72229524-72229524 on Build GRCh38
rs986072907 Chr.11: 72229429-72229429 on Build GRCh38
rs374049262 Chr.11: 72229660-72229660 on Build GRCh38
rs900076023 Chr.11: 72229846-72229846 on Build GRCh38
rs527470737 Chr.11: 72229837-72229837 on Build GRCh38
rs910375345 Chr.11: 72229456-72229456 on Build GRCh38
rs761978331 Chr.11: 72229731-72229731 on Build GRCh38
rs759259120 Chr.11: 72229595-72229595 on Build GRCh38
rs1005517470 Chr.11: 72229776-72229776 on Build GRCh38
rs770890568 Chr.11: 72229507-72229507 on Build GRCh38
rs770212675 Chr.11: 72229441-72229441 on Build GRCh38
rs374410580 Chr.11: 72229780-72229780 on Build GRCh38
rs915157319 Chr.11: 72229639-72229639 on Build GRCh38
rs573385475 Chr.11: 72229584-72229584 on Build GRCh38
rs774501149 Chr.11: 72229624-72229624 on Build GRCh38
rs766877002 Chr.11: 72229658-72229658 on Build GRCh38
rs1052407023 Chr.11: 72229592-72229592 on Build GRCh38
rs891799988 Chr.11: 72229654-72229654 on Build GRCh38
rs758146582 Chr.11: 72229676-72229676 on Build GRCh38
rs12795135 Chr.11: 72229605-72229605 on Build GRCh38
rs746647683 Chr.11: 72229677-72229678 on Build GRCh38
rs760513714 Chr.11: 72229527-72229527 on Build GRCh38
rs750527388 Chr.11: 72229646-72229646 on Build GRCh38
rs753707673 Chr.11: 72229532-72229532 on Build GRCh38
rs775043885 Chr.11: 72229655-72229655 on Build GRCh38
rs951723805 Chr.11: 72229792-72229792 on Build GRCh38
rs566700152 Chr.11: 72229822-72229822 on Build GRCh38
rs773348902 Chr.11: 72229526-72229526 on Build GRCh38
rs770929582 Chr.11: 72229692-72229692 on Build GRCh38
rs745668155 Chr.11: 72229707-72229707 on Build GRCh38
rs371206323 Chr.11: 72229715-72229715 on Build GRCh38
rs753006035 Chr.11: 72229414-72229414 on Build GRCh38
rs756452267 Chr.11: 72229559-72229559 on Build GRCh38
rs755346094 Chr.11: 72229778-72229778 on Build GRCh38
rs971735121 Chr.11: 72229405-72229405 on Build GRCh38
rs756574741 Chr.11: 72229651-72229651 on Build GRCh38
rs747802572 Chr.11: 72229671-72229671 on Build GRCh38
rs1003397653 Chr.11: 72229693-72229693 on Build GRCh38
rs771023319 Chr.11: 72229593-72229593 on Build GRCh38
rs887200823 Chr.11: 72229732-72229732 on Build GRCh38
rs539048364 Chr.11: 72229767-72229767 on Build GRCh38
rs80156982 Chr.11: 72229705-72229705 on Build GRCh38
rs773871418 Chr.11: 72229488-72229488 on Build GRCh38
rs763192141 Chr.11: 72229537-72229537 on Build GRCh38
rs757335516 Chr.11: 72229543-72229543 on Build GRCh38
rs774695734 Chr.11: 72229544-72229544 on Build GRCh38
rs758385012 Chr.11: 72229416-72229416 on Build GRCh38
rs61736300 Chr.11: 72229710-72229710 on Build GRCh38
rs529907144 Chr.11: 72229772-72229772 on Build GRCh38
rs776487134 Chr.11: 72229703-72229703 on Build GRCh38
rs768216134 Chr.11: 72229485-72229485 on Build GRCh38
rs45506896 Chr.11: 72229589-72229589 on Build GRCh38
rs749646417 Chr.11: 72229578-72229578 on Build GRCh38
rs780351983 Chr.11: 72229576-72229576 on Build GRCh38
rs768343273 Chr.11: 72229776-72229776 on Build GRCh38
rs867029347 Chr.11: 72229652-72229652 on Build GRCh38
rs369528817 Chr.11: 72229831-72229831 on Build GRCh38
rs764222606 Chr.11: 72229539-72229539 on Build GRCh38
rs552436243 Chr.11: 72229847-72229847 on Build GRCh38
rs750970470 Chr.11: 72229549-72229549 on Build GRCh38
rs543320087 Chr.11: 72229429-72229429 on Build GRCh38
rs554820987 Chr.11: 72229540-72229540 on Build GRCh38
rs878853120 Chr.11: 72229558-72229558 on Build GRCh38
rs575979068 Chr.11: 72229428-72229428 on Build GRCh38
rs773210890 Chr.11: 72229740-72229740 on Build GRCh38

View Full Product Details