Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77872087-77872553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAMDC
Entrez Gene ID
28971
Gene Name
adipogenesis associated Mth938 domain containing
Gene Aliases
C11orf67, CK067, PTD015
UniGene
Hs.745040
Gene Chromosome Location
Chr.11: 77821114-77901281 on Build GRCh38
Transcripts
NM_001316957.1 NM_001316958.1 NM_001316960.1 NM_001316961.1 NM_001316962.1 NM_024684.3 NR_133637.1 XM_011544968.2 XM_011544969.2 XM_017017620.1 XM_017017923.1

Gene Symbol
RSF1
Entrez Gene ID
51773
Gene Name
remodeling and spacing factor 1
Gene Aliases
HBXAP, RSF-1, XAP8, p325
UniGene
Hs.420229
Gene Chromosome Location
Chr.11: 77660009-77872232 on Build GRCh38
Transcripts
NM_001316957.1 NM_001316958.1 NM_001316960.1 NM_001316961.1 NM_001316962.1 NM_024684.3 NR_133637.1 XM_011544968.2 XM_011544969.2 XM_017017620.1 XM_017017923.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5982388 c.300A>C p.A100A Chr.11: 77872246-77872246 on Build GRCh38
932213 c.268A>G p.I90V Chr.11: 77872214-77872214 on Build GRCh38
5982389 c.342G>A p.V114V Chr.11: 77872288-77872288 on Build GRCh38
147295 c.274G>A p.V92M Chr.11: 77872220-77872220 on Build GRCh38
932214 c.277C>T p.R93W Chr.11: 77872223-77872223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558708060 Chr.11: 77872427-77872427 on Build GRCh38
rs1056526975 Chr.11: 77872384-77872384 on Build GRCh38
rs756693045 Chr.11: 77872215-77872215 on Build GRCh38
rs957907207 Chr.11: 77872130-77872130 on Build GRCh38
rs779087535 Chr.11: 77872350-77872350 on Build GRCh38
rs781672405 Chr.11: 77872124-77872124 on Build GRCh38
rs751418968 Chr.11: 77872129-77872129 on Build GRCh38
rs745307773 Chr.11: 77872373-77872373 on Build GRCh38
rs746754321 Chr.11: 77872254-77872254 on Build GRCh38
rs771928726 Chr.11: 77872166-77872166 on Build GRCh38
rs555078639 Chr.11: 77872449-77872449 on Build GRCh38
rs1022022854 Chr.11: 77872507-77872507 on Build GRCh38
rs752285965 Chr.11: 77872201-77872201 on Build GRCh38
rs758966519 Chr.11: 77872194-77872194 on Build GRCh38
rs768012112 Chr.11: 77872186-77872186 on Build GRCh38
rs200436609 Chr.11: 77872295-77872295 on Build GRCh38
rs1003220584 Chr.11: 77872375-77872375 on Build GRCh38
rs746302505 Chr.11: 77872126-77872126 on Build GRCh38
rs764930310 Chr.11: 77872199-77872199 on Build GRCh38
rs148604757 Chr.11: 77872333-77872333 on Build GRCh38
rs752803733 Chr.11: 77872355-77872355 on Build GRCh38
rs777763915 Chr.11: 77872244-77872244 on Build GRCh38
rs2186564 Chr.11: 77872220-77872220 on Build GRCh38
rs756808773 Chr.11: 77872352-77872352 on Build GRCh38
rs762166963 Chr.11: 77872203-77872203 on Build GRCh38
rs779283881 Chr.11: 77872226-77872226 on Build GRCh38
rs377044804 Chr.11: 77872131-77872131 on Build GRCh38
rs776241998 Chr.11: 77872521-77872521 on Build GRCh38
rs754956539 Chr.11: 77872224-77872224 on Build GRCh38
rs749305430 Chr.11: 77872149-77872149 on Build GRCh38
rs968985680 Chr.11: 77872121-77872121 on Build GRCh38
rs1020662652 Chr.11: 77872170-77872170 on Build GRCh38
rs750839929 Chr.11: 77872332-77872332 on Build GRCh38
rs762540643 Chr.11: 77872314-77872314 on Build GRCh38
rs763251965 Chr.11: 77872524-77872524 on Build GRCh38
rs772202811 Chr.11: 77872423-77872423 on Build GRCh38
rs780132780 Chr.11: 77872132-77872132 on Build GRCh38
rs770969978 Chr.11: 77872259-77872259 on Build GRCh38
rs146747352 Chr.11: 77872267-77872267 on Build GRCh38
rs201384470 Chr.11: 77872172-77872172 on Build GRCh38
rs778498206 Chr.11: 77872368-77872368 on Build GRCh38
rs588217 Chr.11: 77872330-77872330 on Build GRCh38
rs748341123 Chr.11: 77872231-77872231 on Build GRCh38
rs754233870 Chr.11: 77872344-77872344 on Build GRCh38
rs774708425 Chr.11: 77872152-77872152 on Build GRCh38
rs768961363 Chr.11: 77872151-77872151 on Build GRCh38
rs368865821 Chr.11: 77872223-77872223 on Build GRCh38
rs773314104 Chr.11: 77872171-77872171 on Build GRCh38
rs750947566 Chr.11: 77872212-77872212 on Build GRCh38
rs766966230 Chr.11: 77872343-77872343 on Build GRCh38
rs768603004 Chr.11: 77872513-77872513 on Build GRCh38
rs766208651 Chr.11: 77872178-77872178 on Build GRCh38
rs373537906 Chr.11: 77872234-77872234 on Build GRCh38
rs755142887 Chr.11: 77872348-77872348 on Build GRCh38
rs747971261 Chr.11: 77872154-77872154 on Build GRCh38
rs769224687 Chr.11: 77872285-77872285 on Build GRCh38
rs897026762 Chr.11: 77872374-77872374 on Build GRCh38
rs775150284 Chr.11: 77872291-77872291 on Build GRCh38
rs61900171 Chr.11: 77872474-77872474 on Build GRCh38
rs668701 Chr.11: 77872430-77872430 on Build GRCh38
rs767825535 Chr.11: 77872210-77872210 on Build GRCh38
rs759096580 Chr.11: 77872269-77872269 on Build GRCh38

View Full Product Details