Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4275865-4276368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND2
Entrez Gene ID
894
Gene Name
cyclin D2
Gene Aliases
KIAK0002, MPPH3
UniGene
Hs.376071
Gene Chromosome Location
Chr.12: 4273736-4305356 on Build GRCh38
Transcripts
NM_001759.3 NR_125790.1

Gene Symbol
CCND2-AS1
Entrez Gene ID
103752584
Gene Name
CCND2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.577711
Gene Chromosome Location
Chr.12: 4248767-4276184 on Build GRCh38
Transcripts
NM_001759.3 NR_125790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6932939 c.232C>T p.P78S Chr.12: 4276041-4276041 on Build GRCh38
6934034 c.233C>T p.P78L Chr.12: 4276042-4276042 on Build GRCh38
5704438 c.307G>A p.A103T Chr.12: 4276116-4276116 on Build GRCh38
6938380 c.349C>T p.P117S Chr.12: 4276158-4276158 on Build GRCh38
4041845 c.228C>T p.V76V Chr.12: 4276037-4276037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750492844 Chr.12: 4275987-4275987 on Build GRCh38
rs867376054 Chr.12: 4276159-4276159 on Build GRCh38
rs1019910889 Chr.12: 4276090-4276090 on Build GRCh38
rs755023974 Chr.12: 4275998-4275999 on Build GRCh38
rs766500677 Chr.12: 4276161-4276161 on Build GRCh38
rs781280428 Chr.12: 4276233-4276240 on Build GRCh38
rs764380553 Chr.12: 4275980-4275980 on Build GRCh38
rs745404167 Chr.12: 4276229-4276229 on Build GRCh38
rs763529306 Chr.12: 4276160-4276160 on Build GRCh38
rs755257315 Chr.12: 4276031-4276031 on Build GRCh38
rs1006222339 Chr.12: 4276207-4276207 on Build GRCh38
rs770583513 Chr.12: 4275974-4275974 on Build GRCh38
rs773631850 Chr.12: 4276250-4276250 on Build GRCh38
rs757187520 Chr.12: 4276218-4276218 on Build GRCh38
rs75088331 Chr.12: 4275901-4275901 on Build GRCh38
rs777614015 Chr.12: 4276079-4276079 on Build GRCh38
rs201572845 Chr.12: 4275969-4275969 on Build GRCh38
rs977221684 Chr.12: 4276265-4276265 on Build GRCh38
rs748823991 Chr.12: 4276088-4276088 on Build GRCh38
rs995287593 Chr.12: 4275915-4275915 on Build GRCh38
rs771743347 Chr.12: 4276124-4276124 on Build GRCh38
rs761009594 Chr.12: 4275977-4275977 on Build GRCh38
rs1055859383 Chr.12: 4276172-4276172 on Build GRCh38
rs765683372 Chr.12: 4275986-4275986 on Build GRCh38
rs748346553 Chr.12: 4276248-4276248 on Build GRCh38
rs776929300 Chr.12: 4276150-4276150 on Build GRCh38
rs773652105 Chr.12: 4276157-4276157 on Build GRCh38
rs879306189 Chr.12: 4275992-4275992 on Build GRCh38
rs754206192 Chr.12: 4275981-4275981 on Build GRCh38
rs567484716 Chr.12: 4275914-4275914 on Build GRCh38
rs867321680 Chr.12: 4276196-4276196 on Build GRCh38
rs530949406 Chr.12: 4276282-4276282 on Build GRCh38
rs781149672 Chr.12: 4275964-4275964 on Build GRCh38
rs910989546 Chr.12: 4275903-4275903 on Build GRCh38
rs748533926 Chr.12: 4276238-4276238 on Build GRCh38
rs762341798 Chr.12: 4275982-4275982 on Build GRCh38
rs762422711 Chr.12: 4275973-4275973 on Build GRCh38
rs772731329 Chr.12: 4275971-4275971 on Build GRCh38
rs769655297 Chr.12: 4275970-4275970 on Build GRCh38
rs372495036 Chr.12: 4275916-4275916 on Build GRCh38
rs745455734 Chr.12: 4276109-4276109 on Build GRCh38
rs986579422 Chr.12: 4275902-4275902 on Build GRCh38
rs767018888 Chr.12: 4276261-4276261 on Build GRCh38
rs973025482 Chr.12: 4275891-4275891 on Build GRCh38
rs774554963 Chr.12: 4276262-4276262 on Build GRCh38
rs769997869 Chr.12: 4276241-4276241 on Build GRCh38
rs777749450 Chr.12: 4276195-4276195 on Build GRCh38
rs536143012 Chr.12: 4275936-4275936 on Build GRCh38
rs747983314 Chr.12: 4276050-4276050 on Build GRCh38
rs989820168 Chr.12: 4275927-4275927 on Build GRCh38
rs560009483 Chr.12: 4276043-4276043 on Build GRCh38
rs751723166 Chr.12: 4276178-4276178 on Build GRCh38
rs780412072 Chr.12: 4276007-4276007 on Build GRCh38
rs955247725 Chr.12: 4276331-4276331 on Build GRCh38
rs894158511 Chr.12: 4276340-4276340 on Build GRCh38
rs200547584 Chr.12: 4276259-4276259 on Build GRCh38
rs928577806 Chr.12: 4276277-4276277 on Build GRCh38
rs758474986 Chr.12: 4275988-4275988 on Build GRCh38
rs955187724 Chr.12: 4275902-4275903 on Build GRCh38
rs768631279 Chr.12: 4275959-4275959 on Build GRCh38
rs958255950 Chr.12: 4276202-4276202 on Build GRCh38
rs762132499 Chr.12: 4276153-4276153 on Build GRCh38
rs771688425 Chr.12: 4276230-4276230 on Build GRCh38
rs755925937 Chr.12: 4276194-4276194 on Build GRCh38
rs1008084448 Chr.12: 4276335-4276335 on Build GRCh38
rs1016654362 Chr.12: 4275890-4275890 on Build GRCh38
rs200880354 Chr.12: 4276103-4276103 on Build GRCh38
rs770403051 Chr.12: 4276091-4276091 on Build GRCh38
rs577370896 Chr.12: 4275944-4275944 on Build GRCh38
rs184590007 Chr.12: 4276190-4276190 on Build GRCh38
rs756058431 Chr.12: 4276064-4276064 on Build GRCh38
rs946354367 Chr.12: 4275995-4275995 on Build GRCh38
rs556741011 Chr.12: 4275937-4275937 on Build GRCh38
rs1056086683 Chr.12: 4276291-4276291 on Build GRCh38
rs759699380 Chr.12: 4276145-4276145 on Build GRCh38
rs765770986 Chr.12: 4276156-4276156 on Build GRCh38
rs550665561 Chr.12: 4276314-4276314 on Build GRCh38
rs775162503 Chr.12: 4276231-4276231 on Build GRCh38
rs878898553 Chr.12: 4276061-4276061 on Build GRCh38
rs1002529328 Chr.12: 4275890-4275890 on Build GRCh38
rs372384965 Chr.12: 4276227-4276227 on Build GRCh38
rs368616285 Chr.12: 4276215-4276215 on Build GRCh38

View Full Product Details