Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7141085-7141605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN3
Entrez Gene ID
9746
Gene Name
calsyntenin 3
Gene Aliases
CDHR14, CSTN3, alcbeta
UniGene
Hs.535378
Gene Chromosome Location
Chr.12: 7130371-7158945 on Build GRCh38
Transcripts
NM_014718.3 XM_006719163.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7206790 c.1460C>T p.A487V Chr.12: 7141378-7141378 on Build GRCh38
1476928 c.1338G>A p.E446E Chr.12: 7141256-7141256 on Build GRCh38
5528783 c.1368C>T p.F456F Chr.12: 7141286-7141286 on Build GRCh38
1705022 c.1467G>A p.M489I Chr.12: 7141385-7141385 on Build GRCh38
348759 c.1383C>T p.L461L Chr.12: 7141301-7141301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753709619 Chr.12: 7141244-7141244 on Build GRCh38
rs752459820 Chr.12: 7141106-7141106 on Build GRCh38
rs912418651 Chr.12: 7141145-7141145 on Build GRCh38
rs755822675 Chr.12: 7141499-7141499 on Build GRCh38
rs758824825 Chr.12: 7141409-7141409 on Build GRCh38
rs770818297 Chr.12: 7141198-7141200 on Build GRCh38
rs761881412 Chr.12: 7141427-7141427 on Build GRCh38
rs776890047 Chr.12: 7141483-7141483 on Build GRCh38
rs947076447 Chr.12: 7141164-7141164 on Build GRCh38
rs759671476 Chr.12: 7141201-7141201 on Build GRCh38
rs977006477 Chr.12: 7141466-7141466 on Build GRCh38
rs758035032 Chr.12: 7141366-7141366 on Build GRCh38
rs977843132 Chr.12: 7141572-7141572 on Build GRCh38
rs764104955 Chr.12: 7141234-7141234 on Build GRCh38
rs989439256 Chr.12: 7141534-7141534 on Build GRCh38
rs139052024 Chr.12: 7141498-7141498 on Build GRCh38
rs948871648 Chr.12: 7141579-7141579 on Build GRCh38
rs780769952 Chr.12: 7141374-7141374 on Build GRCh38
rs776643907 Chr.12: 7141339-7141339 on Build GRCh38
rs146448907 Chr.12: 7141380-7141380 on Build GRCh38
rs866743185 Chr.12: 7141286-7141286 on Build GRCh38
rs115260521 Chr.12: 7141159-7141159 on Build GRCh38
rs760573563 Chr.12: 7141232-7141232 on Build GRCh38
rs2305341 Chr.12: 7141200-7141200 on Build GRCh38
rs750199694 Chr.12: 7141348-7141348 on Build GRCh38
rs117852387 Chr.12: 7141268-7141268 on Build GRCh38
rs995073709 Chr.12: 7141333-7141333 on Build GRCh38
rs770302756 Chr.12: 7141510-7141510 on Build GRCh38
rs780284175 Chr.12: 7141391-7141391 on Build GRCh38
rs1027608950 Chr.12: 7141334-7141334 on Build GRCh38
rs145584691 Chr.12: 7141322-7141322 on Build GRCh38
rs986614674 Chr.12: 7141121-7141121 on Build GRCh38
rs771224492 Chr.12: 7141305-7141305 on Build GRCh38
rs79390230 Chr.12: 7141151-7141151 on Build GRCh38
rs201416693 Chr.12: 7141280-7141280 on Build GRCh38
rs775639634 Chr.12: 7141223-7141223 on Build GRCh38
rs149527518 Chr.12: 7141570-7141570 on Build GRCh38
rs776838306 Chr.12: 7141426-7141426 on Build GRCh38
rs570506390 Chr.12: 7141150-7141150 on Build GRCh38
rs779189003 Chr.12: 7141385-7141385 on Build GRCh38
rs149721212 Chr.12: 7141310-7141310 on Build GRCh38
rs758063912 Chr.12: 7141255-7141255 on Build GRCh38
rs757760011 Chr.12: 7141383-7141383 on Build GRCh38
rs773995517 Chr.12: 7141226-7141226 on Build GRCh38
rs761716029 Chr.12: 7141344-7141344 on Build GRCh38
rs139596561 Chr.12: 7141308-7141308 on Build GRCh38
rs906661473 Chr.12: 7141281-7141281 on Build GRCh38
rs199820992 Chr.12: 7141210-7141210 on Build GRCh38
rs914874847 Chr.12: 7141547-7141547 on Build GRCh38
rs938904198 Chr.12: 7141529-7141529 on Build GRCh38
rs886476541 Chr.12: 7141342-7141342 on Build GRCh38
rs746506042 Chr.12: 7141269-7141269 on Build GRCh38
rs150595257 Chr.12: 7141307-7141307 on Build GRCh38
rs201380732 Chr.12: 7141294-7141300 on Build GRCh38
rs778209104 Chr.12: 7141289-7141289 on Build GRCh38
rs377691953 Chr.12: 7141405-7141405 on Build GRCh38
rs749653032 Chr.12: 7141293-7141293 on Build GRCh38
rs1038159844 Chr.12: 7141582-7141582 on Build GRCh38
rs373349765 Chr.12: 7141341-7141341 on Build GRCh38
rs370124618 Chr.12: 7141422-7141422 on Build GRCh38
rs779918332 Chr.12: 7141148-7141148 on Build GRCh38
rs764988030 Chr.12: 7141249-7141249 on Build GRCh38
rs764534617 Chr.12: 7141464-7141464 on Build GRCh38
rs757077930 Chr.12: 7141245-7141245 on Build GRCh38
rs916080777 Chr.12: 7141574-7141574 on Build GRCh38
rs144059185 Chr.12: 7141378-7141378 on Build GRCh38
rs772209938 Chr.12: 7141390-7141390 on Build GRCh38
rs754398323 Chr.12: 7141382-7141382 on Build GRCh38
rs749987986 Chr.12: 7141253-7141253 on Build GRCh38
rs938811087 Chr.12: 7141224-7141224 on Build GRCh38
rs968761108 Chr.12: 7141476-7141476 on Build GRCh38
rs772308702 Chr.12: 7141309-7141309 on Build GRCh38
rs747090351 Chr.12: 7141411-7141411 on Build GRCh38
rs764158378 Chr.12: 7141326-7141326 on Build GRCh38
rs140338155 Chr.12: 7141323-7141323 on Build GRCh38
rs953927619 Chr.12: 7141533-7141533 on Build GRCh38
rs914336451 Chr.12: 7141447-7141447 on Build GRCh38
rs140816333 Chr.12: 7141387-7141387 on Build GRCh38
rs766006539 Chr.12: 7141367-7141367 on Build GRCh38

View Full Product Details