Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6415738-6416265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HES2
Entrez Gene ID
54626
Gene Name
hes family bHLH transcription factor 2
Gene Aliases
bHLHb40
UniGene
Hs.118727
Gene Chromosome Location
Chr.1: 6415232-6419919 on Build GRCh38
Transcripts
NM_019089.4

SNP Detail

SNP ID Chromosome Location
rs758885895 Chr.1: 6415850-6415850 on Build GRCh38
rs945407122 Chr.1: 6416107-6416107 on Build GRCh38
rs1045995816 Chr.1: 6415874-6415874 on Build GRCh38
rs981252786 Chr.1: 6415771-6415771 on Build GRCh38
rs113269467 Chr.1: 6416226-6416226 on Build GRCh38
rs974398673 Chr.1: 6416046-6416046 on Build GRCh38
rs936752640 Chr.1: 6415812-6415812 on Build GRCh38
rs551244338 Chr.1: 6416133-6416133 on Build GRCh38
rs370825813 Chr.1: 6415903-6415903 on Build GRCh38
rs1034855146 Chr.1: 6415927-6415927 on Build GRCh38
rs1049207261 Chr.1: 6415840-6415840 on Build GRCh38
rs951802378 Chr.1: 6416058-6416058 on Build GRCh38
rs965691323 Chr.1: 6416037-6416037 on Build GRCh38
rs754946135 Chr.1: 6416073-6416073 on Build GRCh38
rs1018810033 Chr.1: 6416030-6416030 on Build GRCh38
rs913885247 Chr.1: 6416081-6416081 on Build GRCh38
rs111617671 Chr.1: 6416130-6416130 on Build GRCh38
rs574006655 Chr.1: 6415842-6415842 on Build GRCh38
rs944374726 Chr.1: 6415783-6415783 on Build GRCh38
rs185579032 Chr.1: 6415836-6415836 on Build GRCh38
rs1041782141 Chr.1: 6416215-6416215 on Build GRCh38
rs368076429 Chr.1: 6416074-6416074 on Build GRCh38
rs778008241 Chr.1: 6416114-6416114 on Build GRCh38
rs770030255 Chr.1: 6416097-6416097 on Build GRCh38
rs147099461 Chr.1: 6415969-6415969 on Build GRCh38
rs748265123 Chr.1: 6416087-6416087 on Build GRCh38
rs907239969 Chr.1: 6415897-6415897 on Build GRCh38
rs528066348 Chr.1: 6416069-6416069 on Build GRCh38
rs940833947 Chr.1: 6416071-6416071 on Build GRCh38
rs890270907 Chr.1: 6415915-6415915 on Build GRCh38
rs1007426452 Chr.1: 6415952-6415952 on Build GRCh38
rs1055773600 Chr.1: 6416158-6416158 on Build GRCh38
rs1038832847 Chr.1: 6416149-6416149 on Build GRCh38
rs550585445 Chr.1: 6416160-6416160 on Build GRCh38
rs921669764 Chr.1: 6415801-6415801 on Build GRCh38
rs928837716 Chr.1: 6416104-6416104 on Build GRCh38
rs1017768648 Chr.1: 6415966-6415966 on Build GRCh38
rs911583226 Chr.1: 6415780-6415783 on Build GRCh38
rs544802996 Chr.1: 6415845-6415845 on Build GRCh38
rs999439675 Chr.1: 6416022-6416022 on Build GRCh38
rs563371408 Chr.1: 6415841-6415841 on Build GRCh38
rs902388673 Chr.1: 6416216-6416216 on Build GRCh38
rs4908889 Chr.1: 6416056-6416056 on Build GRCh38
rs971297878 Chr.1: 6416057-6416057 on Build GRCh38
rs974932597 Chr.1: 6415798-6415798 on Build GRCh38
rs568952398 Chr.1: 6415866-6415866 on Build GRCh38
rs1047537687 Chr.1: 6416198-6416198 on Build GRCh38
rs899008583 Chr.1: 6416171-6416171 on Build GRCh38
rs745638382 Chr.1: 6416195-6416195 on Build GRCh38
rs1011173936 Chr.1: 6415935-6415935 on Build GRCh38
rs983014914 Chr.1: 6416055-6416055 on Build GRCh38
rs980922571 Chr.1: 6416060-6416060 on Build GRCh38
rs898876372 Chr.1: 6415967-6415967 on Build GRCh38
rs949605133 Chr.1: 6415766-6415766 on Build GRCh38
rs926764036 Chr.1: 6415772-6415772 on Build GRCh38
rs779435577 Chr.1: 6416070-6416070 on Build GRCh38
rs950383513 Chr.1: 6416038-6416038 on Build GRCh38
rs1025957460 Chr.1: 6415978-6415978 on Build GRCh38
rs141712069 Chr.1: 6415839-6415839 on Build GRCh38
rs577939012 Chr.1: 6415930-6415930 on Build GRCh38
rs187733679 Chr.1: 6415905-6415905 on Build GRCh38
rs930855561 Chr.1: 6415924-6415924 on Build GRCh38

View Full Product Details