Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70366903-70367401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNMB4
Entrez Gene ID
27345
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 4
Gene Aliases
-
UniGene
Hs.525529
Gene Chromosome Location
Chr.12: 70366220-70434292 on Build GRCh38
Transcripts
NM_014505.5 XM_011538188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1299836 c.234C>T p.G78G Chr.12: 70366968-70366968 on Build GRCh38
1606578 c.189C>T p.G63G Chr.12: 70366923-70366923 on Build GRCh38
4817662 c.190G>A p.E64K Chr.12: 70366924-70366924 on Build GRCh38
376606 c.301C>G p.H101D Chr.12: 70367035-70367035 on Build GRCh38
3936114 c.277G>A p.E93K Chr.12: 70367011-70367011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753114704 Chr.12: 70367087-70367087 on Build GRCh38
rs570785511 Chr.12: 70367231-70367231 on Build GRCh38
rs11178200 Chr.12: 70366983-70366983 on Build GRCh38
rs3782388 Chr.12: 70367345-70367345 on Build GRCh38
rs769440472 Chr.12: 70367048-70367048 on Build GRCh38
rs902721759 Chr.12: 70367350-70367350 on Build GRCh38
rs950440566 Chr.12: 70367158-70367158 on Build GRCh38
rs574187097 Chr.12: 70367248-70367248 on Build GRCh38
rs751389729 Chr.12: 70367080-70367081 on Build GRCh38
rs541631620 Chr.12: 70367259-70367259 on Build GRCh38
rs992552434 Chr.12: 70367169-70367169 on Build GRCh38
rs989526356 Chr.12: 70367292-70367292 on Build GRCh38
rs746435834 Chr.12: 70367017-70367017 on Build GRCh38
rs767797869 Chr.12: 70367094-70367095 on Build GRCh38
rs376983672 Chr.12: 70367218-70367218 on Build GRCh38
rs779568548 Chr.12: 70367004-70367004 on Build GRCh38
rs558285950 Chr.12: 70367079-70367079 on Build GRCh38
rs758048301 Chr.12: 70367106-70367106 on Build GRCh38
rs775907060 Chr.12: 70367311-70367311 on Build GRCh38
rs754683619 Chr.12: 70367083-70367084 on Build GRCh38
rs940081117 Chr.12: 70367184-70367184 on Build GRCh38
rs773614651 Chr.12: 70367055-70367055 on Build GRCh38
rs185269693 Chr.12: 70367234-70367234 on Build GRCh38
rs745499444 Chr.12: 70366964-70366964 on Build GRCh38
rs537420950 Chr.12: 70367213-70367213 on Build GRCh38
rs11178201 Chr.12: 70367294-70367294 on Build GRCh38
rs768650937 Chr.12: 70367019-70367019 on Build GRCh38
rs759524842 Chr.12: 70367086-70367086 on Build GRCh38
rs920015226 Chr.12: 70367236-70367236 on Build GRCh38
rs763288315 Chr.12: 70367065-70367065 on Build GRCh38
rs753166736 Chr.12: 70366933-70366933 on Build GRCh38
rs1017555615 Chr.12: 70367129-70367129 on Build GRCh38
rs778577165 Chr.12: 70366957-70366957 on Build GRCh38
rs766643802 Chr.12: 70367067-70367067 on Build GRCh38
rs754163598 Chr.12: 70367103-70367103 on Build GRCh38
rs945419907 Chr.12: 70367308-70367308 on Build GRCh38
rs764466266 Chr.12: 70367100-70367100 on Build GRCh38
rs760194449 Chr.12: 70367265-70367265 on Build GRCh38
rs936651527 Chr.12: 70367291-70367291 on Build GRCh38
rs1001013809 Chr.12: 70367364-70367364 on Build GRCh38
rs767956663 Chr.12: 70366995-70366995 on Build GRCh38
rs970635398 Chr.12: 70367091-70367091 on Build GRCh38
rs959103639 Chr.12: 70366986-70366986 on Build GRCh38
rs546044677 Chr.12: 70367336-70367336 on Build GRCh38
rs188524736 Chr.12: 70367284-70367284 on Build GRCh38
rs776418718 Chr.12: 70367022-70367022 on Build GRCh38
rs756431148 Chr.12: 70366938-70366938 on Build GRCh38
rs772715409 Chr.12: 70367153-70367153 on Build GRCh38
rs766275506 Chr.12: 70367078-70367079 on Build GRCh38
rs898822236 Chr.12: 70367134-70367134 on Build GRCh38
rs748037323 Chr.12: 70367028-70367028 on Build GRCh38
rs368037458 Chr.12: 70367160-70367160 on Build GRCh38
rs994535126 Chr.12: 70367139-70367139 on Build GRCh38
rs757999060 Chr.12: 70366987-70366987 on Build GRCh38
rs1038488826 Chr.12: 70367205-70367205 on Build GRCh38
rs756551493 Chr.12: 70367097-70367097 on Build GRCh38
rs779623390 Chr.12: 70367115-70367115 on Build GRCh38
rs1008514497 Chr.12: 70367328-70367328 on Build GRCh38

View Full Product Details