Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98744054-98744564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
UniGene
Hs.506458
Gene Chromosome Location
Chr.12: 98729904-99984727 on Build GRCh38
Transcripts
NM_001204065.1 NM_001204066.1 NM_001204067.1 NM_001204068.1 NM_001204069.1 NM_001204070.1 NM_001204079.1 NM_001204080.1 NM_001204081.1 NM_020140.3 NM_152788.4 NM_181670.3 XM_006719507.3 XM_006719514.3 XM_017019651.1 XM_017019653.1 XM_017019654.1 XM_017019658.1

SNP Detail

SNP ID Chromosome Location
rs80297723 Chr.12: 98744482-98744482 on Build GRCh38
rs548666666 Chr.12: 98744443-98744443 on Build GRCh38
rs991535950 Chr.12: 98744383-98744383 on Build GRCh38
rs398044706 Chr.12: 98744382-98744383 on Build GRCh38
rs981187422 Chr.12: 98744342-98744342 on Build GRCh38
rs571574107 Chr.12: 98744409-98744409 on Build GRCh38
rs577091942 Chr.12: 98744413-98744413 on Build GRCh38
rs1002111548 Chr.12: 98744199-98744199 on Build GRCh38
rs947291753 Chr.12: 98744450-98744450 on Build GRCh38
rs765655912 Chr.12: 98744348-98744351 on Build GRCh38
rs947239360 Chr.12: 98744131-98744131 on Build GRCh38
rs934776302 Chr.12: 98744080-98744080 on Build GRCh38
rs34975028 Chr.12: 98744375-98744376 on Build GRCh38
rs951145836 Chr.12: 98744277-98744278 on Build GRCh38
rs1018542089 Chr.12: 98744210-98744210 on Build GRCh38
rs903020961 Chr.12: 98744472-98744472 on Build GRCh38
rs1007268180 Chr.12: 98744209-98744209 on Build GRCh38
rs1052666 Chr.12: 98744096-98744096 on Build GRCh38
rs893146978 Chr.12: 98744098-98744098 on Build GRCh38
rs989316370 Chr.12: 98744489-98744489 on Build GRCh38
rs962700887 Chr.12: 98744332-98744332 on Build GRCh38
rs933114849 Chr.12: 98744473-98744473 on Build GRCh38
rs1029657388 Chr.12: 98744190-98744190 on Build GRCh38
rs912754252 Chr.12: 98744465-98744465 on Build GRCh38
rs745479268 Chr.12: 98744429-98744429 on Build GRCh38
rs1015313927 Chr.12: 98744279-98744279 on Build GRCh38
rs1003980675 Chr.12: 98744271-98744271 on Build GRCh38
rs550731204 Chr.12: 98744108-98744109 on Build GRCh38
rs181420833 Chr.12: 98744150-98744150 on Build GRCh38
rs1011550937 Chr.12: 98744119-98744119 on Build GRCh38
rs769654481 Chr.12: 98744431-98744431 on Build GRCh38
rs541621334 Chr.12: 98744516-98744516 on Build GRCh38
rs987001001 Chr.12: 98744454-98744454 on Build GRCh38
rs1053114474 Chr.12: 98744093-98744093 on Build GRCh38
rs893012618 Chr.12: 98744123-98744123 on Build GRCh38
rs1034878397 Chr.12: 98744241-98744241 on Build GRCh38
rs963265663 Chr.12: 98744218-98744218 on Build GRCh38
rs905071754 Chr.12: 98744185-98744185 on Build GRCh38
rs974262636 Chr.12: 98744235-98744235 on Build GRCh38
rs1025805292 Chr.12: 98744265-98744265 on Build GRCh38
rs1041564956 Chr.12: 98744460-98744460 on Build GRCh38
rs954329618 Chr.12: 98744426-98744426 on Build GRCh38
rs896300453 Chr.12: 98744268-98744268 on Build GRCh38
rs76399902 Chr.12: 98744365-98744365 on Build GRCh38
rs999556110 Chr.12: 98744182-98744182 on Build GRCh38
rs544211127 Chr.12: 98744418-98744418 on Build GRCh38
rs1041511661 Chr.12: 98744145-98744145 on Build GRCh38
rs902633194 Chr.12: 98744181-98744181 on Build GRCh38

View Full Product Details