Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37009342-37009886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC8
Entrez Gene ID
11340
Gene Name
exosome component 8
Gene Aliases
CIP3, EAP2, OIP2, PCH1C, RRP43, Rrp43p, bA421P11.3, p9
UniGene
Hs.294041
Gene Chromosome Location
Chr.13: 37000373-37009614 on Build GRCh38
Transcripts
NM_181503.2 XM_006719763.1 NM_001014286.2 NM_001278480.1 NM_001278481.1 NM_001278482.1 NM_017569.3 XM_005266447.2 XM_005266449.3 XM_005266451.3 XM_005266454.3 XM_005266455.2 XM_005266456.3 XM_005266457.3 XM_005266458.3 XM_005266460.2 XM_005266461.3 XM_005266462.4 XM_005266464.3 XM_005266465.2 XM_005266467.3 XM_017020653.1 XM_017020654.1 XM_017020655.1 XM_017020656.1 XM_017020657.1 XM_017020658.1 XM_017020659.1 XM_017020660.1 XM_017020661.1

Gene Symbol
SUPT20H
Entrez Gene ID
55578
Gene Name
SPT20 homolog, SAGA complex component
Gene Aliases
C13, C13orf19, FAM48A, FP757, P38IP, SPT20
UniGene
Hs.435815
Gene Chromosome Location
Chr.13: 37009312-37059714 on Build GRCh38
Transcripts
NM_181503.2 XM_006719763.1 NM_001014286.2 NM_001278480.1 NM_001278481.1 NM_001278482.1 NM_017569.3 XM_005266447.2 XM_005266449.3 XM_005266451.3 XM_005266454.3 XM_005266455.2 XM_005266456.3 XM_005266457.3 XM_005266458.3 XM_005266460.2 XM_005266461.3 XM_005266462.4 XM_005266464.3 XM_005266465.2 XM_005266467.3 XM_017020653.1 XM_017020654.1 XM_017020655.1 XM_017020656.1 XM_017020657.1 XM_017020658.1 XM_017020659.1 XM_017020660.1 XM_017020661.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947166 c.2336T>G p.F779C Chr.13: 37009676-37009676 on Build GRCh38
3885307 c.2140A>G p.S714G Chr.13: 37009771-37009771 on Build GRCh38
3813775 c.2214C>T p.F738F Chr.13: 37009798-37009798 on Build GRCh38
1206364 c.2287A>G p.S763G Chr.13: 37009725-37009725 on Build GRCh38
6793242 c.2108C>T p.A703V Chr.13: 37009803-37009803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs9594185 Chr.13: 37009675-37009675 on Build GRCh38
rs780703867 Chr.13: 37009724-37009725 on Build GRCh38
rs766826291 Chr.13: 37009767-37009767 on Build GRCh38
rs1039836040 Chr.13: 37009541-37009541 on Build GRCh38
rs367722311 Chr.13: 37009859-37009859 on Build GRCh38
rs9469 Chr.13: 37009694-37009694 on Build GRCh38
rs774070601 Chr.13: 37009601-37009601 on Build GRCh38
rs970475975 Chr.13: 37009485-37009485 on Build GRCh38
rs185586458 Chr.13: 37009514-37009514 on Build GRCh38
rs760534356 Chr.13: 37009616-37009616 on Build GRCh38
rs768129917 Chr.13: 37009570-37009570 on Build GRCh38
rs1048479353 Chr.13: 37009614-37009614 on Build GRCh38
rs776643456 Chr.13: 37009728-37009728 on Build GRCh38
rs762892647 Chr.13: 37009730-37009730 on Build GRCh38
rs750307293 Chr.13: 37009588-37009589 on Build GRCh38
rs9566173 Chr.13: 37009820-37009820 on Build GRCh38
rs766802613 Chr.13: 37009856-37009856 on Build GRCh38
rs79506869 Chr.13: 37009831-37009831 on Build GRCh38
rs773864661 Chr.13: 37009808-37009808 on Build GRCh38
rs934339019 Chr.13: 37009466-37009466 on Build GRCh38
rs750722882 Chr.13: 37009747-37009747 on Build GRCh38
rs1016903370 Chr.13: 37009594-37009597 on Build GRCh38
rs981852250 Chr.13: 37009554-37009554 on Build GRCh38
rs918297140 Chr.13: 37009560-37009560 on Build GRCh38
rs149036783 Chr.13: 37009737-37009737 on Build GRCh38
rs764165058 Chr.13: 37009647-37009647 on Build GRCh38
rs745866589 Chr.13: 37009818-37009818 on Build GRCh38
rs773195358 Chr.13: 37009699-37009699 on Build GRCh38
rs1007337116 Chr.13: 37009537-37009537 on Build GRCh38
rs999543827 Chr.13: 37009774-37009774 on Build GRCh38
rs915739391 Chr.13: 37009651-37009651 on Build GRCh38
rs746937023 Chr.13: 37009700-37009700 on Build GRCh38
rs191078501 Chr.13: 37009815-37009815 on Build GRCh38
rs750919009 Chr.13: 37009669-37009669 on Build GRCh38
rs754411778 Chr.13: 37009778-37009778 on Build GRCh38
rs974581658 Chr.13: 37009857-37009857 on Build GRCh38
rs902550232 Chr.13: 37009755-37009755 on Build GRCh38
rs924299547 Chr.13: 37009382-37009382 on Build GRCh38
rs200334194 Chr.13: 37009821-37009821 on Build GRCh38
rs751828292 Chr.13: 37009654-37009654 on Build GRCh38
rs892863838 Chr.13: 37009506-37009506 on Build GRCh38
rs746928009 Chr.13: 37009593-37009597 on Build GRCh38
rs201986233 Chr.13: 37009664-37009664 on Build GRCh38
rs990684783 Chr.13: 37009381-37009381 on Build GRCh38
rs11559100 Chr.13: 37009639-37009639 on Build GRCh38
rs752891385 Chr.13: 37009644-37009644 on Build GRCh38
rs758187788 Chr.13: 37009786-37009786 on Build GRCh38
rs998782618 Chr.13: 37009783-37009783 on Build GRCh38
rs929611937 Chr.13: 37009569-37009569 on Build GRCh38
rs770923289 Chr.13: 37009714-37009714 on Build GRCh38
rs775408573 Chr.13: 37009627-37009627 on Build GRCh38
rs981883340 Chr.13: 37009823-37009823 on Build GRCh38
rs756682989 Chr.13: 37009670-37009670 on Build GRCh38
rs1054194738 Chr.13: 37009505-37009505 on Build GRCh38
rs773248513 Chr.13: 37009636-37009636 on Build GRCh38
rs764275231 Chr.13: 37009735-37009735 on Build GRCh38
rs143011049 Chr.13: 37009780-37009780 on Build GRCh38
rs370974587 Chr.13: 37009802-37009802 on Build GRCh38
rs754403317 Chr.13: 37009772-37009772 on Build GRCh38
rs778815592 Chr.13: 37009582-37009583 on Build GRCh38
rs773448527 Chr.13: 37009834-37009834 on Build GRCh38
rs771928651 Chr.13: 37009623-37009623 on Build GRCh38
rs758257851 Chr.13: 37009589-37009589 on Build GRCh38
rs769476262 Chr.13: 37009755-37009756 on Build GRCh38
rs779988673 Chr.13: 37009589-37009592 on Build GRCh38
rs371008112 Chr.13: 37009840-37009840 on Build GRCh38
rs918328251 Chr.13: 37009824-37009824 on Build GRCh38
rs748692017 Chr.13: 37009680-37009680 on Build GRCh38
rs768238164 Chr.13: 37009683-37009683 on Build GRCh38
rs981364745 Chr.13: 37009666-37009666 on Build GRCh38
rs763096378 Chr.13: 37009646-37009646 on Build GRCh38
rs768174756 Chr.13: 37009595-37009597 on Build GRCh38
rs377592079 Chr.13: 37009779-37009779 on Build GRCh38
rs997594858 Chr.13: 37009734-37009734 on Build GRCh38
rs747779312 Chr.13: 37009736-37009737 on Build GRCh38
rs746740892 Chr.13: 37009803-37009803 on Build GRCh38
rs761470265 Chr.13: 37009621-37009621 on Build GRCh38
rs916358328 Chr.13: 37009470-37009470 on Build GRCh38
rs538492102 Chr.13: 37009667-37009667 on Build GRCh38
rs750163497 Chr.13: 37009578-37009581 on Build GRCh38
rs778385572 Chr.13: 37009687-37009687 on Build GRCh38
rs540927550 Chr.13: 37009431-37009431 on Build GRCh38
rs747870909 Chr.13: 37009693-37009693 on Build GRCh38
rs756486975 Chr.13: 37009759-37009759 on Build GRCh38
rs551506424 Chr.13: 37009629-37009629 on Build GRCh38
rs762154234 Chr.13: 37009736-37009736 on Build GRCh38
rs368307804 Chr.13: 37009833-37009833 on Build GRCh38

View Full Product Details