Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48233021-48233512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITM2B
Entrez Gene ID
9445
Gene Name
integral membrane protein 2B
Gene Aliases
ABRI, BRI, BRI2, BRICD2B, E25B, E3-16, FBD, RDGCA, imBRI2
UniGene
Hs.643683
Gene Chromosome Location
Chr.13: 48233138-48262096 on Build GRCh38
Transcripts
NM_021999.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7372715 c.70G>A p.E24K Chr.13: 48233430-48233430 on Build GRCh38
5498107 c.110A>G p.D37G Chr.13: 48233470-48233470 on Build GRCh38
6255029 c.105G>T p.A35A Chr.13: 48233465-48233465 on Build GRCh38
6406224 c.26C>A p.A9D Chr.13: 48233386-48233386 on Build GRCh38
4964228 c.68G>C p.G23A Chr.13: 48233428-48233428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778187781 Chr.13: 48233420-48233420 on Build GRCh38
rs919771742 Chr.13: 48233293-48233293 on Build GRCh38
rs1017472591 Chr.13: 48233212-48233212 on Build GRCh38
rs1025618824 Chr.13: 48233206-48233206 on Build GRCh38
rs922869576 Chr.13: 48233106-48233106 on Build GRCh38
rs11556903 Chr.13: 48233298-48233298 on Build GRCh38
rs539024187 Chr.13: 48233219-48233219 on Build GRCh38
rs536989630 Chr.13: 48233256-48233256 on Build GRCh38
rs912917846 Chr.13: 48233079-48233079 on Build GRCh38
rs751333259 Chr.13: 48233453-48233453 on Build GRCh38
rs1008439046 Chr.13: 48233101-48233101 on Build GRCh38
rs748552422 Chr.13: 48233385-48233385 on Build GRCh38
rs572688262 Chr.13: 48233251-48233251 on Build GRCh38
rs909030865 Chr.13: 48233045-48233045 on Build GRCh38
rs942475327 Chr.13: 48233253-48233253 on Build GRCh38
rs150336652 Chr.13: 48233452-48233452 on Build GRCh38
rs910348564 Chr.13: 48233301-48233320 on Build GRCh38
rs11556899 Chr.13: 48233441-48233441 on Build GRCh38
rs963233596 Chr.13: 48233215-48233215 on Build GRCh38
rs1016942995 Chr.13: 48233402-48233402 on Build GRCh38
rs945727692 Chr.13: 48233235-48233235 on Build GRCh38
rs1004041059 Chr.13: 48233072-48233072 on Build GRCh38
rs908183138 Chr.13: 48233276-48233276 on Build GRCh38
rs746714921 Chr.13: 48233426-48233426 on Build GRCh38
rs772385941 Chr.13: 48233415-48233415 on Build GRCh38
rs745799929 Chr.13: 48233395-48233397 on Build GRCh38
rs779515110 Chr.13: 48233447-48233447 on Build GRCh38
rs531403426 Chr.13: 48233324-48233325 on Build GRCh38
rs761013044 Chr.13: 48233465-48233465 on Build GRCh38
rs565013505 Chr.13: 48233399-48233399 on Build GRCh38
rs991806046 Chr.13: 48233260-48233260 on Build GRCh38
rs1047366668 Chr.13: 48233354-48233354 on Build GRCh38
rs769217205 Chr.13: 48233232-48233232 on Build GRCh38
rs932769468 Chr.13: 48233285-48233285 on Build GRCh38
rs189827935 Chr.13: 48233053-48233053 on Build GRCh38
rs552198156 Chr.13: 48233351-48233351 on Build GRCh38
rs765109304 Chr.13: 48233316-48233316 on Build GRCh38
rs375837803 Chr.13: 48233203-48233203 on Build GRCh38
rs11556905 Chr.13: 48233403-48233403 on Build GRCh38
rs543509091 Chr.13: 48233375-48233375 on Build GRCh38
rs939930230 Chr.13: 48233290-48233290 on Build GRCh38
rs991878027 Chr.13: 48233472-48233472 on Build GRCh38
rs886979110 Chr.13: 48233069-48233069 on Build GRCh38
rs182153640 Chr.13: 48233082-48233082 on Build GRCh38
rs576743598 Chr.13: 48233355-48233355 on Build GRCh38
rs926317666 Chr.13: 48233189-48233189 on Build GRCh38
rs532372951 Chr.13: 48233264-48233264 on Build GRCh38
rs1053067147 Chr.13: 48233329-48233329 on Build GRCh38
rs955521190 Chr.13: 48233098-48233098 on Build GRCh38
rs879791684 Chr.13: 48233163-48233163 on Build GRCh38
rs938607697 Chr.13: 48233057-48233057 on Build GRCh38
rs1002096122 Chr.13: 48233438-48233438 on Build GRCh38
rs1005614518 Chr.13: 48233209-48233209 on Build GRCh38
rs990266865 Chr.13: 48233115-48233120 on Build GRCh38
rs970598991 Chr.13: 48233240-48233240 on Build GRCh38
rs757960036 Chr.13: 48233394-48233395 on Build GRCh38
rs1039949603 Chr.13: 48233198-48233198 on Build GRCh38
rs891194263 Chr.13: 48233323-48233323 on Build GRCh38
rs1056685580 Chr.13: 48233073-48233073 on Build GRCh38
rs549899044 Chr.13: 48233195-48233195 on Build GRCh38
rs978533715 Chr.13: 48233246-48233246 on Build GRCh38
rs1007510253 Chr.13: 48233194-48233194 on Build GRCh38
rs967842571 Chr.13: 48233149-48233149 on Build GRCh38
rs763943060 Chr.13: 48233350-48233350 on Build GRCh38
rs775954363 Chr.13: 48233051-48233051 on Build GRCh38
rs766663901 Chr.13: 48233466-48233466 on Build GRCh38
rs866736004 Chr.13: 48233391-48233391 on Build GRCh38
rs954129082 Chr.13: 48233077-48233078 on Build GRCh38
rs752549116 Chr.13: 48233336-48233336 on Build GRCh38
rs537928273 Chr.13: 48233102-48233102 on Build GRCh38
rs769773974 Chr.13: 48233448-48233448 on Build GRCh38
rs758987944 Chr.13: 48233445-48233445 on Build GRCh38
rs535639014 Chr.13: 48233059-48233059 on Build GRCh38
rs746700992 Chr.13: 48233446-48233447 on Build GRCh38
rs866375807 Chr.13: 48233388-48233388 on Build GRCh38
rs992716865 Chr.13: 48233227-48233227 on Build GRCh38
rs914998660 Chr.13: 48233125-48233126 on Build GRCh38
rs904672718 Chr.13: 48233182-48233182 on Build GRCh38
rs984622932 Chr.13: 48233044-48233044 on Build GRCh38
rs554126468 Chr.13: 48233221-48233221 on Build GRCh38
rs898292453 Chr.13: 48233210-48233210 on Build GRCh38
rs571519655 Chr.13: 48233207-48233207 on Build GRCh38
rs11556906 Chr.13: 48233356-48233356 on Build GRCh38
rs115420240 Chr.13: 48233048-48233048 on Build GRCh38
rs1041291165 Chr.13: 48233100-48233100 on Build GRCh38
rs959924836 Chr.13: 48233224-48233224 on Build GRCh38
rs1056356124 Chr.13: 48233056-48233056 on Build GRCh38
rs980307830 Chr.13: 48233220-48233220 on Build GRCh38
rs758061168 Chr.13: 48233346-48233346 on Build GRCh38
rs940240491 Chr.13: 48233095-48233095 on Build GRCh38
rs770539704 Chr.13: 48233433-48233433 on Build GRCh38
rs934215670 Chr.13: 48233307-48233307 on Build GRCh38
rs891169153 Chr.13: 48233214-48233214 on Build GRCh38
rs560135420 Chr.13: 48233126-48233126 on Build GRCh38
rs753982738 Chr.13: 48233469-48233469 on Build GRCh38
rs998443443 Chr.13: 48233205-48233205 on Build GRCh38
rs780316156 Chr.13: 48233358-48233358 on Build GRCh38
rs980902008 Chr.13: 48233157-48233157 on Build GRCh38
rs986390108 Chr.13: 48233114-48233130 on Build GRCh38
rs779234032 Chr.13: 48233380-48233380 on Build GRCh38
rs1043227717 Chr.13: 48233180-48233180 on Build GRCh38
rs11556904 Chr.13: 48233349-48233349 on Build GRCh38
rs901362811 Chr.13: 48233292-48233292 on Build GRCh38
rs939010646 Chr.13: 48233192-48233192 on Build GRCh38

View Full Product Details