Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 96838648-96839177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS6ST3
Entrez Gene ID
266722
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
HS6ST-3
UniGene
Hs.171001
Gene Chromosome Location
Chr.13: 96090634-96839562 on Build GRCh38
Transcripts
NM_153456.3

SNP Detail

SNP ID Chromosome Location
rs544911825 Chr.13: 96839090-96839090 on Build GRCh38
rs114193572 Chr.13: 96838679-96838679 on Build GRCh38
rs144098829 Chr.13: 96838816-96838816 on Build GRCh38
rs79521595 Chr.13: 96838893-96838893 on Build GRCh38
rs141305557 Chr.13: 96838949-96838949 on Build GRCh38
rs77275541 Chr.13: 96838896-96838896 on Build GRCh38
rs534652440 Chr.13: 96839110-96839110 on Build GRCh38
rs971569082 Chr.13: 96838831-96838831 on Build GRCh38
rs539912396 Chr.13: 96838717-96838717 on Build GRCh38
rs932670744 Chr.13: 96839047-96839047 on Build GRCh38
rs981603033 Chr.13: 96838975-96838975 on Build GRCh38
rs956379725 Chr.13: 96838997-96838997 on Build GRCh38
rs897608503 Chr.13: 96838909-96838909 on Build GRCh38
rs916136842 Chr.13: 96838683-96838683 on Build GRCh38
rs1018783381 Chr.13: 96838821-96838821 on Build GRCh38
rs569810317 Chr.13: 96838846-96838846 on Build GRCh38
rs892010694 Chr.13: 96838804-96838804 on Build GRCh38
rs771437218 Chr.13: 96839071-96839071 on Build GRCh38
rs146474611 Chr.13: 96838871-96838871 on Build GRCh38
rs951761412 Chr.13: 96838935-96838935 on Build GRCh38
rs1054417590 Chr.13: 96839068-96839068 on Build GRCh38
rs1049755873 Chr.13: 96839051-96839051 on Build GRCh38
rs1001705395 Chr.13: 96838848-96838848 on Build GRCh38
rs1024931571 Chr.13: 96838818-96838818 on Build GRCh38
rs750897676 Chr.13: 96839129-96839129 on Build GRCh38
rs759283431 Chr.13: 96839038-96839038 on Build GRCh38
rs1050373241 Chr.13: 96838782-96838782 on Build GRCh38
rs771514133 Chr.13: 96838887-96838888 on Build GRCh38
rs1054886215 Chr.13: 96838721-96838721 on Build GRCh38
rs182239262 Chr.13: 96839035-96839035 on Build GRCh38
rs990262634 Chr.13: 96839001-96839001 on Build GRCh38
rs867873035 Chr.13: 96839086-96839086 on Build GRCh38
rs889796443 Chr.13: 96838713-96838713 on Build GRCh38
rs888995270 Chr.13: 96838797-96838797 on Build GRCh38
rs1012965239 Chr.13: 96838944-96838944 on Build GRCh38
rs975979296 Chr.13: 96838970-96838970 on Build GRCh38
rs914606372 Chr.13: 96839041-96839041 on Build GRCh38
rs557781301 Chr.13: 96839125-96839125 on Build GRCh38
rs773846043 Chr.13: 96839029-96839029 on Build GRCh38
rs765578414 Chr.13: 96838962-96838962 on Build GRCh38
rs924730527 Chr.13: 96838711-96838711 on Build GRCh38
rs932614551 Chr.13: 96838740-96838740 on Build GRCh38
rs950229105 Chr.13: 96838703-96838703 on Build GRCh38
rs770260692 Chr.13: 96838910-96838910 on Build GRCh38
rs764270243 Chr.13: 96838724-96838724 on Build GRCh38
rs535261871 Chr.13: 96838942-96838942 on Build GRCh38
rs924763180 Chr.13: 96838986-96838986 on Build GRCh38
rs188363339 Chr.13: 96838710-96838710 on Build GRCh38
rs1027795098 Chr.13: 96838913-96838913 on Build GRCh38
rs911270784 Chr.13: 96839057-96839057 on Build GRCh38
rs934094931 Chr.13: 96838698-96838698 on Build GRCh38
rs762106147 Chr.13: 96838915-96838915 on Build GRCh38
rs910505346 Chr.13: 96839058-96839058 on Build GRCh38

View Full Product Details