Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23117228-23117738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPE
Entrez Gene ID
1053
Gene Name
CCAAT/enhancer binding protein epsilon
Gene Aliases
C/EBP-epsilon, CRP1
UniGene
Hs.558308
Gene Chromosome Location
Chr.14: 23117306-23119611 on Build GRCh38
Transcripts
NM_001805.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625966 c.717C>T p.Y239Y Chr.14: 23117616-23117616 on Build GRCh38
6625964 c.622C>T p.R208W Chr.14: 23117711-23117711 on Build GRCh38
5560821 c.603C>G p.N201K Chr.14: 23117730-23117730 on Build GRCh38
6625962 c.618G>A p.E206E Chr.14: 23117715-23117715 on Build GRCh38
6625960 c.761C>T p.T254I Chr.14: 23117572-23117572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367795312 Chr.14: 23117622-23117622 on Build GRCh38
rs923787623 Chr.14: 23117638-23117638 on Build GRCh38
rs755654290 Chr.14: 23117561-23117561 on Build GRCh38
rs747524697 Chr.14: 23117680-23117680 on Build GRCh38
rs370718175 Chr.14: 23117500-23117500 on Build GRCh38
rs774467197 Chr.14: 23117435-23117435 on Build GRCh38
rs1051640040 Chr.14: 23117603-23117603 on Build GRCh38
rs780345897 Chr.14: 23117654-23117654 on Build GRCh38
rs772501158 Chr.14: 23117594-23117594 on Build GRCh38
rs74036916 Chr.14: 23117248-23117248 on Build GRCh38
rs746944840 Chr.14: 23117486-23117486 on Build GRCh38
rs750263943 Chr.14: 23117459-23117459 on Build GRCh38
rs754942176 Chr.14: 23117552-23117552 on Build GRCh38
rs779209834 Chr.14: 23117587-23117587 on Build GRCh38
rs766104489 Chr.14: 23117547-23117547 on Build GRCh38
rs56087057 Chr.14: 23117286-23117289 on Build GRCh38
rs953065043 Chr.14: 23117323-23117323 on Build GRCh38
rs774719241 Chr.14: 23117523-23117523 on Build GRCh38
rs143030663 Chr.14: 23117359-23117359 on Build GRCh38
rs374712257 Chr.14: 23117610-23117610 on Build GRCh38
rs1015773527 Chr.14: 23117532-23117534 on Build GRCh38
rs201609537 Chr.14: 23117621-23117621 on Build GRCh38
rs143753215 Chr.14: 23117505-23117505 on Build GRCh38
rs71425065 Chr.14: 23117248-23117249 on Build GRCh38
rs112420613 Chr.14: 23117472-23117472 on Build GRCh38
rs141185489 Chr.14: 23117709-23117709 on Build GRCh38
rs746230262 Chr.14: 23117588-23117588 on Build GRCh38
rs756067667 Chr.14: 23117625-23117625 on Build GRCh38
rs761128015 Chr.14: 23117600-23117600 on Build GRCh38
rs538148561 Chr.14: 23117525-23117525 on Build GRCh38
rs750122474 Chr.14: 23117637-23117637 on Build GRCh38
rs775604463 Chr.14: 23117537-23117537 on Build GRCh38
rs768742384 Chr.14: 23117604-23117604 on Build GRCh38
rs748972715 Chr.14: 23117582-23117582 on Build GRCh38
rs764284282 Chr.14: 23117455-23117455 on Build GRCh38
rs566415335 Chr.14: 23117394-23117394 on Build GRCh38
rs375672870 Chr.14: 23117694-23117694 on Build GRCh38
rs933197835 Chr.14: 23117260-23117260 on Build GRCh38
rs541785349 Chr.14: 23117312-23117313 on Build GRCh38
rs867269245 Chr.14: 23117522-23117522 on Build GRCh38
rs3222520 Chr.14: 23117250-23117251 on Build GRCh38
rs959995195 Chr.14: 23117256-23117257 on Build GRCh38
rs915188324 Chr.14: 23117440-23117440 on Build GRCh38
rs57969698 Chr.14: 23117289-23117290 on Build GRCh38
rs771232876 Chr.14: 23117518-23117518 on Build GRCh38
rs775939975 Chr.14: 23117599-23117599 on Build GRCh38
rs769963026 Chr.14: 23117703-23117703 on Build GRCh38
rs372651475 Chr.14: 23117653-23117653 on Build GRCh38
rs777380202 Chr.14: 23117580-23117580 on Build GRCh38
rs768064135 Chr.14: 23117532-23117532 on Build GRCh38
rs984341714 Chr.14: 23117346-23117346 on Build GRCh38
rs757448387 Chr.14: 23117543-23117543 on Build GRCh38
rs889079447 Chr.14: 23117301-23117301 on Build GRCh38
rs763046578 Chr.14: 23117710-23117710 on Build GRCh38
rs149444043 Chr.14: 23117491-23117491 on Build GRCh38
rs756917368 Chr.14: 23117636-23117636 on Build GRCh38
rs769087828 Chr.14: 23117682-23117682 on Build GRCh38
rs908701161 Chr.14: 23117355-23117355 on Build GRCh38
rs761795732 Chr.14: 23117439-23117439 on Build GRCh38
rs756394880 Chr.14: 23117448-23117448 on Build GRCh38
rs961119813 Chr.14: 23117360-23117360 on Build GRCh38
rs1013141996 Chr.14: 23117289-23117289 on Build GRCh38
rs1026384076 Chr.14: 23117402-23117402 on Build GRCh38
rs374665903 Chr.14: 23117478-23117478 on Build GRCh38
rs968427857 Chr.14: 23117497-23117497 on Build GRCh38
rs1035655436 Chr.14: 23117256-23117256 on Build GRCh38
rs754087983 Chr.14: 23117540-23117540 on Build GRCh38
rs776758085 Chr.14: 23117557-23117558 on Build GRCh38
rs777425212 Chr.14: 23117630-23117630 on Build GRCh38
rs978378493 Chr.14: 23117635-23117635 on Build GRCh38
rs758156324 Chr.14: 23117468-23117468 on Build GRCh38
rs778542611 Chr.14: 23117498-23117498 on Build GRCh38
rs961511817 Chr.14: 23117535-23117535 on Build GRCh38
rs747660576 Chr.14: 23117504-23117504 on Build GRCh38
rs899230959 Chr.14: 23117392-23117392 on Build GRCh38
rs529617986 Chr.14: 23117288-23117288 on Build GRCh38
rs914888795 Chr.14: 23117611-23117611 on Build GRCh38
rs1049404497 Chr.14: 23117405-23117405 on Build GRCh38
rs55722931 Chr.14: 23117586-23117586 on Build GRCh38
rs751372334 Chr.14: 23117615-23117615 on Build GRCh38
rs199967428 Chr.14: 23117539-23117539 on Build GRCh38
rs113340378 Chr.14: 23117305-23117305 on Build GRCh38
rs767445685 Chr.14: 23117711-23117711 on Build GRCh38
rs117800045 Chr.14: 23117447-23117447 on Build GRCh38
rs746255618 Chr.14: 23117645-23117645 on Build GRCh38
rs1037676059 Chr.14: 23117373-23117373 on Build GRCh38
rs761948458 Chr.14: 23117614-23117614 on Build GRCh38
rs751304427 Chr.14: 23117551-23117551 on Build GRCh38
rs1007967803 Chr.14: 23117313-23117313 on Build GRCh38
rs772978298 Chr.14: 23117517-23117517 on Build GRCh38
rs763610496 Chr.14: 23117601-23117601 on Build GRCh38
rs753381359 Chr.14: 23117631-23117631 on Build GRCh38

View Full Product Details