Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34535944-34536438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAPP
Entrez Gene ID
55837
Gene Name
E2F associated phosphoprotein
Gene Aliases
BM036, C14orf11
UniGene
Hs.433269
Gene Chromosome Location
Chr.14: 34515929-34539737 on Build GRCh38
Transcripts
NM_001318916.1 NM_018453.3 XM_011536958.1 XM_017021460.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2153659 c.124C>T p.R42* Chr.14: 34536226-34536226 on Build GRCh38
3886026 c.112C>T p.P38S Chr.14: 34536238-34536238 on Build GRCh38
3495798 c.247C>T p.L83L Chr.14: 34536103-34536103 on Build GRCh38
3495799 c.117C>T p.D39D Chr.14: 34536233-34536233 on Build GRCh38
6651111 c.123delA p.K41fs*29 Chr.14: 34536227-34536227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775468089 Chr.14: 34536068-34536068 on Build GRCh38
rs964841410 Chr.14: 34536308-34536308 on Build GRCh38
rs369013236 Chr.14: 34536415-34536415 on Build GRCh38
rs995014676 Chr.14: 34536246-34536246 on Build GRCh38
rs761848134 Chr.14: 34536124-34536124 on Build GRCh38
rs932407736 Chr.14: 34536064-34536064 on Build GRCh38
rs554875909 Chr.14: 34536104-34536104 on Build GRCh38
rs771879397 Chr.14: 34536162-34536162 on Build GRCh38
rs1045904088 Chr.14: 34536415-34536416 on Build GRCh38
rs369451332 Chr.14: 34536264-34536264 on Build GRCh38
rs754374098 Chr.14: 34536226-34536226 on Build GRCh38
rs946541301 Chr.14: 34536385-34536385 on Build GRCh38
rs761217728 Chr.14: 34536113-34536114 on Build GRCh38
rs915031597 Chr.14: 34536350-34536350 on Build GRCh38
rs946602287 Chr.14: 34536015-34536015 on Build GRCh38
rs372745064 Chr.14: 34536282-34536282 on Build GRCh38
rs558498853 Chr.14: 34536394-34536394 on Build GRCh38
rs545037407 Chr.14: 34536387-34536387 on Build GRCh38
rs745631390 Chr.14: 34536045-34536045 on Build GRCh38
rs760920658 Chr.14: 34536001-34536001 on Build GRCh38
rs756786792 Chr.14: 34536141-34536141 on Build GRCh38
rs779767989 Chr.14: 34536258-34536258 on Build GRCh38
rs922323959 Chr.14: 34536040-34536040 on Build GRCh38
rs775855553 Chr.14: 34535992-34535992 on Build GRCh38
rs756679003 Chr.14: 34536135-34536135 on Build GRCh38
rs767817127 Chr.14: 34536125-34536125 on Build GRCh38
rs535088063 Chr.14: 34536051-34536051 on Build GRCh38
rs761042244 Chr.14: 34536214-34536214 on Build GRCh38
rs778260137 Chr.14: 34536137-34536137 on Build GRCh38
rs1021285310 Chr.14: 34536240-34536240 on Build GRCh38
rs534860351 Chr.14: 34536407-34536409 on Build GRCh38
rs768655782 Chr.14: 34536185-34536185 on Build GRCh38
rs974538139 Chr.14: 34536319-34536319 on Build GRCh38
rs779772751 Chr.14: 34536169-34536169 on Build GRCh38
rs745547243 Chr.14: 34536144-34536144 on Build GRCh38
rs760988801 Chr.14: 34536307-34536307 on Build GRCh38
rs546267694 Chr.14: 34536268-34536268 on Build GRCh38
rs953906827 Chr.14: 34536334-34536334 on Build GRCh38
rs566054960 Chr.14: 34536020-34536020 on Build GRCh38
rs773357203 Chr.14: 34536097-34536097 on Build GRCh38
rs201467477 Chr.14: 34536142-34536142 on Build GRCh38
rs202029009 Chr.14: 34536140-34536140 on Build GRCh38
rs941014170 Chr.14: 34536353-34536353 on Build GRCh38
rs200558084 Chr.14: 34536199-34536199 on Build GRCh38
rs557668954 Chr.14: 34536205-34536205 on Build GRCh38
rs750080859 Chr.14: 34536237-34536237 on Build GRCh38
rs758055434 Chr.14: 34536243-34536243 on Build GRCh38
rs764482656 Chr.14: 34536317-34536317 on Build GRCh38
rs923894973 Chr.14: 34536389-34536389 on Build GRCh38
rs771170644 Chr.14: 34536194-34536194 on Build GRCh38
rs985155749 Chr.14: 34536338-34536338 on Build GRCh38
rs776731287 Chr.14: 34536188-34536188 on Build GRCh38
rs751761180 Chr.14: 34536119-34536119 on Build GRCh38
rs765064665 Chr.14: 34536200-34536200 on Build GRCh38
rs766658190 Chr.14: 34536108-34536108 on Build GRCh38
rs781240330 Chr.14: 34536276-34536276 on Build GRCh38
rs772130011 Chr.14: 34536293-34536293 on Build GRCh38
rs774359363 Chr.14: 34536285-34536285 on Build GRCh38
rs754231049 Chr.14: 34536313-34536313 on Build GRCh38
rs756733657 Chr.14: 34536232-34536232 on Build GRCh38
rs759733040 Chr.14: 34536201-34536201 on Build GRCh38
rs763086927 Chr.14: 34536191-34536191 on Build GRCh38
rs778340198 Chr.14: 34536233-34536233 on Build GRCh38
rs975011172 Chr.14: 34535982-34535982 on Build GRCh38
rs572050715 Chr.14: 34536399-34536399 on Build GRCh38
rs35580813 Chr.14: 34536330-34536331 on Build GRCh38
rs764537607 Chr.14: 34536225-34536225 on Build GRCh38
rs746822873 Chr.14: 34536178-34536178 on Build GRCh38
rs200733425 Chr.14: 34536132-34536132 on Build GRCh38
rs753187237 Chr.14: 34536065-34536066 on Build GRCh38
rs895832721 Chr.14: 34536404-34536404 on Build GRCh38
rs748144937 Chr.14: 34536279-34536279 on Build GRCh38
rs202040456 Chr.14: 34536210-34536210 on Build GRCh38
rs764409095 Chr.14: 34536127-34536127 on Build GRCh38
rs754847724 Chr.14: 34536275-34536275 on Build GRCh38
rs1023529879 Chr.14: 34536181-34536181 on Build GRCh38
rs775805233 Chr.14: 34536302-34536302 on Build GRCh38
rs202103758 Chr.14: 34536074-34536074 on Build GRCh38
rs769983811 Chr.14: 34536091-34536091 on Build GRCh38

View Full Product Details