Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73498704-73499203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT1
Entrez Gene ID
641371
Gene Name
acyl-CoA thioesterase 1
Gene Aliases
ACH2, CTE-1, LACH2
UniGene
Hs.446685
Gene Chromosome Location
Chr.14: 73493021-73543799 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1707572 c.2330G>A p.W777* Chr.14: 73499097-73499097 on Build GRCh38
135655 c.2146G>A p.V716M Chr.14: 73499140-73499140 on Build GRCh38
5015250 c.2155T>A p.C719S Chr.14: 73499131-73499131 on Build GRCh38
3706381 c.2204T>C p.F735S Chr.14: 73499082-73499082 on Build GRCh38
5015249 c.2296T>A p.C766S Chr.14: 73499131-73499131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547279159 Chr.14: 73498753-73498753 on Build GRCh38
rs766813563 Chr.14: 73499064-73499064 on Build GRCh38
rs1014087 Chr.14: 73498870-73498870 on Build GRCh38
rs778326392 Chr.14: 73499132-73499132 on Build GRCh38
rs754044635 Chr.14: 73499103-73499103 on Build GRCh38
rs184273633 Chr.14: 73499109-73499109 on Build GRCh38
rs114232158 Chr.14: 73499074-73499074 on Build GRCh38
rs149937111 Chr.14: 73498875-73498875 on Build GRCh38
rs750514189 Chr.14: 73499080-73499080 on Build GRCh38
rs565598534 Chr.14: 73498761-73498761 on Build GRCh38
rs564488568 Chr.14: 73499078-73499078 on Build GRCh38
rs900211144 Chr.14: 73499077-73499077 on Build GRCh38
rs543489401 Chr.14: 73499127-73499127 on Build GRCh38
rs766774208 Chr.14: 73499092-73499092 on Build GRCh38
rs1019346795 Chr.14: 73498938-73498938 on Build GRCh38
rs957931620 Chr.14: 73499019-73499019 on Build GRCh38
rs758869003 Chr.14: 73499130-73499130 on Build GRCh38
rs59763915 Chr.14: 73498871-73498871 on Build GRCh38
rs926198864 Chr.14: 73498978-73498978 on Build GRCh38
rs756248510 Chr.14: 73499085-73499085 on Build GRCh38
rs368350559 Chr.14: 73499172-73499172 on Build GRCh38
rs996242309 Chr.14: 73499105-73499105 on Build GRCh38
rs759016309 Chr.14: 73499033-73499033 on Build GRCh38
rs961469299 Chr.14: 73498799-73498799 on Build GRCh38
rs925106156 Chr.14: 73499006-73499006 on Build GRCh38
rs555426913 Chr.14: 73498932-73498932 on Build GRCh38
rs1007917637 Chr.14: 73498891-73498891 on Build GRCh38
rs530901532 Chr.14: 73499115-73499115 on Build GRCh38
rs762524136 Chr.14: 73499106-73499107 on Build GRCh38
rs913377701 Chr.14: 73499020-73499020 on Build GRCh38
rs554518069 Chr.14: 73498991-73498991 on Build GRCh38
rs374170978 Chr.14: 73499143-73499143 on Build GRCh38
rs7142466 Chr.14: 73498974-73498974 on Build GRCh38
rs944482563 Chr.14: 73499038-73499038 on Build GRCh38
rs780748065 Chr.14: 73498859-73498863 on Build GRCh38
rs374354466 Chr.14: 73499047-73499047 on Build GRCh38
rs573674426 Chr.14: 73498942-73498942 on Build GRCh38
rs905297991 Chr.14: 73498727-73498727 on Build GRCh38
rs572606057 Chr.14: 73499017-73499017 on Build GRCh38
rs775131632 Chr.14: 73499173-73499173 on Build GRCh38
rs775113615 Chr.14: 73499052-73499052 on Build GRCh38
rs539622875 Chr.14: 73498838-73498838 on Build GRCh38
rs936230952 Chr.14: 73499008-73499008 on Build GRCh38
rs751535322 Chr.14: 73499073-73499073 on Build GRCh38
rs372763948 Chr.14: 73499058-73499058 on Build GRCh38
rs755408967 Chr.14: 73499107-73499107 on Build GRCh38
rs536730338 Chr.14: 73498927-73498927 on Build GRCh38

View Full Product Details