Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77799944-77800455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK1
Entrez Gene ID
57143
Gene Name
aarF domain containing kinase 1
Gene Aliases
-
UniGene
Hs.413208
Gene Chromosome Location
Chr.14: 77800058-77933954 on Build GRCh38
Transcripts
NM_001142545.1 NM_020421.3 XM_005267885.3 XM_005267886.3 XM_011536987.2 XM_011536988.2 XM_017021474.1 XM_017021475.1 XM_017021476.1

SNP Detail

SNP ID Chromosome Location
rs560181943 Chr.14: 77800044-77800044 on Build GRCh38
rs978860109 Chr.14: 77800194-77800194 on Build GRCh38
rs993263017 Chr.14: 77800070-77800070 on Build GRCh38
rs993503538 Chr.14: 77800425-77800425 on Build GRCh38
rs552569327 Chr.14: 77800393-77800393 on Build GRCh38
rs557819366 Chr.14: 77800042-77800042 on Build GRCh38
rs28699349 Chr.14: 77800293-77800293 on Build GRCh38
rs887597582 Chr.14: 77800377-77800377 on Build GRCh38
rs1009006627 Chr.14: 77800000-77800000 on Build GRCh38
rs757778900 Chr.14: 77800281-77800281 on Build GRCh38
rs1049952022 Chr.14: 77800237-77800237 on Build GRCh38
rs1030398689 Chr.14: 77800138-77800138 on Build GRCh38
rs115018137 Chr.14: 77800264-77800264 on Build GRCh38
rs980666726 Chr.14: 77800142-77800142 on Build GRCh38
rs1006026547 Chr.14: 77799992-77799992 on Build GRCh38
rs572993129 Chr.14: 77800295-77800295 on Build GRCh38
rs566948487 Chr.14: 77800306-77800306 on Build GRCh38
rs890133667 Chr.14: 77800298-77800298 on Build GRCh38
rs1027453982 Chr.14: 77800072-77800072 on Build GRCh38
rs945421220 Chr.14: 77800191-77800191 on Build GRCh38
rs1019994029 Chr.14: 77800365-77800365 on Build GRCh38
rs925430649 Chr.14: 77800206-77800206 on Build GRCh38
rs28549360 Chr.14: 77799999-77799999 on Build GRCh38
rs1027430883 Chr.14: 77800308-77800308 on Build GRCh38
rs926556444 Chr.14: 77800152-77800152 on Build GRCh38
rs545374094 Chr.14: 77800239-77800239 on Build GRCh38
rs527478724 Chr.14: 77800303-77800303 on Build GRCh38
rs960732331 Chr.14: 77800159-77800159 on Build GRCh38
rs28461444 Chr.14: 77800254-77800254 on Build GRCh38
rs866980977 Chr.14: 77800240-77800240 on Build GRCh38
rs978985415 Chr.14: 77800393-77800393 on Build GRCh38
rs951878606 Chr.14: 77800105-77800105 on Build GRCh38
rs900446014 Chr.14: 77800274-77800274 on Build GRCh38
rs577630516 Chr.14: 77800167-77800167 on Build GRCh38
rs979501094 Chr.14: 77800192-77800192 on Build GRCh38
rs539156244 Chr.14: 77800349-77800349 on Build GRCh38
rs886289179 Chr.14: 77799989-77799989 on Build GRCh38
rs1033593473 Chr.14: 77800395-77800395 on Build GRCh38
rs937581635 Chr.14: 77800221-77800221 on Build GRCh38
rs929935861 Chr.14: 77800243-77800243 on Build GRCh38
rs547316642 Chr.14: 77800305-77800305 on Build GRCh38
rs12880840 Chr.14: 77800248-77800248 on Build GRCh38
rs1037932491 Chr.14: 77800271-77800271 on Build GRCh38
rs913908750 Chr.14: 77800180-77800180 on Build GRCh38
rs927606450 Chr.14: 77800219-77800219 on Build GRCh38
rs114721104 Chr.14: 77800223-77800223 on Build GRCh38
rs1019963076 Chr.14: 77800170-77800170 on Build GRCh38
rs1018689299 Chr.14: 77800018-77800018 on Build GRCh38
rs959008856 Chr.14: 77800414-77800414 on Build GRCh38

View Full Product Details